Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstituted by adding 1.0 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20°C.
Cross-reactivities against enzymes of other sources may occur but have not been determined.
Prior to use, an aliquot is thawed slowly at ambient temperature, spun down again and used to prepare working dilutions by adding sterile phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. Working dilutions should be stored at 4°C, not refrozen, and preferably used the same day. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance of the product.
Hexokinases phosphorylate glucose to produce glucose-6-phosphate, the first step in most glucose metabolism pathways. This gene encodes a ubiquitous form of hexokinase which localizes to the outer membrane of mitochondria. Mutations in this gene have been associated with hemolytic anemia due to hexokinase deficiency. Alternative splicing of this gene results in five transcript variants which encode different isoforms, some of which are tissue-specific. Each isoform has a distinct N-terminus; the remainder of the protein is identical among all the isoforms. A sixth transcript variant has been described, but due to the presence of several stop codons, it is not thought to encode a protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hexokinase PI; Hexokinase-1; Hexokinase-A; HK1-ta; HK1-tb; HK1-tc; HKI
Gene Aliases: HKA; HXK1; YFR053C
UniProt ID: (Baker's yeast) P04806
Entrez Gene ID: (Baker's yeast) 850614
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support