Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Histones H1 are necessary for the condensation of nucleosome chains into higher-order structures.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H1 histone family, member X; Histone H1.10; Histone H1x
Gene Aliases: H1-10; H1.10; H1FX; H1X
UniProt ID: (Human) Q92522
Entrez Gene ID: (Human) 8971
Molecular Function: histone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support