Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Hrk, a pro-apoptotic member of the Bcl-2 homology domain-3 (BH3)-only group of the Bcl-2 family of proteins, was also identified as novel protein induced during programmed neuronal death. It lacks significant homology to other Bcl-2 family members except for an 8-amino acid region that is similar to the BH3 motif of Bik. Hrk regulates apoptosis through interaction with the anti-apoptotic proteins Bcl-2 and Bcl-XL via this domain. It does not interact with the pro-apoptotic proteins Bax, Bak, or Bcl-XS. Hrk localizes to mitochondrial membranes in a pattern similar to that previously reported for Bcl-2 and Bcl-XL.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activator of apoptosis harakiri; activator of apoptosis Hrk; BCL2-interacting protein; BH3 interacting (with BCL2 family) domain, apoptosis agonist; BH3-interacting domain-containing protein 3; death protein 5; harakiri, BCL2 interacting protein (contains only BH3 domain); Neuronal death protein DP5
Gene Aliases: AI838259; BID3; DP5; HARAKIRI; HRK
UniProt ID: (Human) O00198, (Rat) P62817, (Mouse) P62816
Entrez Gene ID: (Human) 8739, (Rat) 117271, (Mouse) 12123
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support