Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a lysosomal hyaluronidase. Hyaluronidases intracellularly degrade hyaluronan, one of the major glycosaminoglycans of the extracellular matrix. Hyaluronan is thought to be involved in cell proliferation, migration and differentiation. This enzyme is active at an acidic pH and is the major hyaluronidase in plasma. Mutations in this gene are associated with mucopolysaccharidosis type IX, or hyaluronidase deficiency. The gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hyal-1; hyaluronidase 1; Hyaluronidase-1; Hyaluronoglucosaminidase-1; LuCa-1; Lung carcinoma protein 1; plasma hyaluronidase; tumor suppressor LUCA-1
Gene Aliases: Hya1; HYAL-1; HYAL1; LUCA1; MPS9; NAT6
UniProt ID: (Human) Q12794, (Rat) Q76HN1, (Mouse) Q91ZJ9
Entrez Gene ID: (Human) 3373, (Rat) 367166, (Mouse) 15586
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support