Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Positive Samples: RAW264.7, THP-1, Mouse testis, Mouse spleen, Rat lung, Rat testis, Rat spleen
Immunogen sequence: MSQSRDLQGG KAFGLLKAQQ EERLEGINKQ FLDDPKYSND EDLPSKLEAF KVKYMEFDLN GNGDIDIMSL KRMLEKLGVP KTHLELKRLI REVSSGSEET
AIF1 is induced by cytokines and interferon. It is thought to be involved in the negative regulation of growth of vascular smooth muscle cells, which contributes to the anti-inflammatory response to vessel wall trauma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AIF-1; Allograft inflammatory factor 1; balloon angioplasty responsive transcript; balloon angioplasty-responsive transcript 1; DADB-70P7.8; DASS-82G15.2; Em:AF129756.17; Iba; interferon gamma responsive transcript; ionized calcium binding adapter molecule 1; Ionized calcium-binding adapter molecule 1; Microglia response factor; MRF-1; protein BART-1; Protein G1; testis specific
Gene Aliases: AI607846; AIF-1; AIF1; BART-1; Bart1; D17H6S50E; G1; IBA1; IRT-1; IRT1; mrf-1; Mrf1
UniProt ID: (Human) P55008, (Mouse) O70200, (Rat) P55009
Entrez Gene ID: (Human) 199, (Mouse) 11629, (Rat) 29427
Molecular Function:
calmodulin-related
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support