Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: GSNFNHVVQN EKSDTHTLVT SGVYAWFRHP SYVGWFYWSI GTQVMLCNPI
Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 79%
This gene encodes the third of three enzymes that post translationally modify isoprenylated C-terminal cysteine residues in certain proteins and target those proteins to the cell membrane. This enzyme localizes to the endoplasmic reticulum. Alternative splicing may result in other transcript variants, but the biological validity of those transcripts has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Isoprenylcysteine carboxylmethyltransferase; MGC39955; pcCMT; PPMT; Prenylated protein carboxyl methyltransferase; Prenylcysteine carboxyl methyltransferase; protein-S isoprenylcysteine O-methyltransferase; Protein-S-isoprenylcysteine O-methyltransferase
Gene Aliases: HSTE14; ICMT; MST098; MSTP098; PCCMT; PCMT; PPMT
UniProt ID: (Human) O60725
Entrez Gene ID: (Human) 23463
Molecular Function:
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support