Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MKAFSPVRSV RKNSLSDHSL GISRSKTPVD DPMSLLYNMN DCYSKLKELV PSIPQNKKVS KMEILQHVID YILDLQIALD SHPTIVSLHH QRPGQNQASR TPLTTLNTDI SILSLQASEF PSELMSNDSK ALCG
The inhibitor of DNA binding 2 (ID2) protein is essential for promoting the development of a number of cell types including natural killer cell, erythroid precursors, as well as, CD8+ and CD4+ T-cell. A lack of ID2 promotes B-cell development. The chief role of this protein is to associate with ubiquitously-expressed E proteins, and other proteins of the basic helix-loop-helix (HLH) family, preventing them from binding to DNA and activating transcription. Activation of ID2 is critical for the development of the natural killer cell lineage.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHb26; cell growth-inhibiting gene 8; Class B basic helix-loop-helix protein 26; DNA-binding protein inhibitor ID-2; DNA-binding protein inhibitor ID2; helix-loop-helix protein ID2; Inhibitor of differentiation 2; Inhibitor of DNA binding 2; inhibitor of DNA binding 2, dominant negative helix-loop-helix protein; MGC26389; OTTHUMP00000140258; OTTHUMP00000200297
Gene Aliases: BHLHB26; GIG8; ID2; ID2A; ID2H
UniProt ID: (Human) Q02363
Entrez Gene ID: (Human) 3398
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support