Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 8
Immunogen sequence: TGGVQTVTLI PGDGIGPEIS AAVMKIFDAA KAPIQWEERN VTAIQGPGGK WMIPSEAKES MDKNKMGLKG PLKTPIAAGH PSMNLLLRKT FDLYANVRPC VSIEGYKTPY TDVNIVTIRE NTEGEYSGIE HVIVDGVVQS IKLITEGASK RIAEFAFEYA RNNHRSNVTA VHKANIMRMS DGLFLQKCRE VAESCKDIKF NEMYLDTVCL NMVQDPSQFD VLVMPNLYGD ILSDLCAGLI GGLGVTPSGN IGANGVAIFE SVHGTAPDIA GKDMANPTAL LLSAVMMLRH MGLFDHAARI EAACFATIKD GKSLTKDLGG NAKCSDFTEE ICRRVKDLD; Positive Samples: Raji, NIH/3T3, Mouse brain, Mouse heart, Mouse kidney; Cellular Location: Mitochondrion
Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step of the tricarboxylic acid cycle. Each isozyme is a heterotetramer that is composed of two alpha subunits, one beta subunit, and one gamma subunit. The protein described here is the alpha subunit of one isozyme of NAD(+)-dependent isocitrate dehydrogenase.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H-IDH alpha; isocitrate dehydrogenase (NAD+) alpha chain; isocitrate dehydrogenase 3 (NAD+) alpha; Isocitrate dehydrogenase [NAD] subunit alpha, mitochondrial; Isocitric dehydrogenase subunit alpha; NAD(+)-specific ICDH subunit alpha; NAD(H)-specific isocitrate dehydrogenase alpha subunit; NAD+-specific ICDH
Gene Aliases: 1110003P10Rik; 1500012E04Rik; AA407078; AI316514; IDH3A
UniProt ID: (Human) P50213, (Mouse) Q9D6R2, (Rat) Q99NA5
Entrez Gene ID: (Human) 3419, (Mouse) 67834, (Rat) 114096
Molecular Function:
dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support