Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MAALSGVRWL TRALVSAGNP GAWRGLSTSA AAHAASRSQA EDVRVEGSFP VTMLPGDGVG PELMHAVKEV FKAAAVPVEF QEHHLSEVQN MASEEKLEQV LSSMKENKVA IIGKIHTPME YKGELASYDM RLRRKLDLFA NVVHVKSLPG YMTRHNNLDL VIIREQTEGE YSSLEHESAR GVIECLKIVT RAKSQRIAKF AFDYATKKGR GKVTAVHKAN IMKLGDGLFL QCCEEVAELY PKIKFETMII DNCCMQLVQN PYQFDVLVMP NLYGNIIDNL AAGLVGGAGV VPGESYSAEY AVFETGARHP FAQAVGRNIA NPTAMLLSAS NMLRHLNLEY HSSMIADAVK KVIKVGKVRT RDMGGYSTTT DFIKSVIGHL QTKGS
Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step of the tricarboxylic acid cycle. Each isozyme is a heterotetramer that is composed of two alpha subunits, one beta subunit, and one gamma subunit. The protein encoded by this gene is the beta subunit of one isozyme of NAD(+)-dependent isocitrate dehydrogenase. Three alternatively spliced transcript variants encoding different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DH3B; H-IDHB; H-IDHG; isocitrate dehydrogenase (NAD(+)) 3 beta; isocitrate dehydrogenase 3 (NAD(+)) beta; isocitrate dehydrogenase 3 (NAD+) beta; isocitrate dehydrogenase 3 beta; isocitrate dehydrogenase 3, beta subunit; Isocitrate dehydrogenase [NAD] subunit beta, mitochondrial; Isocitric dehydrogenase subunit beta; mitochondrial NAD+isocitrate dehydrogenase 3 beta variant 1; mitochondrial NAD+isocitrate dehydrogenase 3 beta variant 2; N14A tumor-related protein; NAD(+)-specific ICDH subunit beta; NAD+-specific isocitrate dehydrogenase b subunit
Gene Aliases: C78231; IDH3B; RP46
UniProt ID: (Human) O43837, (Rat) Q68FX0
Entrez Gene ID: (Rabbit) 100342113, (Human) 3420, (Pig) 733641, (Rat) 94173, (Mouse) 170718
Molecular Function:
dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support