Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MALKVATVA GSAAKAVLGP ALLCRPWEVL GAHEVPSRNI FSEQTIPPSA KYGGRHTVTM IPGDGIGPEL MLHVKSVFRH ACVPVDFEEV HVSSNADEED IRNAIMAIRR NRVALKGNIE TNHNLPPSHK SRNNILRTSL DLYANVIHCK SLPGVVTRHK DIDILIVREN TEGEYSSLEH ESVAGVVESL KIITKAKSLR IAEYAFKLAQ ESGRKKVTAV HKANIMKLGD GLFLQCCREV AARYPQITFE NMIVDNTTMQ LVSRPQQFDV MVMPNLYGNI VNNVCAGLVG GPGLVAGANY GHVYAVFETA TRNTGKSIAN KNIANPTATL LASCMMLDHL KLHSYATSIR KAVLASMDNE NMHTPDIGGQ GTTSEAIQDV IRHIRVINGR AVEA (1-393 aa encoded by BC000933)
Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step of the tricarboxylic acid cycle. Each isozyme is a heterotetramer that is composed of two alpha subunits, one beta subunit, and one gamma subunit. The protein encoded by this gene is the gamma subunit of one isozyme of NAD(+)-dependent isocitrate dehydrogenase. This gene is a candidate gene for periventricular heterotopia. Several alternatively spliced transcript variants of this gene have been described, but only some of their full length natures have been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IDH-gamma; iso; isocitrate dehydrogenase 3 (NAD+) gamma; isocitrate dehydrogenase 3 gamma; Isocitrate dehydrogenase [NAD] subunit gamma 1, mitochondrial; Isocitrate dehydrogenase [NAD] subunit gamma, mitochondrial; isocitrate dehydrogenase, NAD(+)-specific, mitochondrial, gamma subunit; Isocitric dehydrogenase subunit gamma; NAD (H)-specific isocitrate dehydrogenase gamma subunit; NAD(+)-specific ICDH subunit gamma; NAD+-specific ICDH
Gene Aliases: H-IDHG; IDH3G
UniProt ID: (Human) P51553, (Mouse) P70404
Entrez Gene ID: (Human) 3421, (Mouse) 15929
Molecular Function:
dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support