Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAHAMENSW TISKEYHIDE EVGFALPNPQ ENLPDFYNDW MFIAKHLPDL IESGQLRERV EKLNMLSIDH LTDHKSQRLA RLVLGCITMA YVWGKGHGDV RKVLPRNIAV PYCQLSKKLE LPPILVYADC VLANWKKKDP NKPLTYENMD VLFSFRDGDC SKGFFLVSLL VEIAAASAIK VIPTVFKAMQ MQERDTLLKA LLEIASCLEK ALQVFHQIHD HVNPKAFFSV LRIYLSGWKG NPQLSDGLVY EGFWEDPKEF AGGSAGQSSV FQCFDVLLGI QQTAGGGHAA QFLQDMRRYM PPAHRNFLCS LESNPSVREF VLSKGDAGLR EAYDACVKAL VSLRSYHLQI VTKYILIPAS QQPKENKTSE DPSKLEAKGT GGTDLMNFLK TVRSTTEKSL LKEG (1-403 aa encoded by BC027882)
IDO1 is an intracellular heme-containing enzyme that catalyzes the oxidative cleavage of the indole ring of several important regulatory molecules like tryptophan, serotonin, and melatonin. By doing this, IDO1 initiates the production of biologically active metabolites, commonly referred to as kynurenines. IDO1 is widely expressed in a variety of human tissues as well as in macrophages and dendritic cells (DCs). In inflammation, interferons (IFNs) act on specific receptors to trigger IDO1 induction. The production of IFN-gamma and induction of IDO1 represent important antimicrobial mechanisms. Degradation and depletion of tryptophan by IDO1 inhibits the growth of viruses, bacteria and parasites. Furthermore, IDO1 plays a complex and crucial role in immunoregulation during infection, pregnancy, autoimmunity, transplantation, and neoplasia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 1.13.11.52; IDO-1; indolamine 2,3 dioxygenase; indole 2,3-dioxygenase; indoleamine; Indoleamine 2,3-dioxygenase 1; indoleamine 23-dioxygenase; Indoleamine-2; indoleamine-pyrrole 2,3 dioxygenase; Indoleamine-pyrrole 2,3-dioxygenase
Gene Aliases: IDO; IDO-1; IDO1; INDO
UniProt ID: (Human) P14902, (Mouse) P28776, (Rat) Q9ERD9
Entrez Gene ID: (Human) 3620, (Mouse) 15930, (Rat) 66029
Molecular Function:
oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support