Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YASDDTRAHP NRSVAVTLRL RGVPPGPGLV YVTRYLDNGL CSPDGEWRRL GRPVFPTAEQ FRRMRAAEDP VAAAPRPLPA GGRLTLRPAL RLPSLLLVHV CARPEKPPGQ VTRLR
IDUA encodes an enzyme that hydrolyzes the terminal alpha-L-iduronic acid residues of two glycosaminoglycans, dermatan sulfate and heparan sulfate. This hydrolysis is required for the lysosomal degradation of these glycosaminoglycans.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-L-iduronidase
Gene Aliases: 6030426D08; IDA; IDUA; MPS1
UniProt ID: (Human) P35475
Entrez Gene ID: (Human) 3425, (Mouse) 15932
Molecular Function:
glycosidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support