Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
IEX-1 is a stress inducible gene that is induced by ionizing radiation, ultraviolet radiation, and a variety of growth factors, i.e., FAS and TNF-a. IEX-1 is widely expressed in epithelial and endocrine tissues, as well as in vascular endothelium. It plays an important role in the regulation of cellular growth, cell death and oncogenesis. IEX-1 is precisely regulated by multiple transcription factors such as p53, NF-kappa-B/rel, Sp1 and c-Myc, to ensure rapid and transient expression of IEX-1 in cells under a variety of stress conditions. IEX-1 is expressed as both a longer form (IEX1L) and a splice variant, designated IEX1S. It is localized to the nucleus and perinuclear region. Overexpression of IEX-1 facilitates apoptosis and cell cycle progression, whereas disruption of IEX-1 expression is associated with decreases in both apoptosis and cell cycle progression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: anti-death protein; cAMP inducible gene 3; DADB-118P11.5; Differentiation-dependent gene 2 protein; expressed in pancreatic carcinoma; gly96, mouse, homolog of; Immediate early protein GLY96; Immediate early response 3 protein; immediately early gene X-1; PACAP-responsive gene 1 protein; Protein DIF-2; Protein PRG1; Radiation-inducible immediate-early gene IEX-1
Gene Aliases: AI663993; cI-3; DIF-2; DIF2; GLY96; IER3; IEX-1; IEX-1L; IEX1; Ler3; PRG1
UniProt ID: (Human) P46695, (Mouse) P46694
Entrez Gene ID: (Human) 8870, (Rat) 294235, (Mouse) 15937
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support