Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VCRNGFLEVY PFTLVADVNA DRNMEIPKGL IRSASVTPKI NQLCSQTKGS FVNGVFEVHK KNVRGEFTYY EIQDNTGKME VVVHGRLTTI NCEEGDKLKL TCFELAPKSG NTGELRSVIH SHIKVIKTRK NKKDILNPDS SMETSPDFFF
IFI16 (gamma-inteferon-inducible protein 16) binds double-stranded DNA. It binds preferentially to supercoiled DNA and cruciform DNA structures. It seems to be involved in transcriptional regulation and as a transcriptional repressor. IFI16 may have a role in the regulation of hematopoietic differentiation through activation of unknown target genes. It controls cellular proliferation by modulating the functions of cell cycle regulatory factors including p53/TP53 and the retinoblastoma protein. IFI16 is also involved in innate immune response by recognizing viral dsDNA in the cytosol and nucleus. After binding to viral DNA in the cytoplasm, IFI16 recruits TMEM173/STING and mediates the induction of IFN-beta. It is necessary to activate the IFR3 signaling cascade suring human herpes simplex virus 1 infection and promotes the assembly of heterochromatin on herpesviral DNA and inhibiton of viral immediate-early gene expression and replication. IFI16 is also involved in the MTA1-mediated epigenetic regulation of ESR1 expression in breast cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gamma-interferon-inducible protein 16; IFI 16C; Ifi-16; interferon, gamma-inducible protein 16; interferon-gamma induced protein IFI 16; Interferon-inducible myeloid differentiation transcriptional activator
Gene Aliases: IFI16; IFNGIP1; PYHIN2
UniProt ID: (Human) H3BLV7
Entrez Gene ID: (Human) 3428
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support