Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MEASALTSSA VTSVAKVVRV ASGSAVVLPL ARIATVVIGG VVAVPMVLSA MGFTAAGIAS SSIAAKMMSA AAIANGGGVA SGSLVATLQS LGATGLSGLT KFILGSIGSA IAAVIARFY
Promotes cell death. Mediates IFN-induced apoptosis characterized by a rapid and robust release of cytochrome C from the mitochondria and activation of BAX and caspases 2, 3, 6, 8 and 9.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2310061N23Rik; Interferon alpha-induced 11.5 kDa protein; Interferon alpha-inducible protein 27, mitochondrial; interferon, alpha-inducible protein 27; Interferon-stimulated gene 12a protein; ISG12(a); p27
Gene Aliases: FAM14D; IFI27; ISG12; ISG12A; P27
UniProt ID: (Human) P40305
Entrez Gene ID: (Human) 3429
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support