Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSAPLDAALH ALQEEQARLK MRLWDLQQLR KELGDSPKDK VPFSVPKIPL VFRGHTQQDP EVPKSLVSNL RIHCPLLAGS ALITFDDPKV AEQVLQQKEH TINMEECRLR VQVQPLELPM VTTIQVMVSS QLSGRRVLVT GFPASLRLSE EELLDKLEIF FGKTRNGGGD VDVRELLPGS VMLGFARDGV AQRLCQIGQF TVPLGGQQVP LRVSPYVNGE IQKAEIRSQP VPRSVLVLNI PDILDGPELH DVLEIHFQKP TRGGGEVEAL TVVPQGQQGL AVFTSESG
Not yet known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ifi-35; IFP 35; Interferon-induced 35 kDa protein; interferon-induced protein 35
Gene Aliases: IFI35; IFP35
UniProt ID: (Human) P80217
Entrez Gene ID: (Human) 3430
Molecular Function:
transcription cofactor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support