Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSEVTKNSLE KILPQLKCHF TWNLFKEDSV SRDLEDRVCN QIEFLNTEFK ATMYNLLAYI KHLDGNNEAA LECLRQAEEL IQQEHADQAE IRSLVTWGNY AWVYYHLGRL SDAQIYVDKV KQTCKKFSNP YSIEYSELDC EEGWTQLKCG RNERAKVCFE KALEEKPNNP EFSSGLAIAM YHLDNHPEKQ FSTDVLKQAI ELSPDNQYVK VLLGLKLQKM NKEAEGEQFV EEALEKSPCQ TDVLRSAAKF YRRKGDLDKA IELFQRVLES TPNNGYLYHQ IGCCYKAKVR QMQNTGESEA SGNKEMIEAL KQYAMDYSNK ALEKGLNPLN AYSDLAEFLE TECYQTPFNK EVPDAEKQQS HQRYCNLQKY NGKSEDTAVQ HGLEGLSISK KSTDKEEIKD QPQNVSENLL PQNAPNYWYL QGLIHKQNGD LLQAAKCYEK ELGRLLRDAP SGIGSIFLSA SELEDGSEEM GQGAVSSSPR ELLSNSEQLN
IFN-induced antiviral protein which acts as an inhibitor of cellular as well as viral processes, cell migration, proliferation, signaling, and viral replication. Enhances MAVS-mediated host antiviral responses by serving as an adapter bridging TBK1 to MAVS which leads to the activation of TBK1 and phosphorylation of IRF3 and phosphorylated IRF3 translocates into nucleus to promote antiviral gene transcription. Exhibits an antiproliferative activity via the up-regulation of cell cycle negative regulators CDKN1A/p21 and CDKN1B/p27. Normally, CDKN1B/p27 turnover is regulated by COPS5, which binds CDKN1B/p27 in the nucleus and exports it to the cytoplasm for ubiquitin-dependent degradation. IFIT3 sequesters COPS5 in the cytoplasm, thereby increasing nuclear CDKN1B/p27 protein levels. Upregulates CDKN1A/p21 by downregulating MYC, a repressor of CDKN1A/p21. Can negatively regulate the apoptotic effects of IFIT2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CIG49; IFI-60K; IFIT-3; IFIT-4; Interferon-induced 60 kDa protein; Interferon-induced protein with tetratricopeptide repeats 3; Interferon-induced protein with tetratricopeptide repeats 4; ISG-60; P60; Retinoic acid-induced gene G protein; RP11-149I23.4
Gene Aliases: CIG-49; cig41; GARG-49; IFI60; IFIT3; IFIT4; IRG2; ISG60; P60; RIG-G
UniProt ID: (Human) O14879
Entrez Gene ID: (Human) 3437
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support