Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSEIRKDTLK AILLELECHF TWNLLKEDID LFEVEDTIGQ QLEFLTTKSR LALYNLLAYV KHLKGQNKDA LECLEQAEEI IQQEHSDKEE VRSLVTWGNY AWVYYHMDQL EEAQKYTGKI GNVCKKLSSP SNYKLECPET DCEKGWALLK FGGKYYQKAK AAFEKALEVE PDNPEFNIGY AITVYRLDDS DREGSVKSFS LGPLRKAVTL NPDNSYIKVF LALKLQDVHA EAEGEKYIEE ILDQISSQPY VLRYAAKFYR RKNSWNKALE LLKKALEVTP TSSFLHHQMG LCYRAQMIQI KKATHNRPKG KDKLKVDELI SSAIFHFKAA MERDSMFAFA YTDLANMYAE GGQYSNAEDI FRKALRLENI TDDHKHQIHY HYGRFQEFHR KSENTAIHHY LEALKVKDRS PLRTKLTSAL KKLSTKRLCH NALDVQSLSA LGFVYKLEGE KRQAAEYYEK AQKIDPENAE FLTALCELRL SI
Interferon-induced RNA-binding protein involved in the human innate immune response. Has a broad and adaptable RNA structure recognition important for RNA recognition specificity in antiviral defense. Binds precursor and processed tRNAs as well as poly-U-tailed tRNA fragments (PubMed:25092312, PubMed:23317505, PubMed:23774268). Specifically binds single-stranded RNA bearing a 5'-triphosphate group (PPP-RNA), thereby acting as a sensor of viral single-stranded RNAs. Single-stranded PPP-RNAs, which lack 2'-O-methylation of the 5' cap and bear a 5'-triphosphate group instead, are specific from viruses, providing a molecular signature to distinguish between self and non-self mRNAs by the host during viral infection. Directly binds PPP-RNA in a non-sequence-specific manner (PubMed:23334420). Also recognizes and selectively binds AT-rich dsDNA (PubMed:23774268). Additionally, as a mediator in innate immunity, regulates positively IKK-NFKB signaling by sinergizing the recruitment of IKK to MAP3K7 (PubMed:26334375). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IFIT-5; Interferon-induced 58 kDa protein; Interferon-induced protein with tetratricopeptide repeats 5; P58; Retinoic acid- and interferon-inducible 58 kDa protein; retinoic acid- and interferon-inducible protein (58kD)
Gene Aliases: IFIT5; ISG58; P58; RI58
UniProt ID: (Human) Q13325
Entrez Gene ID: (Human) 24138
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support