Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
FIGURE: 1 / 1
IFN-induced antiviral protein that mediates cellular innate immunity to at least three major human pathogens, namely influenza A H1N1 virus, West Nile virus (WNV), and dengue virus (WNV), by inhibiting the early step(s) of replication.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dispanin subfamily A member 2b; Dispanin subfamily A member 2c; DSPA2b; DSPA2c; ifm2; Interferon-induced transmembrane protein 2; Interferon-induced transmembrane protein 3; Interferon-inducible protein 1-8D; Interferon-inducible protein 1-8U
Gene Aliases: 1-8D; 1-8U; DSPA2b; DSPA2c; IFITM2; IFITM3; IP15
UniProt ID: (Human) Q01629, (Human) Q01628
Entrez Gene ID: (Human) 10581, (Human) 10410
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support