Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Leinco Technologies
Specificity: Clone MAR1-5A3 recognizes an epitope on mouse IFNAR1.
Endotoxin Level: ≤ 1.0 EU/mg as determined by the LAL method. Purity: >95% by SDS Page; ≥95% monomer by analytical SEC.
Product Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using only In vitro protein free cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Storage and Handling: Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80 C. Avoid Repeated Freeze Thaw Cycles.
This antibody has been tested in the following applications: Western Blotting, Immunoprecipitation, Flow Cytometry, ELISA, Blocking.
Interferons are widely used therapeutic agents because of their anti tumor and antiviral effects and because of their modulatory effects on the immune system (Biron,2001, Kirkwood, 2002). These cytokines produce their effects by binding to the Type 1 Interferon-& Receptor (IFNAR1). Down regulation of this receptor plays a key role in determining the magnitude and duration of cytokine signaling. This down regulation is thought to be influenced by phosphorylation of Serine 535 and 539 in the IFNAR1 (Kumar et al., 2003).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H3K1; IFN R1; IFN-alpha/beta receptor 1; IFN-alpha/betaR; IFN-alpha/betaR1; IFN-alpha/betaRalpha; IFN-R-1; IFNalpha/beta R1; IFNalpha/beta-Ralpha; IFNalpha/betaR1; IFNR1; INF-a receptor; Interferon alpha/beta receptor 1; interferon-alpha/beta receptor 1; Type I interferon receptor 1
Gene Aliases: CD118; Ifar; Ifnar; Ifnar1; Ifrc; Infar
UniProt ID: (Mouse) P33896
Entrez Gene ID: (Mouse) 15975
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support