Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Predicted to react with Mouse samples.
IFRD2 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Interferon-related developmental regulator 2; interferon-related protein; Protein SKMC15
Gene Aliases: 1810034A24Rik; AA470234; AI838527; IFNRP; IFRD2; SKMc15; SM15
UniProt ID: (Human) Q12894, (Mouse) Q9D8U0
Entrez Gene ID: (Human) 7866, (Mouse) 15983
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support