Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MPRARKGNTL RKGGQRRGGG ARSSAQADSG SSDDEAASEA RSTASECPSL LSTTAEDSLG GDVVDEQGQQ EDLEEKLKEY VDCLTDKSAK TRQGALESLR LALASRLLPD FLLERRLTLA DALEKCLKKG KGEEQALAAA VLGLLCVQLG PGPKGEELFH SLQPLLVSVL SDSTASPAAR LHCASALGLG CYVAAADIQD LVSCLACLES VFSRFYGLGG SSTSPVVPAS LHGLLSAALQ AWALLLTICP STQISHILDR QLPRLPQLLS SESVNQRIAA GETIALLFEL ARDLEEEFVY EDMEALCSVL RTLATDSNKY RAKADRRRQR STFRAVLHSV EGGECEEEIV RFGFEVLYMD SWARHRIYAA FKEVLGSGMH HHLQNNELLR DIFGLGPVLL LDATALKACK VPRFEKHLYN AAAFKARTKA RSRVRDKQAD IL
IFRD2 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Interferon-related developmental regulator 2; interferon-related protein; Protein SKMC15
Gene Aliases: IFNRP; IFRD2; SKMc15; SM15
UniProt ID: (Human) Q12894
Entrez Gene ID: (Human) 7866
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support