Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MRAVLTWRD KAEHCINDIA FKPDGTQLIL AAGSRLLVYD TSDGTLLQPL KGHKDTVYCV AYAKDGLWSP EQKSVSKHKS SSKIICCSWT NDGQYLALGM FNGIISIRNK NGEEKVKIER PGGSLSPIWS ICWNPSREER NDILAVADWG QKVSFYQLSG KQIGKDRALN FDPCCISYFT KGEYILLGGS DKQVSLFTKD GVRLGTVGEQ NSWVWTCQAK PDSNYVVVGC QDGTISFYQL IFSTVHGLYK DRYAYRDSMT DVIVQHLITE QKVRIKCKEL VKKIAIYRNR LAIQLPEKIL IYELYSEDLS DMHYRVKEKI IKKFECNLLV VCANHIILCQ EKRLQCLSFS GVKEREWQME (1-359 aa encoded by BC004238)
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This cytoplasmic protein contains seven WD repeats and an AF-2 domain which function by recruiting coregulatory molecules and in transcriptional activation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: intraflagellar transport 122 homolog; Intraflagellar transport protein 122 homolog; WD repeat domain 10; WD repeat-containing protein 10; WD repeat-containing protein 140
Gene Aliases: C86139; CED; CED1; IFT122; sopb; SPG; WDR10; WDR10p; WDR140
UniProt ID: (Human) Q9HBG6, (Mouse) Q6NWV3
Entrez Gene ID: (Human) 55764, (Mouse) 81896
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support