Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: SIGDEDPFTA KLSFEQGGSE FVPVVVSRLV LRSMSRRDVL IKRWPPPLRW QYFRSLLPDA SITMCPSCFQ MFHSEDYELL VLQHGCCPYC RRCKDDPG
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This cytoplasmic protein contains seven WD repeats and an AF-2 domain which function by recruiting coregulatory molecules and in transcriptional activation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: intraflagellar transport 122 homolog; Intraflagellar transport protein 122 homolog; WD repeat domain 10; WD repeat-containing protein 10; WD repeat-containing protein 140
Gene Aliases: CED; CED1; IFT122; SPG; WDR10; WDR10p; WDR140
UniProt ID: (Human) Q9HBG6
Entrez Gene ID: (Human) 55764
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support