Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEDLLDLDE ELRYSLATSR AKMGRRAQQE SAQAENHLNG KNSSLTLTGE TSSAKLPRCR QGGWAGDSVK ASNGTQTGKQ QLDLNACYHK THHRNLGLAS LEEADIPIIP DLEEVQEEDF VLQVAAPPSI QIKRVMTYRD LDNDLMKYSA IQTLDGEIDL KLLTKVLAPE HEVREDDVGW DWDHLFTEVS SEVLTEWDPL QTEKEDPAGQ ARHT (1-213 aa encoded by BC010436)
IFT43, encoded by C14ORF179, is a subunit of the intraflagellar transport complex A (IFT-A) of primary cilia. The IFT-A is implicated in retrograde ciliary transport along axonemal microtubules. Defects in subunits of IFT-A has been associated with skeletal ciliopathies, including Sensenbrenner syndrome, Jeune syndrome, and Ellis-van-Creveld syndrome. Mutations in C14ORF179 has been linked to Sensenbrenner syndrome. (21378380).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IFT complex A subunit; intraflagellar transport 43 homolog; Intraflagellar transport protein 43 homolog
Gene Aliases: C14orf179; CED3; IFT43
UniProt ID: (Human) Q96FT9
Entrez Gene ID: (Human) 112752
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support