Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Isoform CDV-1 may be involved in cardiac hypertrophy caused by carnitine deficiency. Isoform CDV-1R appears to play a role in development of the testis and spermatogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: carnitine deficiency-associated gene expressed in ventricle 1; Carnitine deficiency-associated protein expressed in ventricle 1; CDV-1; intraflagellar transport 81 homolog; Intraflagellar transport protein 81 homolog
Gene Aliases: CDV-1; CDV-1R; CDV1; CDV1R; DV1; IFT81
UniProt ID: (Human) Q8WYA0
Entrez Gene ID: (Human) 28981, (Dog) 100856715
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support