Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
AA Sequence of recombinant protein: MAPWQCAPCS AEKLALCPPV SASCSEVTRS AGCGCCPMCA LPLGAACGVA TARCARGLSC RALPGEQQPL HALTRGQGAC VQESDASAPH AAEAGSPESP ESTEITEEEL LDNFHLMAPS EEDHSILWDA ISTYDGSKAL HVTNIKKWKE PCRIELYRVV ESLAKAQETS GEEISKFYLP NCNKNGFYHS RQCETSMDGE AGLCWCVYPW NGKRIPGSPE IRGDPNCQIY FNVQN.
Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IGF-BP1. Anti-Human IGF-BP1-specific antibody was purified by affinity chromatography and then biotinylated.
Sandwich ELISA: To detect hIGF-BP1 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25-1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IGF-BP1 (500-P228) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hIGF-BP1.
Western Blot: To detect hIGF-BP1 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hIGF-BP1 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
500-P228BT-1MG will be provided as 2 x 500 µg
This gene is a member of the insulin-like growth factor binding protein (IGFBP) family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors (IGFs) I and II and circulates in the plasma. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-pregnancy-associated endometrial globulin; amniotic fluid binding protein; Binding protein 25; Binding protein 26; Binding protein 28; binding protein-25; binding protein-26; binding protein-28; growth hormone independent-binding protein; hIGFBP 1; hIGFBP1; IBP 1; IBP-1; IGF BP25; IGF-binding protein 1; IGFBP; IGFBP 1; IGFBP-1; Insulin like growth factor binding protein; insulin-like growth factor binding protein 1; Insulin-like growth factor-binding protein 1; Placental protein 12; PP 12; PP12
Gene Aliases: AFBP; hIGFBP-1; IBP1; IGF-BP25; IGFBP1; PP12
UniProt ID: (Human) P08833
Entrez Gene ID: (Human) 3484
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support