Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLPLCLVAA LLLAAGPGPS LGDEAIHCPP CSEEKLARCR PPVGCEELVR EPGCGCCATC ALGLGMPCGV YTPRCGSGLR CYPPRGVEKP LHTLMHGQGV CMELAEIEAI QESLQPSDKD EGDHPNNSFS PCSAHDRRCL QKHFAKIRDR STSGGKMKVN GAPREDARPV PQGSCQSELH RALERLAASQ SRTHEDLYII PIPNCDRNGN FHPKQCHPAL DGQRGKCWCV DRKTGVKLPG GLEPKGELDC HQLADSFRE (1-258 aa encoded by BC016041 )
This gene is a member of the insulin-like growth factor binding protein family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors I and II and circulates in the plasma in both glycosylated and non-glycosylated forms. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IBP-4; IGF-binding protein 4; IGFBP; IGFBP-4; Insulin like growth factor binding protein; insulin-like growth factor binding protein 4; Insulin-like growth factor-binding protein 4
Gene Aliases: BP-4; HT29-IGFBP; IBP4; IGF-BP4; IGFBP-4; IGFBP4
UniProt ID: (Human) P22692, (Rat) P21744
Entrez Gene ID: (Human) 3487, (Rat) 360622
Molecular Function:
protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support