Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody detects the heavy chain of human IgM (~75 kDa).
Immunogen sequence: FGNPRKSKL ICQATGFSPR QIQVSWLREG KQVGSGVTTD QVQAEAKESG PTTYKVTSTL TIKESDWLSQ SMFTCRVDHR GLTFQQNASS MCVPDQDTAI RVFAIPPSFA SIFLTKSTKL TCLVTDLTTY DSVTISWTRQ NGEAVKTHTN ISESHPNATF SAVGEASICE DDWNSGERFT CTVTHTDLPS PLKQTISRPK GVALHRPDVY LLPPAREQLN LRESATITCL VTGFSPADVF VQWMQRGQPL SPEKYVTSAP MPEPQAPGRY FAHSILTVSE EEWNTGETYT CVVAHEALPN RVTERTVDKS TEGEVSADEE GFENLWATAS TFIVLFLLSL FYSTTVTLFK VK (266-614 aa encoded by BC009851)
IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene Aliases: AGM1; MU; VH
Entrez Gene ID: (Human) 3507
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support