Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAWVSFYLLP FIFSTGLCAL PVLTQPPSAS AFLGASIKLT CTLSREHSSY TIEWYQQRPG RSPQYIMKVK SDGSHNKGDG IPDRFMGSSS GADRYLTLSN LQSDDEAEYH CGESHTIDGQ VGWVFGGGTK LTVLSQPKAA PSVTLFPPSS EELQANKATL VCLISDFYPG AVTVAWKADS SPVKAGVETT TPSKQSNNKY AASSYLSLTP EQWKSHRSYS CQVTHEGSTV EKTVAPTECS
Predicted to be involved in immune response. Predicted to be located in plasma membrane. Predicted to be part of immunoglobulin complex. Predicted to be active in extracellular space. [provided by Alliance of Genome Resources, Apr 2022]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene Aliases: IGLV43; V5-1
Entrez Gene ID: (Human) 28786
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support