Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: VVAGLYPKPT LTAHPGPIMA PGESLNLRCQ GPIYGMTFAL MRVEDLEKSF YHKKTIKNEA NFFFQSLKIQ DTGHYLCFYY DASYRGSLLS D
This gene encodes a member of the immunoglobulin-like domain-containing superfamily. Proteins in this superfamily contain varying numbers of immunoglobulin-like domains and are thought to participate in the regulation of interactions between cells. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IgSF1; Immunoglobulin superfamily member 1; immunoglobulin superfamily, member 1; Immunoglobulin-like domain-containing protein 1; InhBP; inhibin binding protein; Inhibin-binding protein; MGC75490; p120; Pituitary gland-specific factor 2
Gene Aliases: CHTE; IGCD1; IGDC1; IGSF1; INHBP; KIAA0364; p120; PGSF2
UniProt ID: (Human) Q8N6C5, (Rat) Q925N6
Entrez Gene ID: (Human) 3547, (Rat) 302822
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support