Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: TLSVTQPWYL EVDYTHEAVT IKCTFSATGC PSEQPTCLWF RYGAHRPENL CLDGCKSEAD KFTVREALKE NQVSLTVNRV TSNDSAIYIC GIAFPSVPEA RAKQTGGGTT LVVREIKLLS KELRNFLTAL VSLLSVYVTG VCVAFILLSK SKSNPLRNKK IKEDSQKKKS ARRIFQEIAQ ELYHKRHVET NQQSEKDNNT YENRRVLSNY ERP
IGSF6 (DORA), a novel member of the immunoglobulin superfamily (IGSF) from human and rat expressed in dendritic and myeloid cells. Using a probe from the open reading frame of the rat cDNA, we isolated a cosmid which contains the entire mouse gene. By comparative analysis and reverse transcriptase polymerase chain reaction, we defined the intron/exon structure and the mRNA of the mouse gene and, with respect to human BAC clones, the human gene. The genes span 10 kb (mouse) and 12 kb (human), with six exons arranged in a manner similar to other members of the IGSF. All intron/exon boundaries follow the GT-AG rule. Expression of the mouse Igsf6 gene is restricted to cells of the immune system, particularly macrophages. Northern blot revealed a single mRNA of 2.5 kb, in contrast to the human gene which is expressed as two mRNAs of 1 and 2.5 kb. The human and mouse genes were localized to a locus associated with inflammatory bowel disease. Analysis of the flanking regions of the Igsf6 gene revealed the presence of an unrelated gene, transcribed from the opposite strand of the DNA and oriented such that the Igsf6 gene is encoded entirely within an intron. An identical organization is seen in human.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: down-regulated by activation (immunoglobulin superfamily); IgSF6; Immunoglobulin superfamily member 6; immunoglobulin superfamily, member 6; Protein DORA
Gene Aliases: DORA; IGSF6
UniProt ID: (Human) O95976
Entrez Gene ID: (Human) 10261
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support