Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: VLFLFEFDEQ GRELQKAFED TLQLMERSLP EIWTLTYQQN SATPVLGPNS TANSIMASYQ QQKTSVPVLD AELFIPPKIN RRTQWKLSLL
The protein encoded by this gene is a scaffold protein and a regulator for 3 different kinases involved in proinflammatory signaling. This encoded protein can bind NF-kappa-B-inducing kinase (NIK) and IKKs through separate domains and assemble them into an active kinase complex. Mutations in this gene have been associated with familial dysautonomia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp781H1425; elongator acetyltransferase complex subunit 1; Elongator complex protein 1; ELP1; FLJ12497; IkappaB kinase complex-associated protein; IKK complex-associated protein; p150; RP11-3J11.4
Gene Aliases: DYS; ELP1; FD; IKAP; IKBKAP; IKI3; TOT1
UniProt ID: (Human) O95163
Entrez Gene ID: (Human) 8518
Molecular Function:
RNA metabolism protein
nucleic acid metabolism protein
general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support