Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
DNA-binding protein IKAROS functions in the specification and the maturation of the T lymphocyte. It also interacts with a critical control element in the terminal deoxynucleotidyl transferase promoter as well as with the promoters for other genes expressed during early stages of B-cell and T-cell development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CLL-associated antigen KW-6; DNA-binding protein Ikaros; hIk-1; IKAROS family zinc finger 1 (Ikaros); Ikaros family zinc finger protein 1; Lymphoid transcription factor LyF-1; protein phosphatase 1, regulatory subunit 92; zinc finger protein, subfamily 1A, 1 (Ikaros)
Gene Aliases: 5832432G11Rik; CVID13; hlk-1; Hs.54452; IK1; IKAROS; IKZF1; LyF-1; LYF1; mKIAA4227; PPP1R92; PRO0758; Zfpn1a1; ZNFN1A1
UniProt ID: (Human) Q53XL2
Entrez Gene ID: (Pig) 100515333, (Human) 10320, (Rabbit) 100356499, (Mouse) 22778
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support