Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: METEAIDGYI TCDNELSPER EHSNMAIDLT SSTPNGQHAS PSHMTSTNSV KLEMQSDEEC DRKPLSREDE IRGHDEGSSL EEPLIESSEV ADNRKVQELQ GEGGIRLPNG ERPFHCNQCG ASFTQKGNLL RHIKLHSGEK PFKCPFCSYA CRRRDALTGH LRTHSVGKPH KCNYCGRSYK QRSSLEEHKE RCHNYLQNVS MEAAGQVMSH HVPPMEDCKE QEPIMDNNIS LVPFERPAVI EKLTGNMGKR KSSTPQKFVG EKLMRFSYPD IHFDMNLTYE KEAELMQSHM MDQAINNAIT YLGAEALHPL MQHPPSTIAE VAPVISSAYS QVYHPNRIER PISRETADSH ENNMDGPISL IRPKSRPQER EASPSNSCLD STDSESSHDD HQSYQGHPAL NPKRKQSPAY MKEDVKALDT TKAPKGSLKD IYKVFNGEGE QIRAFKCEHC RVLFLDHVMY TIHMGCHGYR DPLECNICGY RSQDRYEFSS HIVRGEHTFH
This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This protein forms homo- or hetero-dimers with other Ikaros family members, and is thought to function predominantly in early hematopoietic development. Multiple transcript variants encoding different isoforms have been found for this gene, but the biological validity of some variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IKAROS family zinc finger 2 (Helios); Ikaros family zinc finger protein 2; zinc finger DNA binding protein Helios; Zinc finger protein Helios; zinc finger protein, subfamily 1A, 2 (Helios)
Gene Aliases: ANF1A2; HELIOS; IKZF2; ZNF1A2; ZNFN1A2
UniProt ID: (Human) Q9UKS7
Entrez Gene ID: (Human) 22807
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support