Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Produced by activated macrophages, IL-1 stimulates thymocyte proliferation by inducing IL-2 release, B-cell maturation and proliferation, and fibroblast growth factor activity. IL-1 proteins are involved in the inflammatory response, being identified as endogenous pyrogens, and are reported to stimulate the release of prostaglandin and collagenase from synovial cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hematopoietin-1; IL-1 alpha; interleukin 1, alpha; Interleukin-1 alpha; preinterleukin 1 alpha; pro-interleukin-1-alpha
Gene Aliases: IL-1 alpha; IL-1A; IL1; IL1-ALPHA; IL1A; IL1F1
UniProt ID: (Human) P01583, (Mouse) P01582, (Rat) P16598
Entrez Gene ID: (Human) 3552, (Mouse) 16175, (Rat) 24493
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support