Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Description: The S4B6 monoclonal antibody reacts with mouse interleukin-2 (IL-2). Mouse IL-2 is a 17 kDa factor produced mainly by activated CD4+ T cells. IL-2 induces cell cycle progression of resting cells in an antigen non-specific manner and allows clonal expansion of activated T cells. IL-2 also acts on activated B cells, monocytes, NK cells, LAK cells, and on oligodendroglial cells in vitro. In addition, IL-2 plays a role in hematopoiesis, tumor surveillance and anti-inflammatory reactions, making this cytokine a central regulator of the immune response. Non-glycosylated IL-2 is biologically active.
The S4B6 antibody has been reported to have neutralizing activity. Studies have demonstrated that the S4B6 antibody binds a different epitope of IL-2 than JES6-1A12. In mice, S4B6 renders IL-2 incapable of binding to the high affinity IL-2R but maintains its ability to interact with the low affinity receptor. However, IL-2 bound to JES6-1A12 can still interact with the high affinity IL-2R.
Applications Reported: The S4B6 antibody has been reported for use in functional assays.
Storage and handling: Use in a sterile environment.
Filtration: 0.2 µm post-manufacturing filtered.
Purity: Greater than 90%, as determined by SDS-PAGE.
Endotoxin Level: Less than 0.001 ng/µg antibody, as determined by LAL assay.
Aggregation: Less than 10%, as determined by HPLC.
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H-IL-2; il 2; IL-2; ILN; Interleukin; Interleukin-2; Interleukin2; Involved in regulation of T cell clonal expansion; M-IL-2; T-cell growth factor; T-cell growth factor (TCGF); TCGF
Gene Aliases: Il-2; Il2
UniProt ID: (Mouse) P04351
Entrez Gene ID: (Mouse) 16183
Molecular Function:
actin or actin-binding cytoskeletal protein
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support