Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Description: This 20RNTC monoclonal antibody recognizes mouse and human IL-20R2 (interleukin-20 receptor 2, IL-20Rb) in either of its two heterodimeric complexes formed by pairing with IL-20R1 or IL-22R1. IL-20R2 belongs to the class II cytokine receptor family that shares a characteristic pattern of cysteine, proline and tryptophan residues in the extracellular domain. IL-20R2 paired with IL-20R1 forms a receptor that binds IL-20, IL-19, or IL-24. IL-20R2 paired with IL22R1 forms a receptor that binds IL-20 or IL-24. The IL-20R complex signals primarily via the JAK-STAT pathway. IL-20R2 is found predominantly on cells of epithelial origin such as gut and skin and can be upregulated during inflammation. Immunofluorescent staining of A375 cells reveals a punctate staining pattern at the cell membrane, as well as diffuse, cytoplasmic staining that is likely associated with protein synthesis and transport in the endoplasmic reticulum and golgi.
Applications Reported: This 20RNTC antibody has been reported for use in immunocytochemistry, immunoblotting (WB) and flow cytometric analysis.
Applications Tested: This 20RNTC antibody has been tested by immunofluorescent staining and flow cytometric analysis of fixed and permeabilized A375 cells. This can be used between 1-10 µg/mL.
Purity: Greater than 90%, as determined by SDS-PAGE.
Aggregation: Less than 10%, as determined by HPLC.
Filtration: 0.2 µm post-manufacturing filtered.
IL20RB and IL20RA (MIM 605620) form a heterodimeric receptor for interleukin-20 (IL20; MIM 605619) (Blumberg et al., 2001 [PubMed 11163236]).[supplied by OMIM, Feb 2009].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fibronectin type III domain containing 6; FNDC6; IL-20 receptor subunit beta; IL-20R-beta; IL-20R2; IL-20RB; interleukin 20 receptor beta subunit; interleukin-20 receptor II; Interleukin-20 receptor subunit beta
Gene Aliases: AV228068; DIRS1; FNDC6; Gm186; IL-20R2; Il20R2; IL20RB; UNQ557/PRO1114
UniProt ID: (Human) Q6UXL0
Entrez Gene ID: (Human) 53833, (Mouse) 213208
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support