Product References
Human IRF1 governs macrophagic IFN-γ immunity to mycobacteria.
Cell
Rosain J,Neehus AL,Manry J,Yang R,Le Pen J,Daher W,Liu Z,Chan YH,Tahuil N,Türel Ö,Bourgey M,Ogishi M,Doisne JM,Izquierdo HM,Shirasaki T,Le Voyer T,Guérin A,Bastard P,Moncada-Vélez M,Han JE,Khan T,Rapaport F,Hong SH,Cheung A,Haake K,Mindt BC,Pérez L,Philippot Q,Lee D,Zhang P,Rinchai D,Al Ali F,Ahmad Ata MM,Rahman M,Peel JN,Heissel S,Molina H,Kendir-Demirkol Y,Bailey R,Zhao S,Bohlen J,Mancini M,Seeleuthner Y,Roelens M,Lorenzo L,Soudée C,Paz MEJ,González ML,Jeljeli M,Soulier J,Romana S,L'Honneur AS,Materna M,Martínez-Barricarte R,Pochon M,Oleaga-Quintas C,Michev A,Migaud M,Lévy R,Alyanakian MA,Rozenberg F,Croft CA,Vogt G,Emile JF,Kremer L,Ma CS,Fritz JH,Lemon SM,Spaan AN,Manel N,Abel L,MacDonald MR,Boisson-Dupuis S,Marr N,Tangye SG,Di Santo JP,Zhang Q,Zhang SY,Rice CM,Béziat V,Lachmann N,Langlais D,Casanova JL,Gros P,Bustamante J
Published figure using IL-21 monoclonal antibody (Product # 12-7219-42) in Flow Cytometry
Thu Feb 02 00:00:00 EST 2023
Functional SARS-CoV-2-Specific Immune Memory Persists after Mild COVID-19.
Cell
Rodda LB,Netland J,Shehata L,Pruner KB,Morawski PA,Thouvenel CD,Takehara KK,Eggenberger J,Hemann EA,Waterman HR,Fahning ML,Chen Y,Hale M,Rathe J,Stokes C,Wrenn S,Fiala B,Carter L,Hamerman JA,King NP,Gale M,Campbell DJ,Rawlings DJ,Pepper M
12-7219 was used in Flow cytometry/Cell sorting to perform a longitudinal assessment of individuals recovered from mild COVID-19 to determine whether they develop and sustain multifaceted SARS-CoV-2-specific immunological memory.
Thu Jan 07 00:00:00 EST 2021
Ageing promotes early T follicular helper cell differentiation by modulating expression of RBPJ.
Aging cell
Webb LMC,Fra-Bido S,Innocentin S,Matheson LS,Attaf N,Bignon A,Novarino J,Fazilleau N,Linterman MA
12-7219-42 was used in Flow Cytometry to show increased formation of Tfh precursors (pre-Tfh) but no associated increase in germinal centre (GC)-Tfh cells in aged mice, suggesting age-driven promotion of only early Tfh cell differentiation.
Fri Jan 01 00:00:00 EST 2021
Ageing promotes early T follicular helper cell differentiation by modulating expression of RBPJ.
Aging cell
Webb LMC,Fra-Bido S,Innocentin S,Matheson LS,Attaf N,Bignon A,Novarino J,Fazilleau N,Linterman MA
12-7219-42 was used in Flow Cytometry to show increased formation of Tfh precursors (pre-Tfh) but no associated increase in germinal centre (GC)-Tfh cells in aged mice, suggesting age-driven promotion of only early Tfh cell differentiation.
Fri Jan 01 00:00:00 EST 2021
A delayed fractionated dose RTS,S AS01 vaccine regimen mediates protection via improved T follicular helper and B cell responses.
eLife
Pallikkuth S,Chaudhury S,Lu P,Pan L,Jongert E,Wille-Reece U,Pahwa S
12-7219 was used in Flow cytometry/Cell sorting to conclude that the DFD regimen favors persistence of functional B cells after the third dose.
Wed Apr 29 00:00:00 EDT 2020
Hepatocyte-derived exosomes promote T follicular regulatory cell expansion during hepatitis C virus infection.
Hepatology (Baltimore, Md.)
Cobb DA,Kim OK,Golden-Mason L,Rosen HR,Hahn YS
12-7219 was used in Flow cytometry/Cell sorting to examine the accumulation of Tfr cells in the liver compartment during chronic HCV infection and defined the cellular and molecular mechanisms underlying their expansion.
Mon Jan 01 00:00:00 EST 2018
Effects of Anti-TNFα Treatment on Mucosal Expression of IL-17A, IL-21, and IL-22 and Cytokine-Producing T Cell Subsets in Crohn's Disease.
Mediators of inflammation
Dige A,Magnusson MK,Uhrenholt C,Rasmussen TK,Kragstrup T,Öhman L,Dahlerup J,Agnholt J
12-7219 was used in Flow cytometry/Cell sorting to show that anti-TNFα treatment has no effect on mucosal IL-17A, IL-21, and IL-22 levels and cytokine-producing T cells in Crohn's diesease.
Mon Oct 01 00:00:00 EDT 2018
C5aR1 regulates T follicular helper differentiation and chronic graft-versus-host disease bronchiolitis obliterans.
JCI insight
Verghese DA,Chun N,Paz K,Fribourg M,Woodruff TM,Flynn R,Hu Y,Xiong H,Zhang W,Yi Z,Du J,Blazar BR,Heeger PS
Published figure using IL-21 monoclonal antibody (Product # 12-7219-42) in Flow Cytometry
Thu Dec 20 00:00:00 EST 2018
Genetic Architecture of Adaptive Immune System Identifies Key Immune Regulators.
Cell reports
Lagou V,Garcia-Perez JE,Smets I,Van Horebeek L,Vandebergh M,Chen L,Mallants K,Prezzemolo T,Hilven K,Humblet-Baron S,Moisse M,Van Damme P,Boeckxstaens G,Bowness P,Dubois B,Dooley J,Liston A,Goris A
12-7219-42 was used in Flow Cytometry to identify eight genome-wide significant associations explaining 6%-20% of variance in immune system regulation.
Tue Oct 16 00:00:00 EDT 2018
Activated T follicular helper-like cells are released into blood after oral vaccination and correlate with vaccine specific mucosal B-cell memory.
Scientific reports
Cárdeno A,Magnusson MK,Quiding-Järbrink M,Lundgren A
12-7219 was used in Flow cytometry/Cell sorting to investigate the frequencies, phenotype and function of circulating Tfh-like CD4+CXCR5+T cells in adults receiving an oral inactivated enterotoxigenic Escherichia coli vaccine.
Fri Feb 09 00:00:00 EST 2018
Th1 is the predominant helper T cell subset that produces GM-CSF in the joint of rheumatoid arthritis.
RMD open
Yamada H,Haraguchi A,Sakuraba K,Okazaki K,Fukushi JI,Mizu-Uchi H,Akasaki Y,Esaki Y,Kamura S,Fujimura K,Kondo M,Miyahara H,Nakashima Y,Yoshikai Y
12-7219 was used in Flow cytometry/Cell sorting to suggest the presence of common developmental pathway for these T cells.
Wed Nov 20 00:00:00 EST 2019
Cytotoxic T Cell Functions Accumulate When CD4 Is Downregulated by CD4+ T Cells in African Green Monkeys.
Journal of immunology (Baltimore, Md. : 1950)
Vinton CL,Ortiz AM,Calantone N,Mudd JC,Deleage C,Morcock DR,Whitted S,Estes JD,Hirsch VM,Brenchley JM
12-7219 was used in Flow cytometry/Cell sorting to study the functional profiles and SIV infection status in vivo of CD4+ T cells, CD8αα T cells, and CD8αΒ T cells of African green monkeys and rhesus macaques.
Thu Jun 01 00:00:00 EDT 2017
TIGIT expressing CD4+T cells represent a tumor-supportive T cell subset in chronic lymphocytic leukemia.
Oncoimmunology
Catakovic K,Gassner FJ,Ratswohl C,Zaborsky N,Rebhandl S,Schubert M,Steiner M,Gutjahr JC,Pleyer L,Egle A,Hartmann TN,Greil R,Geisberger R
12-7219 was used in Flow cytometry/Cell sorting to reveal that TIGIT+CD4+T cells provide a supportive microenvironment for CLL cells.
Sat Jan 29 00:00:00 EST 2022
Overexpression of microRNA-155 increases IL-21 mediated STAT3 signaling and IL-21 production in systemic lupus erythematosus.
Arthritis research & therapy
Rasmussen TK,Andersen T,Bak RO,Yiu G,Sørensen CM,Stengaard-Pedersen K,Mikkelsen JG,Utz PJ,Holm CK,Deleuran B
12721942 was used in flow cytometry to investigate the effect of miRNA-155 in IL-21 signaling in white cells and it role in systemic lupues erythematosus.
Tue Jun 09 00:00:00 EDT 2015
Impaired Phenotype and Function of T Follicular Helper Cells in HIV-1-Infected Children Receiving ART.
Medicine
Bekele Y,Amu S,Bobosha K,Lantto R,Nilsson A,Endale B,Gebre M,Aseffa A,Rethi B,Howe R,Chiodi F
12-7219 was used in Flow cytometry/Cell sorting to show that function and phenotype of Tfh cells, are impaired during HIV-1 infection in children, preventing adaptive B cell responses.
Wed Jul 01 00:00:00 EDT 2015
Galectin-9 and IL-21 mediate cross-regulation between Th17 and Treg cells during acute hepatitis C.
PLoS pathogens
Kared H,Fabre T,Bédard N,Bruneau J,Shoukry NH
12-7219 was used in Flow cytometry/Cell sorting to longitudinally analyse CD4 helper T cell function as well as the interaction between CD4 and CD8 T cells during acute HCV infection in individuals with spontaneous resolution vs persistent viremia.
Fri Jan 17 00:00:00 EST 2014
Increased frequency of circulating IL-21 producing Th-cells in patients with granulomatosis with polyangiitis (GPA).
Arthritis research & therapy
Abdulahad WH,Lepse N,Stegeman CA,Huitema MG,Doornbos-van der Meer B,Tadema H,Rutgers A,Limburg PC,Kallenberg CG,Heeringa P
12-7219 was used in Flow cytometry/Cell sorting to explore a possible role for IL-21 producing Th-cells in the immunopathogenesis of granulomatosis with polyangiitis.
Fri Nov 14 00:00:00 EST 2014
Engineering lymph node homing of ex vivo-expanded human natural killer cells via trogocytosis of the chemokine receptor CCR7.
Blood
Somanchi SS,Somanchi A,Cooper LJ,Lee DA
12-7219 was used in Flow cytometry/Cell sorting to explore the use of trogocytosis as a tool to transiently express CCR7 on expanded natural killer cells to enhance homing to lymph nodes.
Thu May 31 00:00:00 EDT 2012
Reprogramming CD19-specific T cells with IL-21 signaling can improve adoptive immunotherapy of B-lineage malignancies.
Cancer research
Singh H,Figliola MJ,Dawson MJ,Huls H,Olivares S,Switzer K,Mi T,Maiti S,Kebriaei P,Lee DA,Champlin RE,Cooper LJ
Sun May 15 00:00:00 EDT 2011
Increase in IL-21 producing T-cells in patients with systemic lupus erythematosus.
Arthritis research & therapy
Dolff S,Abdulahad WH,Westra J,Doornbos-van der Meer B,Limburg PC,Kallenberg CG,Bijl M
12-7219 was used in Flow cytometry/Cell sorting to reveal a pivotal role of IL-21 in the pathogenesis of SLE.
Thu Dec 13 00:00:00 EST 2012
1,25-Dihydroxyvitamin D3 and IL-2 combine to inhibit T cell production of inflammatory cytokines and promote development of regulatory T cells expressing CTLA-4 and FoxP3.
Journal of immunology (Baltimore, Md. : 1950)
Jeffery LE,Burke F,Mura M,Zheng Y,Qureshi OS,Hewison M,Walker LS,Lammas DA,Raza K,Sansom DM
12-7219 was used in Flow cytometry/Cell sorting to assess the effects of the active form of vitamin D, 1,25-dihydroxyvitamin D(3) on human CD4(+)CD25(-) T cells.
Sun Nov 01 00:00:00 EDT 2009