Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
In ELISAs, this antibody does not cross-react with recombinant human (rh) IL-17E, rmIL-17, IL-17B, C, D, or F.
Reconstitute at 0.5 mg/mL in sterile PBS.
Interleukin 25 (IL-25) is a member of the IL-17 family and is identified as a proinflammatory cytokine which is produced by Th17 effector cells thus enabling the Th17 subset to initiate or maintain the autoimmune process in diseases such as Rheumatoid Arthritis. IL-25 also induces activation of NF-kappa-B and stimulates production of the proinflammatory chemokine IL-8. IL-25 itself is activated through IL-23, IL-5, and TGF-beta.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: il 17e; IL-17E; IL-25; ILN; Interleukin; interleukin 17E; Interleukin-17E; Interleukin-25
Gene Aliases: IL-17e; Il17e; Il25
UniProt ID: (Mouse) Q8VHH8
Entrez Gene ID: (Mouse) 140806
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support