Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
IL26 is a member of the IL10 family of cytokines. It is a secreted protein and may function as a homodimer. This protein is thought to contribute to the transformed phenotype of T cells after infection by herpesvirus samimiri.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: il 26; IL-26; ILN; Interleukin; Interleukin-26; Protein AK155
Gene Aliases: AK155; IL-26; IL26
UniProt ID: (Human) Q9NPH9
Entrez Gene ID: (Human) 55801
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support