Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Leinco Technologies
Specificity: This non-therapeutic biosimilar antibody uses the same variable region sequence as the therapeutic antibody Basiliximab. Basiliximab recognizes human CD25. This product is for research use only.
Endotoxin Level: ≤ 1.0 EU/mg as determined by the LAL method. Purity: >95% by SDS-PAGE and HPLC.
Product Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using only In vitro protein free cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Pathogen Testing: To protect mouse colonies from infection by pathogens and to assure that experimental preclinical data is not affected by such pathogens, all of Leinco recombinant biosimilar antibodies are tested and guaranteed to be negative for all pathogens in the IDEXX IMPACT I Mouse Profile.
Storage and Handling: Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80 C. Avoid Repeated Freeze Thaw Cycles.
This antibody has been tested in the following applications: Western Blotting, Immunohistochemistry, Immunofluorescence, Flow Cytometry, CyTOF®, Blocking.
A recombinant chimeric (murine/human) monoclonal antibody (IgG1k) that functions as an immunosuppressive agent, specifically binding to and blocking the interleukin-2 receptor a-chain (IL-2R alpha, also known as CD25 antigen) on the surface of activated T-lymphocytes. It is a 144 kDa glycoprotein obtained from fermentation of an established mouse myeloma cell line genetically engineered to express plasmids containing the human heavy and light chain constant region genes and mouse heavy and light chain variable region genes encoding the RFT5 antibody that binds selectively to the IL-2R alpha. Basiliximab binds with high-affinity to the alpha-subunit (CD25) of the high-affinity IL-2 receptor. This inhibits IL-2 binding, which inhibits T-cell activation and prevents the body from mounting an immune response against the foreign kidney.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CHI621
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support