Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In Western blots, no cross-reactivity with recombinant human (rh) IL-3 or recombinant rat IL-3 is observed. This antibody does not neutralize the biological activity of rhIL-3.
Reconstitute at 0.5 mg/mL in sterile PBS.
Endoxin level is <0.10 EU per 1 µg of the antibody by the LAL method.
Interleukin 3 (IL-3), also called Mast Cell Growth Factor (MCGF), Multi-Colony Stimulating Factor (multi-CSF), or Eosinophil-CSF (E-CSF), is produced by activated T cells, mast cells and eosinophils. IL-3 is a hematopoietic growth factor that stimulates colony formation of erythroid, megakaryocyte, neutrophil, eosinophil, basophil, mast cell and monocytic lineages. Most of these functions are enhanced or dependent on co-stimulation with other cytokines. Human and mouse IL-3 share only 29% sequence identity at the amino acid level. The bioactivity of IL-3 is highly species specific. For example, human IL-3 has no activity on mouse cells. IL-3 is a potent growth promoting cytokine. IL-3 is capable of supporting the proliferation of a broad range of hematopoietic cell types, and is involved in a variety of cell activities such as cell growth, differentiation and apoptosis. IL-3 has been shown to also possess neurotrophic activity, and it may be associated with neurologic disorders.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BP/BPA; Colony stimulating factor multiple; Eo-CSF; Hematopoietic growth factor; IL-3; ILN; Interleukin; Interleukin 3 (colony stimulating factor; Interleukin-3; Interleukin3; Mast cell growth factor; MCGF; MGC79398; MGC79399; Multi CSF; Multilineage colony stimulating factor; multiple); Multipotential colony-stimulating factor; P cell stimulating factor; P-cell-stimulating factor; PCSA
Gene Aliases: BPA; Csfmu; HCGF; Il-3; Il3; MCGF; PSF
UniProt ID: (Mouse) P01586
Entrez Gene ID: (Mouse) 16187
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support