Product References
The significance of nanofiber polyglycolic acid for promoting tissue repair in a rat subcutaneous implantation model.
Journal of biomedical materials research. Part B, Applied biomaterials
Sueyoshi Y,Niwa A,Nishikawa Y,Isogai N
Sun Jan 01 00:00:00 EST 2023
Effects of a Macroporous Resin Extract of Dendrobium officinale Leaves in Rats with Hyperuricemia Induced by Anthropomorphic Unhealthy Lifestyle.
Evidence-based complementary and alternative medicine : eCAM
Li LZ,Wang XM,Feng XJ,Liu K,Li B,Zhu LJ,Xu WF,Zheng X,Dong YJ,He XL,Guan HR,Ding YY,Wu HS,Zhou CJ,Ye SY,Zhang BB,Lv GY,Chen SH
Tue Jan 17 00:00:00 EST 2023
PKCθ Regulates Pituitary Adenoma Bone Invasion by Activating Osteoclast in NF-κB/IL-1β-Dependent Manner.
Cancers
Wang Q,Lei Z,Wang Z,Jiang Q,Zhang Z,Liu X,Xing B,Li S,Guo X,Liu Y,Li X,Shu K,Zhang H,Huang Y,Lei T
Mon Mar 06 00:00:00 EST 2023
Enzymatic one-pot preparation of carboxylmethyl chitosan-based hydrogel with inherent antioxidant and antibacterial properties for accelerating wound healing.
International journal of biological macromolecules
Wei Q,Ma J,Jia L,Zhao H,Dong Y,Jiang Y,Zhang W,Hu Z
Tue Jan 31 00:00:00 EST 2023
YY1 complex in M2 macrophage promotes prostate cancer progression by upregulating IL-6.
Journal for immunotherapy of cancer
Chen S,Lu K,Hou Y,You Z,Shu C,Wei X,Wu T,Shi N,Zhang G,Wu J,Chen S,Zhang L,Li W,Zhang D,Ju S,Chen M,Xu B
Sat Apr 01 00:00:00 EDT 2023
Aryl hydrocarbon receptor dependent anti-inflammation and neuroprotective effects of tryptophan metabolites on retinal ischemia/reperfusion injury.
Cell death & disease
Yang Y,Wang N,Xu L,Liu Y,Huang L,Gu M,Wu Y,Guo W,Sun H
Wed Feb 08 00:00:00 EST 2023
Patrinia villosa treat colorectal cancer by activating PI3K/Akt signaling pathway.
Journal of ethnopharmacology
Li XC,Wang S,Yang XX,Li TJ,Gu JX,Zhao L,Bao YR,Meng XS
Mon Jun 12 00:00:00 EDT 2023
Mechanism of piR-1245/PIWI-like protein-2 regulating Janus kinase-2/signal transducer and activator of transcription-3/vascular endothelial growth factor signaling pathway in retinal neovascularization.
Neural regeneration research
Yu Y,Xia LK,Di Y,Nie QZ,Chen XL
Mon May 01 00:00:00 EDT 2023
Targeting DCLK1 attenuates tumor stemness and evokes antitumor immunity in triple-negative breast cancer by inhibiting IL-6/STAT3 signaling.
Breast cancer research : BCR
Liu H,Yan R,Xiao Z,Huang X,Yao J,Liu J,An G,Ge Y
Mon Apr 17 00:00:00 EDT 2023
Targeting DCLK1 attenuates tumor stemness and evokes antitumor immunity in triple-negative breast cancer by inhibiting IL-6/STAT3 signaling.
Breast cancer research : BCR
Liu H,Yan R,Xiao Z,Huang X,Yao J,Liu J,An G,Ge Y
Mon Apr 17 00:00:00 EDT 2023
Targeting DCLK1 attenuates tumor stemness and evokes antitumor immunity in triple-negative breast cancer by inhibiting IL-6/STAT3 signaling.
Breast cancer research : BCR
Liu H,Yan R,Xiao Z,Huang X,Yao J,Liu J,An G,Ge Y
Mon Apr 17 00:00:00 EDT 2023
Targeting DCLK1 attenuates tumor stemness and evokes antitumor immunity in triple-negative breast cancer by inhibiting IL-6/STAT3 signaling.
Breast cancer research : BCR
Liu H,Yan R,Xiao Z,Huang X,Yao J,Liu J,An G,Ge Y
Mon Apr 17 00:00:00 EDT 2023
TIGAR deficiency induces caspase-1-dependent trophoblasts pyroptosis through NLRP3-ASC inflammasome.
Frontiers in immunology
Guo J,Zhou M,Zhao M,Li S,Fang Z,Li A,Zhang M
Tue May 02 00:00:00 EDT 2023
Role of the Peli1-RIPK1 Signaling Axis in Methamphetamine-Induced Neuroinflammation.
ACS chemical neuroscience
Xu W,Yang T,Lou X,Chen J,Wang X,Hu M,An D,Gao R,Wang J,Chen X
Wed Mar 01 00:00:00 EST 2023
Role of the Peli1-RIPK1 Signaling Axis in Methamphetamine-Induced Neuroinflammation.
ACS chemical neuroscience
Xu W,Yang T,Lou X,Chen J,Wang X,Hu M,An D,Gao R,Wang J,Chen X
Wed Mar 01 00:00:00 EST 2023
STAT1/3 signaling suppresses axon degeneration and neuronal cell death through regulation of NAD+-biosynthetic and consuming enzymes.
Cellular signalling
Murata H,Yasui Y,Oiso K,Ochi T,Tomonobu N,Yamamoto KI,Kinoshita R,Sakaguchi M
Tue Aug 01 00:00:00 EDT 2023
Effect of stem cell conditional medium-loading adhesive hydrogel on TGF-β1-induced endometrial stromal cell fibrosis.
Frontiers in bioengineering and biotechnology
Zhu Y,Wang T,Bao MJ,Qu XH,Li ZM
Tue May 23 00:00:00 EDT 2023
Extracellular Vesicles from Carcinoma-associated Fibroblasts Promote EMT of Salivary Adenoid Cystic Carcinoma Via IL-6.
Archives of medical research
Al-Raimi HAI,Kong J,Ran Y,Zhu L,Li J,Liu X,Yang X,Qi D,Liu T
Sun Jan 01 00:00:00 EST 2023
Network pharmacology analysis combined with experimental validation to explore the therapeutic mechanism of Schisandra Chinensis Mixture on diabetic nephropathy.
Journal of ethnopharmacology
Ma Y,Deng Y,Li N,Dong A,Li H,Chen S,Zhang S,Zhang M
Fri Feb 10 00:00:00 EST 2023
Early preclinical plasma protein biomarkers of brain trauma are influenced by early seizures and levetiracetam.
Epilepsia open
Saletti PG,Mowrey WB,Liu W,Li Q,McCullough J,Aniceto R,Lin IH,Eklund M,Casillas-Espinosa PM,Ali I,Santana-Gomez C,Coles L,Shultz SR,Jones N,Staba R,O'Brien TJ,Moshé SL,Agoston DV,Galanopoulou AS
Thu Jun 01 00:00:00 EDT 2023
Akkermansia muciniphila protects mice against an emerging tick-borne viral pathogen.
Nature microbiology
Xie J,Li H,Zhang X,Yang T,Yue M,Zhang Y,Chen S,Cui N,Yuan C,Li J,Zhu SJ,Liu W
Sun Jan 01 00:00:00 EST 2023
Akkermansia muciniphila protects mice against an emerging tick-borne viral pathogen.
Nature microbiology
Xie J,Li H,Zhang X,Yang T,Yue M,Zhang Y,Chen S,Cui N,Yuan C,Li J,Zhu SJ,Liu W
Sun Jan 01 00:00:00 EST 2023
Akkermansia muciniphila protects mice against an emerging tick-borne viral pathogen.
Nature microbiology
Xie J,Li H,Zhang X,Yang T,Yue M,Zhang Y,Chen S,Cui N,Yuan C,Li J,Zhu SJ,Liu W
Sun Jan 01 00:00:00 EST 2023
Akkermansia muciniphila protects mice against an emerging tick-borne viral pathogen.
Nature microbiology
Xie J,Li H,Zhang X,Yang T,Yue M,Zhang Y,Chen S,Cui N,Yuan C,Li J,Zhu SJ,Liu W
Sun Jan 01 00:00:00 EST 2023
Rat tight junction proteins are disrupted after subchronic exposure to okadaic acid.
Environmental science and pollution research international
Huang L,Liu B,Yu XW,Pan GQ,Xu JY,Yan D,Wang YL,Guo QN
Mon May 01 00:00:00 EDT 2023
Macrophage Inactivation by Small Molecule Wedelolactone via Targeting sEH for the Treatment of LPS-Induced Acute Lung Injury.
ACS central science
Zhang J,Zhang M,Huo XK,Ning J,Yu ZL,Morisseau C,Sun CP,Hammock BD,Ma XC
Wed Mar 22 00:00:00 EDT 2023
Cartilage Damage Pathological Characteristics of Diabetic Neuropathic Osteoarthropathy.
Analytical cellular pathology (Amsterdam)
Liu PL,Diao JY,Wang Q,Liu H,Zhang Y,Liang JQ,Zhang F,Liang XJ,Zhao HM
Fri May 19 00:00:00 EDT 2023
EEF1A2 accelerates the protein translation of chemokine in rat myocardial cells induced by ischemia-reperfusion.
Heliyon
Zhang ZJ,Sun ZX,Liu HJ
Sat Apr 01 00:00:00 EDT 2023
Knockdown of LINC01138 protects human chondrocytes against IL-1β-induced damage by regulating the hsa-miR-1207-5p/KIAA0101 axis.
Immunity, inflammation and disease
Zhang J,Lv G
Sun Jan 01 00:00:00 EST 2023
Metformin Attenuates the Inflammatory Response via the Regulation of Synovial M1 Macrophage in Osteoarthritis.
International journal of molecular sciences
Zheng M,Zhu Y,Wei K,Pu H,Peng R,Xiao J,Liu C,Sun X
Fri Mar 10 00:00:00 EST 2023
Geraniol prevents Helicobacterium pylori-induced human gastric cancer signalling by enhancing peroxiredoxin-1 expression in GES-1 cells.
Microbial pathogenesis
Zhuang K,Tang H,Guo H,Yuan S
Sun Jan 01 00:00:00 EST 2023
Macrophage microvesicle-derived circ_YTHDF2 in methamphetamine-induced chronic lung injury.
The Journal of physiology
Chen L,Gu YJ,Zhang XG,Cheng L,Zhou MY,Yang Y,Wang Y
Wed Nov 01 00:00:00 EDT 2023
Magnify is a universal molecular anchoring strategy for expansion microscopy.
Nature biotechnology
Klimas A,Gallagher BR,Wijesekara P,Fekir S,DiBernardo EF,Cheng Z,Stolz DB,Cambi F,Watkins SC,Brody SL,Horani A,Barth AL,Moore CI,Ren X,Zhao Y
Thu Jun 01 00:00:00 EDT 2023
Engineered exosomes derived from miR-132-overexpresssing adipose stem cells promoted diabetic wound healing and skin reconstruction.
Frontiers in bioengineering and biotechnology
Ge L,Wang K,Lin H,Tao E,Xia W,Wang F,Mao C,Feng Y
Tue Mar 21 00:00:00 EDT 2023
Integration of proteomics and network toxicology reveals the mechanism of mercury chloride induced hepatotoxicity, in mice and HepG2 cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Cao X,Mao K,Zhang Y,Yang M,Liu H,Wang X,Hao L
Sat Jul 01 00:00:00 EDT 2023
M2 Macrophage Membrane-Mediated Biomimetic-Nanoparticle Carrying COX-siRNA Targeted Delivery for Prevention of Tendon Adhesions by Inhibiting Inflammation.
Small (Weinheim an der Bergstrasse, Germany)
Sun J,Ju F,Jin J,Wang HL,Li ZJ,Sun YC,Chen QZ,Yang QQ,Tan J,Zhou YL
Tue Aug 01 00:00:00 EDT 2023
Transcription factor Dmrt1 triggers the SPRY1-NF-κB pathway to maintain testicular immune homeostasis and male fertility.
Zoological research
Zhang MF,Wan SC,Chen WB,Yang DH,Liu WQ,Li BL,Aierken A,Du XM,Li YX,Wu WP,Yang XC,Wei YD,Li N,Peng S,Li XL,Li GP,Hua JL
Thu May 18 00:00:00 EDT 2023
Transcription factor Dmrt1 triggers the SPRY1-NF-κB pathway to maintain testicular immune homeostasis and male fertility.
Zoological research
Zhang MF,Wan SC,Chen WB,Yang DH,Liu WQ,Li BL,Aierken A,Du XM,Li YX,Wu WP,Yang XC,Wei YD,Li N,Peng S,Li XL,Li GP,Hua JL
Thu May 18 00:00:00 EDT 2023
Scutellaria baicalensis Pith-decayed Root Inhibits Macrophage-related Inflammation Through the NF-κB/NLRP3 Pathway to Alleviate LPS-induced Acute Lung Injury.
Planta medica
Zhang F,Ke C,Zhou Z,Xu K,Wang Y,Liu Y,Tu J
Sat Apr 01 00:00:00 EDT 2023
Antidepression of Xingpijieyu formula targets gut microbiota derived from depressive disorder.
CNS neuroscience & therapeutics
Li Y,Yang L,Li J,Gao W,Zhao Z,Dong K,Duan W,Dai B,Guo R
Wed Feb 01 00:00:00 EST 2023
RNF128 regulates neutrophil infiltration and myeloperoxidase functions to prevent acute lung injury.
Cell death & disease
Liu PY,Chen CY,Lin YL,Lin CM,Tsai WC,Tsai YL,Lin GJ,Chen YG,Wang SY,Sun RN,Huang YC,Chang H,Chen YC
Wed Jun 21 00:00:00 EDT 2023
A novel immunogenic cell death-related subtype classification and risk signature for predicting prognosis and immunotherapy efficacy in gastric cancer.
Frontiers in immunology
Dong B,Wu Y,Zhang J,Gu Y,Xie R,He X,Pang X,Wang X,Cui Y
Wed May 24 00:00:00 EDT 2023
Blockage of TRPV4 Downregulates the Nuclear Factor-Kappa B Signaling Pathway to Inhibit Inflammatory Responses and Neuronal Death in Mice with Pilocarpine-Induced Status Epilepticus.
Cellular and molecular neurobiology
An D,Qi X,Li K,Xu W,Wang Y,Chen X,Sha S,Wu C,Du Y,Chen L
Sat Apr 01 00:00:00 EDT 2023
Integrating network pharmacology and experimental verification to explore the pharmacological mechanisms of asparagus against polycystic ovary syndrome.
Journal of ovarian research
Xing J,Luo X,Jia K,Liu S,Chen S,Qiao G,Zhang C,Yi J
Sat Jul 01 00:00:00 EDT 2023
Long-Term High-Fat Diet Consumption Induces Cognitive Decline Accompanied by Tau Hyper-Phosphorylation and Microglial Activation in Aging.
Nutrients
Liang Z,Gong X,Ye R,Zhao Y,Yu J,Zhao Y,Bao J
Tue Jan 03 00:00:00 EST 2023
The Effect of Gut Microbiota on the Progression of Intervertebral Disc Degeneration.
Orthopaedic surgery
Yao B,Cai Y,Wang W,Deng J,Zhao L,Han Z,Wan L
Wed Mar 01 00:00:00 EST 2023
Design, synthesis, and SAR study of novel flavone 1,2,4-oxadiazole derivatives with anti-inflammatory activities for the treatment of Parkinson's disease.
European journal of medicinal chemistry
Shen ZB,Meng HW,Meng XS,Lv ZK,Fang MY,Zhang LL,Lv ZL,Li MS,Liu AK,Han JH,Li QS,Duan YJ
Wed Jul 05 00:00:00 EDT 2023
Regulatory T cells showed characteristics of T helper-17(Th17) cells in mice periodontitis model.
Oral diseases
Kou Y,Jiang Y,Liu S,Yang P,Lu Y,Liu H,Li M
Sat Apr 01 00:00:00 EDT 2023
Interleukin 6 promotes BMP9-induced osteoblastic differentiation through Stat3/mTORC1 in mouse embryonic fibroblasts.
Aging
Wang SY,Jiang JH,Liu SY,Zhang J,Gao X,Liu H,Ke KX,Jiang Y,Liu L,He BC
Wed Feb 01 00:00:00 EST 2023
Inhibition of STAT3 signaling as critical molecular event in HUC-MSCs suppressed Glioblastoma Cells.
Journal of Cancer
Wang M,Zhang Y,Liu M,Jia Y,He J,Xu X,Shi H,Zhang Y,Zhang J,Liu Y
Sat Apr 15 00:00:00 EDT 2023
Inhibition of mGluR5 ameliorates lipid accumulation and inflammation in HepG2 cells.
Biochemical and biophysical research communications
Zeng S,Tao M,Yuan L,Zhang L,Luo X
Sun Apr 23 00:00:00 EDT 2023
Hydrogen sulfide attenuates TMAO‑induced macrophage inflammation through increased SIRT1 sulfhydration.
Molecular medicine reports
Liu MH,Lin XL,Xiao LL
Sat Jul 01 00:00:00 EDT 2023
MDM2 upregulation induces mitophagy deficiency via Mic60 ubiquitination in fetal microglial inflammation and consequently neuronal DNA damage caused by exposure to ZnO-NPs during pregnancy.
Journal of hazardous materials
Zhang Y,Zhang Y,Lei Y,Wu J,Kang Y,Zheng S,Shao L
Tue Sep 05 00:00:00 EDT 2023
The upregulation of peripheral blood polyamine metabolites spermidine and spermine in children with hand, foot, mouth disease is related to enterovirus 71 capsid protein VP1, but not VP4.
Translational pediatrics
Li C,Zhang W,Chang X,Di X,Xie Q,Lin B,Zhang H,Ye Z,Lan M,Lian J,Zhang H,Qiu X,Zeng J,Huang M
Tue Feb 28 00:00:00 EST 2023
Reversing inflammatory microenvironment by a single intra-articular injection of multi-stimulus responsive lipogel to relieve rheumatoid arthritis and promote joint repair.
Materials today. Bio
Tao J,Yang P,Gao M,Zhang F,Wu Y,Jiang Y,Ning Y,Li Z,Ai F
Thu Jun 01 00:00:00 EDT 2023
Fibroblast growth factor 10 delays the progression of osteoarthritis by attenuating synovial fibrosis via inhibition of IL-6/JAK2/STAT3 signaling in vivo and in vitro.
Molecular immunology
Su W,Zheng X,Zhou H,Yang S,Zhu X
Sat Jul 01 00:00:00 EDT 2023
Up-regulation of PPAR-γ involved in the therapeutic effect of icariin on cigarette smoke-induced inflammation.
Pulmonary pharmacology & therapeutics
Li Q,Zhang H,Yan X,Zhao Z,Qiu J,Hu L,Jiang S,Kong Q,Sun J,Li L
Sat Apr 01 00:00:00 EDT 2023
Epsin1-mediated exosomal sorting of Dll4 modulates the tubular-macrophage crosstalk in diabetic nephropathy.
Molecular therapy : the journal of the American Society of Gene Therapy
Liu JL,Zhang L,Huang Y,Li XH,Liu YF,Zhang SM,Zhao YE,Chen XJ,Liu Y,He LY,Dong Z,Liu FY,Sun L,Xiao L
Wed May 03 00:00:00 EDT 2023
Role of NF-κB in lead exposure-induced activation of astrocytes based on bioinformatics analysis of hippocampal proteomics.
Chemico-biological interactions
Fan S,Weixuan W,Han H,Liansheng Z,Gang L,Jierui W,Yanshu Z
Wed Jan 25 00:00:00 EST 2023
Role of nuclear factor of activated T Cells-1 in Sepsis-induced behavioral deficits in mice.
Brain research
Jiang F,Guo Y,Hu L,Zhang M,Meng J,Si Y,Shi H
Mon May 01 00:00:00 EDT 2023
Intestinal edema induced by LPS-induced endotoxemia is associated with an inflammasome adaptor ASC.
PloS one
Yamamoto T,Kurata M,Kaneko N,Masumoto J
Wed Feb 22 00:00:00 EST 2023
Astragalus polysaccharides alleviate type 1 diabetes via modulating gut microbiota in mice.
International journal of biological macromolecules
Yang B,Xiong Z,Lin M,Yang Y,Chen Y,Zeng J,Jia X,Feng L
Sat Apr 15 00:00:00 EDT 2023
FNDC5/Irisin Inhibits the Inflammatory Response and Mediates the Aerobic Exercise-Induced Improvement of Liver Injury after Myocardial Infarction.
International journal of molecular sciences
Wang T,Yu M,Li H,Qin S,Ren W,Ma Y,Bo W,Xi Y,Cai M,Tian Z
Sun Feb 19 00:00:00 EST 2023
Chronic Microcystin-LR-Induced α-Synuclein Promotes Neuroinflammation Through Activation of the NLRP3 Inflammasome in Microglia.
Molecular neurobiology
Yan M,Jin H,Pan C,Han X
Wed Feb 01 00:00:00 EST 2023
STING/TBK1 Regulates Inflammation in Macrophages and Titanium Particles-Induced Osteolysis.
ACS biomaterials science & engineering
Ouyang Z,Xu J,Liu T,Lin S,Sun Y,Huang Y,Zheng Z,Zeng G,Li C,Li S,Ding Y
Mon Jun 12 00:00:00 EDT 2023
Astragaloside IV attenuates high glucose-induced NF-κB-mediated inflammation through activation of PI3K/AKT-ERK-dependent Nrf2/ARE signaling pathway in glomerular mesangial cells.
Phytotherapy research : PTR
Su X,Guo H,Zhou Y,Cao A,Shen Q,Zhu B,Yao X,Wang Y,Wang H,Wang L
Fri Sep 01 00:00:00 EDT 2023
Glucose Transporter Glut1-Dependent Metabolic Reprogramming Regulates Lipopolysaccharide-Induced Inflammation in RAW264.7 Macrophages.
Biomolecules
Cornwell A,Ziółkowski H,Badiei A
Sat Apr 29 00:00:00 EDT 2023
SM22α Deletion Contributes to Neurocognitive Impairment in Mice through Modulating Vascular Smooth Muscle Cell Phenotypes.
International journal of molecular sciences
Xu X,Liu XQ,Liu XL,Wang X,Zhang WD,Huang XF,Jia FY,Kong P,Han M
Wed Apr 12 00:00:00 EDT 2023
Autophagy supports mitochondrial metabolism through the regulation of iron homeostasis in pancreatic cancer.
Science advances
Mukhopadhyay S,Encarnación-Rosado J,Lin EY,Sohn ASW,Zhang H,Mancias JD,Kimmelman AC
Fri Apr 21 00:00:00 EDT 2023
Verniciflavanol A, a profisetinidin-type-4-arylflavan-3-ol from toxicodendron vernicifluum protects SH-SY5Y cells against H2O2-Induced oxidative stress.
Phytochemistry
Li M,Zhang Y,Han T,Guan L,Fan D,Wu Q,Liu J,Xu Y,Fan Y
Sun Jan 01 00:00:00 EST 2023
Dietary cholesterol drives the development of nonalcoholic steatohepatitis by altering gut microbiota mediated bile acid metabolism in high-fat diet fed mice.
The Journal of nutritional biochemistry
Gao X,Lin X,Xin Y,Zhu X,Li X,Chen M,Huang Z,Guo H
Sat Jul 01 00:00:00 EDT 2023
Ginsenoside Rg1 alleviates learning and memory impairments and Aβ disposition through inhibiting NLRP1 inflammasome and autophagy dysfunction in APP/PS1 mice.
Molecular medicine reports
Li X,Huang L,Kong L,Su Y,Zhou H,Ji P,Sun R,Wang C,Li W,Li W
Sun Jan 01 00:00:00 EST 2023
Onychiol B attenuates lipopolysaccharide-induced inflammation via MAPK/NF-κB pathways and acute lung injury in vivo.
Bioorganic chemistry
Pei X,Zhang Z,Wang N,Huang G,Min X,Yang Y,Cao J
Wed Mar 01 00:00:00 EST 2023
Effects of gradient high-field static magnetic fields on diabetic mice.
Zoological research
Yu B,Song C,Feng CL,Zhang J,Wang Y,Zhu YM,Zhang L,Ji XM,Tian XF,Cheng GF,Chen WL,Zablotskii V,Wang H,Zhang X
Sat Mar 18 00:00:00 EDT 2023
Polydopamine-Pd nanozymes as potent ROS scavengers in combination with near-infrared irradiation for osteoarthritis treatment.
iScience
Hu H,Yang J,Zhong Y,Wang J,Cai J,Luo C,Jin Z,Gao M,He M,Zheng L
Fri May 19 00:00:00 EDT 2023
Fisetin treatment alleviates kidney injury in mice with diabetes-exacerbated atherosclerosis through inhibiting CD36/fibrosis pathway.
Acta pharmacologica Sinica
Zou TF,Liu ZG,Cao PC,Zheng SH,Guo WT,Wang TX,Chen YL,Duan YJ,Li QS,Liao CZ,Xie ZL,Han JH,Yang XX
Sun Oct 01 00:00:00 EDT 2023
Neuro death through autophagy via the acetylation of FoxO1 by SIRT2 in the hippocampus of mice in a autism spectrum disorder mice model.
Journal of cellular physiology
Guo L,Jiang ZM,Zhan YJ,Pan W,Wu QW,Song FX,Zhou X,Zhou XY,Liu LJ,Wang JT
Thu Jun 01 00:00:00 EDT 2023
Direct targeting of sEH with alisol B alleviated the apoptosis, inflammation, and oxidative stress in cisplatin-induced acute kidney injury.
International journal of biological sciences
Zhang J,Luan ZL,Huo XK,Zhang M,Morisseau C,Sun CP,Hammock BD,Ma XC
Wed Jan 04 00:00:00 EST 2023
NQO1 alleviates renal fibrosis by inhibiting the TLR4/NF-κB and TGF-β/Smad signaling pathways in diabetic nephropathy.
Cellular signalling
Qiu D,Song S,Chen N,Bian Y,Yuan C,Zhang W,Duan H,Shi Y
Tue Aug 01 00:00:00 EDT 2023
NQO1 alleviates renal fibrosis by inhibiting the TLR4/NF-κB and TGF-β/Smad signaling pathways in diabetic nephropathy.
Cellular signalling
Qiu D,Song S,Chen N,Bian Y,Yuan C,Zhang W,Duan H,Shi Y
Tue Aug 01 00:00:00 EDT 2023
NQO1 alleviates renal fibrosis by inhibiting the TLR4/NF-κB and TGF-β/Smad signaling pathways in diabetic nephropathy.
Cellular signalling
Qiu D,Song S,Chen N,Bian Y,Yuan C,Zhang W,Duan H,Shi Y
Tue Aug 01 00:00:00 EDT 2023
NQO1 alleviates renal fibrosis by inhibiting the TLR4/NF-κB and TGF-β/Smad signaling pathways in diabetic nephropathy.
Cellular signalling
Qiu D,Song S,Chen N,Bian Y,Yuan C,Zhang W,Duan H,Shi Y
Tue Aug 01 00:00:00 EDT 2023
Sodium butyrate alleviates pre-eclampsia in pregnant rats by improving the gut microbiota and short-chain fatty acid metabolites production.
Journal of applied microbiology
Yong W,Zhao Y,Jiang X,Li P
Tue Feb 01 00:00:00 EST 2022
Betaine supplementation alleviates dextran sulfate sodium-induced colitis via regulating the inflammatory response, enhancing the intestinal barrier, and altering gut microbiota.
Food & function
Zhao N,Yang Y,Chen C,Jing T,Hu Y,Xu H,Wang S,He Y,Liu E,Cui J
Tue Dec 13 00:00:00 EST 2022
Activation of PGC-1α via isoliquiritigenin-induced downregulation of miR-138-5p alleviates nonalcoholic fatty liver disease.
Phytotherapy research : PTR
Wang L,Wang X,Kong L,Li Y,Huang K,Wu J,Wang C,Sun H,Sun P,Gu J,Luo H,Liu K,Meng Q
Tue Feb 01 00:00:00 EST 2022
Design, Synthesis and Evaluation of Novel 1,4-Disubstituted Piperazine-2,5-dione Derivatives as Antioxidants against H2O2-Induced Oxidative Injury via the IL-6/Nrf2 Loop Pathway.
Antioxidants (Basel, Switzerland)
Xiong L,Wu H,Zhong T,Luo F,Li Q,Li M,Fan Y
Wed Oct 12 00:00:00 EDT 2022
Red Nucleus Interleukin-6 Evokes Tactile Allodynia in Male Rats Through Modulating Spinal Pro-inflammatory and Anti-inflammatory Cytokines.
Frontiers in molecular neuroscience
Yang QQ,Li HN,Xia YT,Tian X,Feng F,Yang J,Xu YL,Guo J,Li XQ,Wang JY,Zeng XY
Fri Apr 29 00:00:00 EDT 2022
Red Nucleus Interleukin-6 Evokes Tactile Allodynia in Male Rats Through Modulating Spinal Pro-inflammatory and Anti-inflammatory Cytokines.
Frontiers in molecular neuroscience
Yang QQ,Li HN,Xia YT,Tian X,Feng F,Yang J,Xu YL,Guo J,Li XQ,Wang JY,Zeng XY
Fri Apr 29 00:00:00 EDT 2022
Magnolol limits NFκB-dependent inflammation by targeting PPARγ relieving retinal ischemia/reperfusion injury.
International immunopharmacology
Wang N,Yang Y,Liu Y,Huang L,Gu M,Wu Y,Xu L,Sun H,Guo W
Tue Nov 01 00:00:00 EDT 2022
Electroacupuncture Alleviates Neuroinflammation by Inhibiting the HMGB1 Signaling Pathway in Rats with Sepsis-Associated Encephalopathy.
Brain sciences
Xin Y,Wang J,Chu T,Zhou Y,Liu C,Xu A
Sat Dec 17 00:00:00 EST 2022
Adrenomedullin Improves Cardiac Remodeling and Function in Obese Rats with Hypertension.
Pharmaceuticals (Basel, Switzerland)
Qian P,Wang Q,Wang FZ,Dai HB,Wang HY,Gao Q,Zhou H,Zhou YB
Mon Jun 06 00:00:00 EDT 2022
Electroacupuncture attenuates brain injury through α7 nicotinic acetylcholine receptor-mediated suppression of neuroinflammation in a rat model of asphyxial cardiac arrest.
Journal of neuroimmunology
Liu Y,Zhang L,Han R,Bai W,Li J,Gao C
Wed Jun 15 00:00:00 EDT 2022
Synergistic Effects of Photobiomodulation Therapy with Combined Wavelength on Diabetic Wound Healing In Vitro and In Vivo.
Photobiomodulation, photomedicine, and laser surgery
Cai W,Hamushan M,Zhang Y,Xu Z,Ren Z,Du J,Ju J,Cheng P,Tan M,Han P
Sat Jan 01 00:00:00 EST 2022
Mesenchymal Stem Cell Exosomes Encapsulated Oral Microcapsules for Acute Colitis Treatment.
Advanced healthcare materials
Gan J,Sun L,Chen G,Ma W,Zhao Y,Sun L
Thu Sep 01 00:00:00 EDT 2022
Exosomes Derived from Baicalin-Pretreated Mesenchymal Stem Cells Alleviate Hepatocyte Ferroptosis after Acute Liver Injury via the Keap1-NRF2 Pathway.
Oxidative medicine and cellular longevity
Zhao S,Huang M,Yan L,Zhang H,Shi C,Liu J,Zhao S,Liu H,Wang B
Tue Aug 02 00:00:00 EDT 2022
One-Step Synthesis of Multifunctional Chitosan Hydrogel for Full-Thickness Wound Closure and Healing.
Advanced healthcare materials
Guo S,Yao M,Zhang D,He Y,Chang R,Ren Y,Guan F
Tue Feb 01 00:00:00 EST 2022
Gastrodin ameliorates the lipopolysaccharide-induced neuroinflammation in mice by downregulating miR-107-3p.
Frontiers in pharmacology
Song JJ,Li H,Wang N,Zhou XY,Liu Y,Zhang Z,Feng Q,Chen YL,Liu D,Liang J,Ma XY,Wen XR,Fu YY
Tue Dec 27 00:00:00 EST 2022
Isoliquiritigenin attenuates neuroinflammation in mice model of Parkinson's disease by promoting Nrf2/NQO-1 pathway.
Translational neuroscience
Huang L,Han Y,Zhou Q,Sun Z,Yan J
Sat Jan 01 00:00:00 EST 2022
Injectable multifunctional CMC/HA-DA hydrogel for repairing skin injury.
Materials today. Bio
Cui L,Li J,Guan S,Zhang K,Zhang K,Li J
Tue Mar 01 00:00:00 EST 2022
Drawn-on-Skin Sensors from Fully Biocompatible Inks toward High-Quality Electrophysiology.
Small (Weinheim an der Bergstrasse, Germany)
Patel S,Ershad F,Lee J,Chacon-Alberty L,Wang Y,Morales-Garza MA,Haces-Garcia A,Jang S,Gonzalez L,Contreras L,Agarwal A,Rao Z,Liu G,Efimov IR,Zhang YS,Zhao M,Isseroff RR,Karim A,Elgalad A,Zhu W,Wu X,Yu C
Thu Sep 01 00:00:00 EDT 2022
A Novel Pyroptosis-Related Gene Signature for Predicting Prognosis in Kidney Renal Papillary Cell Carcinoma.
Frontiers in genetics
Hu J,Chen Y,Gao L,Ge C,Xie X,Lei P,Zhang Y,Liang P
Wed Apr 13 00:00:00 EDT 2022
Moderate Aerobic Exercise Regulates Follicular Dysfunction by Initiating Brain-Derived Neurotrophic Factor (BDNF)-Mediated Anti-Apoptotic Signaling Pathways in Polycystic Ovary Syndrome.
Journal of clinical medicine
Zhang Y,Chen D,Wang D,Wang L,Weng Y,Wang H,Wu X,Wang Y
Fri Sep 23 00:00:00 EDT 2022
Protection of CAPE-pNO2 Against Chronic Myocardial Ischemia by the TGF-Β1/Galectin-3 Pathway In Vivo and In Vitro.
Inflammation
Wan Q,Zhang L,Zhou Q,Han Y,Li Z,Li B
Wed Jun 01 00:00:00 EDT 2022
Protection of CAPE-pNO2 Against Chronic Myocardial Ischemia by the TGF-Β1/Galectin-3 Pathway In Vivo and In Vitro.
Inflammation
Wan Q,Zhang L,Zhou Q,Han Y,Li Z,Li B
Wed Jun 01 00:00:00 EDT 2022
Protection of CAPE-pNO2 Against Chronic Myocardial Ischemia by the TGF-Β1/Galectin-3 Pathway In Vivo and In Vitro.
Inflammation
Wan Q,Zhang L,Zhou Q,Han Y,Li Z,Li B
Wed Jun 01 00:00:00 EDT 2022
Sonodynamic Therapy Promotes Efferocytosis via CD47 Down-Regulation in Advanced Atherosclerotic Plaque.
International heart journal
Cao Y,Yao J,Gao W,Cao Z,Diabakte K,Wang L,Sun X,Tian Y
Sat Jan 29 00:00:00 EST 2022
Apigenin Ameliorates Hyperuricemia and Renal Injury through Regulation of Uric Acid Metabolism and JAK2/STAT3 Signaling Pathway.
Pharmaceuticals (Basel, Switzerland)
Liu T,Gao H,Zhang Y,Wang S,Lu M,Dai X,Liu Y,Shi H,Xu T,Yin J,Gao S,Wang L,Zhang D
Mon Nov 21 00:00:00 EST 2022
Mechanisms of Dendrobium officinale polysaccharides in repairing gastric mucosal injuries based on mitogen-activated protein kinases (MAPK) signaling pathway.
Bioengineered
Ma S,Wu Q,Zhao Z,Xiong J,Niu J,Liu C,Liu T,Chai Y,Qu X,Ma Z,Zhang L,Pu X
Sat Jan 01 00:00:00 EST 2022
The keratin 17/YAP/IL6 axis contributes to E-cadherin loss and aggressiveness of diffuse gastric cancer.
Oncogene
Li M,Rao X,Cui Y,Zhang L,Li X,Wang B,Zheng Y,Teng L,Zhou T,Zhuo W
Tue Feb 01 00:00:00 EST 2022
Maresin 1 alleviates sevoflurane-induced neuroinflammation in neonatal rats via JAK2/STAT3/IL-6 pathways.
International immunopharmacology
Wu Y,Sun Z,Wang T,Wang Y,Zhong L,Kong F
Fri Jul 01 00:00:00 EDT 2022
Gene expression and methylation profiles show the involvement of POMC in primary hyperparathyroidsm.
Journal of translational medicine
Zhou WX,Wang S,Wu TC,Cheng LC,Du Y,Wu W,Lin C,Li XY,Hu ZL
Tue Aug 16 00:00:00 EDT 2022
Intra-Articular Injection of Adipose-Derived Stem Cells Ameliorates Pain and Cartilage Anabolism/Catabolism in Osteoarthritis: Preclinical and Clinical Evidences.
Frontiers in pharmacology
Yan B,Lv S,Tong P,Yan L,Chen Z,Zhou L,Yuan Q,Guo L,Shan L
Fri Apr 08 00:00:00 EDT 2022
Synergistic anti-inflammatory effects of peimine, peiminine, and forsythoside a combination on LPS-induced acute lung injury by inhibition of the IL-17-NF-κB/MAPK pathway activation.
Journal of ethnopharmacology
Liu C,Zhen D,Du H,Gong G,Wu Y,Ma Q,Quan ZS
Thu Sep 15 00:00:00 EDT 2022
Epac activation ameliorates tubulointerstitial inflammation in diabetic nephropathy.
Acta pharmacologica Sinica
Yang WX,Liu Y,Zhang SM,Wang HF,Liu YF,Liu JL,Li XH,Zeng MR,Han YZ,Liu FY,Sun L,Xiao L
Tue Mar 01 00:00:00 EST 2022
Epac activation ameliorates tubulointerstitial inflammation in diabetic nephropathy.
Acta pharmacologica Sinica
Yang WX,Liu Y,Zhang SM,Wang HF,Liu YF,Liu JL,Li XH,Zeng MR,Han YZ,Liu FY,Sun L,Xiao L
Tue Mar 01 00:00:00 EST 2022
Epac activation ameliorates tubulointerstitial inflammation in diabetic nephropathy.
Acta pharmacologica Sinica
Yang WX,Liu Y,Zhang SM,Wang HF,Liu YF,Liu JL,Li XH,Zeng MR,Han YZ,Liu FY,Sun L,Xiao L
Tue Mar 01 00:00:00 EST 2022
Epac activation ameliorates tubulointerstitial inflammation in diabetic nephropathy.
Acta pharmacologica Sinica
Yang WX,Liu Y,Zhang SM,Wang HF,Liu YF,Liu JL,Li XH,Zeng MR,Han YZ,Liu FY,Sun L,Xiao L
Tue Mar 01 00:00:00 EST 2022
Using Network Pharmacology and Animal Experiment to Investigate the Therapeutic Mechanisms of Polydatin against Vincristine-Induced Neuropathic Pain.
Mediators of inflammation
Xi P,Mao R,Wu S,Liu L,Cai C,Lu L,Zhang C,Li Y
Wed Oct 26 00:00:00 EDT 2022
Diminished miR-374c-5p negatively regulates IL (interleukin)-6 in unexplained recurrent spontaneous abortion.
Journal of molecular medicine (Berlin, Germany)
Zhao L,Han L,Hei G,Wei R,Zhang Z,Zhu X,Guo Q,Chu C,Fu X,Xu K,Li X
Fri Jul 01 00:00:00 EDT 2022
Rice protein suppresses 4-hydroxy-2-nonenal-induced inflammation owing to methionine availability.
Applied physiology, nutrition, and metabolism = Physiologie appliquee, nutrition et metabolisme
Wang Z,Liang M,Li H,Liu B,Yang L
Mon Aug 01 00:00:00 EDT 2022
DPSCs Protect Architectural Integrity and Alleviate Intervertebral Disc Degeneration by Regulating Nucleus Pulposus Immune Status.
Stem cells international
Dong X,Hu F,Yi J,Zhang Y,Liu C,Geng P,Duan H,Wu CT,Zhang X,Wang H
Fri Oct 28 00:00:00 EDT 2022
Ganluyin ameliorates DSS-induced ulcerative colitis by inhibiting the enteric-origin LPS/TLR4/NF-κB pathway.
Journal of ethnopharmacology
Xiong T,Zheng X,Zhang K,Wu H,Dong Y,Zhou F,Cheng B,Li L,Xu W,Su J,Huang J,Jiang Z,Li B,Zhang B,Lv G,Chen S
Tue May 10 00:00:00 EDT 2022
Procyanidin B2 ameliorates the progression of osteoarthritis: An in vitro and in vivo study.
International immunopharmacology
Cai W,Zhang Y,Jin W,Wei S,Chen J,Zhong C,Zhong Y,Tu C,Peng H
Thu Dec 01 00:00:00 EST 2022
Laser-assisted nanocarrier delivery to achieve cutaneous siRNA targeting for attenuating psoriasiform dermatitis.
Journal of controlled release : official journal of the Controlled Release Society
Lee WR,Chou WL,Lin ZC,Sung CT,Lin CY,Fang JY
Fri Jul 01 00:00:00 EDT 2022
Isoliquiritigenin-mediated miR-23a-3p inhibition activates PGC-1α to alleviate alcoholic liver injury.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Wang L,Kong L,Xu S,Wang X,Huang K,Wang S,Wu J,Wang C,Sun H,Liu K,Meng Q
Tue Feb 01 00:00:00 EST 2022
Trehalose inhibits ferroptosis via NRF2/HO-1 pathway and promotes functional recovery in mice with spinal cord injury.
Aging
Gong F,Ge T,Liu J,Xiao J,Wu X,Wang H,Zhu Y,Xia D,Hu B
Sun Apr 10 00:00:00 EDT 2022
JQ1 attenuates neuroinflammation by inhibiting the inflammasome-dependent canonical pyroptosis pathway in SAE.
Brain research bulletin
Zhong X,Chen Z,Wang Y,Mao M,Deng Y,Shi M,Xu Y,Chen L,Cao W
Sat Oct 15 00:00:00 EDT 2022
Fecal Microbiota Transplantation Ameliorates Active Ulcerative Colitis by Downregulating Pro-inflammatory Cytokines in Mucosa and Serum.
Frontiers in microbiology
Zhang WH,Jin ZY,Yang ZH,Zhang JY,Ma XH,Guan J,Sun BL,Chen X
Sat Jul 16 00:00:00 EDT 2022
Effects of BMAL1 on dentinogenic differentiation of dental pulp stem cells via PI3K/Akt/mTOR pathway.
International endodontic journal
Xu H,Zhao J,Chen G,Yuan Z,Liu J
Sun May 01 00:00:00 EDT 2022
Tumor-associated macrophages promote migration and invasion via modulating IL-6/STAT3 signaling in renal cell carcinoma.
International immunopharmacology
Chen S,Qian S,Zhang L,Pan X,Qu F,Yu Y,Gu Z,Cui X,Shen H
Sat Oct 01 00:00:00 EDT 2022
Hepatoprotective mechanism of Silybum marianum on nonalcoholic fatty liver disease based on network pharmacology and experimental verification.
Bioengineered
Jiang G,Sun C,Wang X,Mei J,Li C,Zhan H,Liao Y,Zhu Y,Mao J
Tue Mar 01 00:00:00 EST 2022
Injectable gelatin/oxidized dextran hydrogel loaded with apocynin for skin tissue regeneration.
Biomaterials advances
Guan S,Zhang K,Cui L,Liang J,Li J,Guan F
Tue Feb 01 00:00:00 EST 2022
Experimental Periodontitis Deteriorates Cognitive Function and Impairs Insulin Signaling in a Streptozotocin-Induced Alzheimer's Disease Rat Model.
Journal of Alzheimer's disease : JAD
Duan L,Qian X,Wang Q,Huang L,Ge S
Thu Jul 07 00:00:00 EDT 2022
Melatonin regulates the immune response and improves Sjögren's syndrome-like symptoms in NOD/Ltj Mice.
Biochemical pharmacology
Liu Y,Weng X,Wei M,Yu S,Ding Y,Cheng B
Fri Jul 01 00:00:00 EDT 2022
Melatonin regulates the immune response and improves Sjögren's syndrome-like symptoms in NOD/Ltj Mice.
Biochemical pharmacology
Liu Y,Weng X,Wei M,Yu S,Ding Y,Cheng B
Fri Jul 01 00:00:00 EDT 2022
In vitro and in vivo effects of hyperglycemia and diabetes mellitus on nucleus pulposus cell senescence.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society
Jiang Z,Jiang C,Jin L,Chen Z,Feng Z,Jiang X,Cao Y
Sat Oct 01 00:00:00 EDT 2022
P38α deficiency in macrophages ameliorates murine experimental colitis by regulating inflammation and immune process.
Pathology, research and practice
Chen W,Liang R,Yi Y,Zhu J,Zhang J
Sun May 01 00:00:00 EDT 2022
Prevotella genus and its related NOD-like receptor signaling pathway in young males with stage III periodontitis.
Frontiers in microbiology
Zhao Y,Ye Q,Feng Y,Chen Y,Tan L,Ouyang Z,Zhao J,Hu J,Chen N,Su X,Dusenge MA,Feng Y,Guo Y
Tue Jan 03 00:00:00 EST 2023
Mussel-inspired adhesive gelatin-polyacrylamide hydrogel wound dressing loaded with tetracycline hydrochloride to enhance complete skin regeneration.
Soft matter
Wang Y,Xiao D,Quan L,Chai H,Sui X,Wang B,Xu H,Mao Z
Wed Jan 19 00:00:00 EST 2022
Effect of metformin in autistic BTBR T + Itpr3tf/J mice administered a high-fat diet.
Brain research bulletin
Deng W,Li F,Ke H,Wang S,Li Z,Lv P,Chen Y
Wed Jun 01 00:00:00 EDT 2022
The protective mechanism of folic acid on hyperhomocysteinemia-related arterial injury in spontaneously hypertensive rats: Folic acid against arterial inflammation.
Vascular
Zhang L,Li Z,Xing C,Ma X,Xu R
Sat Oct 01 00:00:00 EDT 2022
Kaempferol From Penthorum chinense Pursh Attenuates Hepatic Ischemia/Reperfusion Injury by Suppressing Oxidative Stress and Inflammation Through Activation of the Nrf2/HO-1 Signaling Pathway.
Frontiers in pharmacology
Chen Y,Li T,Tan P,Shi H,Cheng Y,Cai T,Bai J,Du Y,Fu W
Sat Jul 16 00:00:00 EDT 2022
Multifunctional biomimetic hydrogel based on graphene nanoparticles and sodium alginate for peripheral nerve injury therapy.
Biomaterials advances
Jin Y,Zhang W,Zhang Y,Yang Y,Fang Z,Song J,Qian Y,Yuan WE
Fri Apr 01 00:00:00 EDT 2022
Artesunate attenuates atherosclerosis by inhibiting macrophage M1-like polarization and improving metabolism.
International immunopharmacology
Wang X,Du H,Li X
Sat Jan 01 00:00:00 EST 2022
Thermosensitive Hydrogel Incorporating Prussian Blue Nanoparticles Promotes Diabetic Wound Healing via ROS Scavenging and Mitochondrial Function Restoration.
ACS applied materials & interfaces
Xu Z,Liu Y,Ma R,Chen J,Qiu J,Du S,Li C,Wu Z,Yang X,Chen Z,Chen T
Wed Mar 30 00:00:00 EDT 2022
Transcriptional landscape of human neuroblastoma cells in response to SARS-CoV-2.
BMC neuroscience
Yang RC,Huang K,Zhang HP,Li L,Tan C,Chen HC,Jin ML,Wang XR
Wed Jul 06 00:00:00 EDT 2022
The Defenders of the Alveolus Succumb in COVID-19 Pneumonia to SARS-CoV-2, Necroptosis, Pyroptosis and Panoptosis.
bioRxiv : the preprint server for biology
Schifanella L,Anderson J,Wieking G,Southern PJ,Antinori S,Galli M,Corbellino M,Lai A,Klatt N,Schacker TW,Haase AT
Mon Aug 08 00:00:00 EDT 2022
Novel Flavan-3,4-diol vernicidin B from Toxicodendron Vernicifluum (Anacardiaceae) as potent antioxidant via IL-6/Nrf2 cross-talks pathways.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Zhong T,Li M,Wu H,Wang D,Liu J,Xu Y,Fan Y
Wed Jun 01 00:00:00 EDT 2022
IL-7-Treated Periodontal Ligament Cells Regulate Local Immune Homeostasis by Modulating Treg/Th17 Cell Polarization.
Frontiers in medicine
Yu XY,Zhang ZQ,Huang JC,Lin JY,Cai XP,Liu CF
Tue Mar 15 00:00:00 EDT 2022
(-)-Epicatechin Reduces Neuroinflammation, Protects Mitochondria Function, and Prevents Cognitive Impairment in Sepsis-Associated Encephalopathy.
Oxidative medicine and cellular longevity
Ling J,Wu Y,Zou X,Chang Y,Li G,Fang M
Mon Jun 06 00:00:00 EDT 2022
Berberine inhibits gastric cancer development and progression by regulating the JAK2/STAT3 pathway and downregulating IL-6.
Life sciences
Xu M,Ren L,Fan J,Huang L,Zhou L,Li X,Ye X
Tue Feb 01 00:00:00 EST 2022
Biotransformation of 18β-Glycyrrhetinic Acid by Human Intestinal Fungus Aspergillus niger RG13B1 and the Potential Anti-Inflammatory Mechanism of Its Metabolites.
Journal of agricultural and food chemistry
Zhang M,Zhang J,Wang C,Yan JK,Yi J,Ning J,Huo XK,Yu ZL,Zhang BJ,Sun CP,Ma XC
Wed Dec 07 00:00:00 EST 2022
Effects of Infrared Combined with Methylcobalamin on the Vibratory Sensory Threshold and Nerve Conduction Velocity of the Lower Extremity in Patients with Diabetic Foot Treatment.
Disease markers
Wang Y,Lin S,Chen Z,Chen Q,Fu M
Fri Sep 09 00:00:00 EDT 2022
Gut-Flora-Dependent Metabolite Trimethylamine-N-Oxide Promotes Atherosclerosis-Associated Inflammation Responses by Indirect ROS Stimulation and Signaling Involving AMPK and SIRT1.
Nutrients
Zhou S,Xue J,Shan J,Hong Y,Zhu W,Nie Z,Zhang Y,Ji N,Luo X,Zhang T,Ma W
Mon Aug 15 00:00:00 EDT 2022
Targeting A-kinase anchoring protein 12 phosphorylation in hepatic stellate cells regulates liver injury and fibrosis in mouse models.
eLife
Ramani K,Mavila N,Abeynayake A,Tomasi ML,Wang J,Matsuda M,Seki E
Tue Oct 04 00:00:00 EDT 2022
LipoxinA4 analog BML-111 protects podocytes cultured in high-glucose medium against oxidative injury via activating Nrf2 pathway.
International immunopharmacology
Hao H,Xie F,Xu F,Wang Q,Wu Y,Zhang D
Sat Oct 01 00:00:00 EDT 2022
LipoxinA4 analog BML-111 protects podocytes cultured in high-glucose medium against oxidative injury via activating Nrf2 pathway.
International immunopharmacology
Hao H,Xie F,Xu F,Wang Q,Wu Y,Zhang D
Sat Oct 01 00:00:00 EDT 2022
Detection of the role of intestinal flora and tryptophan metabolism involved in antidepressant-like actions of crocetin based on a multi-omics approach.
Psychopharmacology
Lin S,Li Q,Xu Z,Chen Z,Tao Y,Tong Y,Wang T,Chen S,Wang P
Tue Nov 01 00:00:00 EDT 2022
Discovery of a novel megakaryopoiesis enhancer, ingenol, promoting thrombopoiesis through PI3K-Akt signaling independent of thrombopoietin.
Pharmacological research
Wang L,Zhang T,Liu S,Mo Q,Jiang N,Chen Q,Yang J,Han YW,Chen JP,Huang FH,Li H,Zhou J,Luo JS,Wu JM
Tue Mar 01 00:00:00 EST 2022
Corn oligopeptides inhibit Akt/NF-κB signaling pathway and inflammatory factors to ameliorate CCl4 -induced hepatic fibrosis in mice.
Journal of food biochemistry
Feng XW,Cheng QL,Fang L,Liu WY,Liu LW,Sun CQ,Lu ZH,Li GM,Gu RZ
Mon Aug 01 00:00:00 EDT 2022
CoQ10 Improves Myocardial Damage in Doxorubicin-Induced Heart Failure in C57BL/6 Mice.
Frontiers in bioscience (Landmark edition)
Pei Z,Ma L,Li Y,Yang J,Yang Q,Yao W,Li S
Mon Aug 15 00:00:00 EDT 2022
Oxybaphus himalaicus Mitigates Lipopolysaccharide-Induced Acute Kidney Injury by Inhibiting TLR4/MD2 Complex Formation.
Antioxidants (Basel, Switzerland)
Zhan H,Pu Q,Long X,Lu W,Wang G,Meng F,Liao Z,Lan X,Chen M
Tue Nov 22 00:00:00 EST 2022
Realgar Alleviated Neuroinflammation Induced by High Protein and High Calorie Diet in Rats via the Microbiota-Gut-Brain Axis.
Nutrients
Feng C,Li A,Yin C,Wang S,Jin W,Liu Y,Huo T,Jiang H
Fri Sep 23 00:00:00 EDT 2022
Realgar Alleviated Neuroinflammation Induced by High Protein and High Calorie Diet in Rats via the Microbiota-Gut-Brain Axis.
Nutrients
Feng C,Li A,Yin C,Wang S,Jin W,Liu Y,Huo T,Jiang H
Fri Sep 23 00:00:00 EDT 2022
Orai1-mediated store-operated Ca2+ entry promotes cervical cancer progression through IL-6 signaling.
Frontiers in molecular biosciences
Pan Y,Huang J,Liu K,Xie C,Chen H,Guo Z,Guo S,Chen Y
Wed Nov 02 00:00:00 EDT 2022
Injectable self-healing hydrogel with siRNA delivery property for sustained STING silencing and enhanced therapy of intervertebral disc degeneration.
Bioactive materials
Chen J,Zhu H,Zhu Y,Zhao C,Wang S,Zheng Y,Xie Z,Jin Y,Song H,Yang L,Zhang J,Dai J,Hu Z,Wang H
Tue Mar 01 00:00:00 EST 2022
Adrenomedullin ameliorates palmitic acid-induced insulin resistance through PI3K/Akt pathway in adipocytes.
Acta diabetologica
Dai HB,Wang HY,Wang FZ,Qian P,Gao Q,Zhou H,Zhou YB
Sun May 01 00:00:00 EDT 2022
Targeting Vascular endothelial growth factor A with soluble vascular endothelial growth factor receptor 1 ameliorates nerve injury-induced neuropathic pain.
Molecular pain
Peng Z,Yang F,Huang S,Tang Y,Wan L
Fri Apr 01 00:00:00 EDT 2022
Morin Inhibits Dox-Induced Vascular Inflammation By Regulating PTEN/AKT/NF-κB Pathway.
Inflammation
Yu J,Qi HL,Zhang H,Zhao ZY,Nie ZY
Thu Dec 01 00:00:00 EST 2022
Morin Inhibits Dox-Induced Vascular Inflammation By Regulating PTEN/AKT/NF-κB Pathway.
Inflammation
Yu J,Qi HL,Zhang H,Zhao ZY,Nie ZY
Thu Dec 01 00:00:00 EST 2022
Morin Inhibits Dox-Induced Vascular Inflammation By Regulating PTEN/AKT/NF-κB Pathway.
Inflammation
Yu J,Qi HL,Zhang H,Zhao ZY,Nie ZY
Thu Dec 01 00:00:00 EST 2022
The Clostridium Metabolite P-Cresol Sulfate Relieves Inflammation of Primary Biliary Cholangitis by Regulating Kupffer Cells.
Cells
Fu HY,Xu JM,Ai X,Dang FT,Tan X,Yu HY,Feng J,Yang WX,Ma HT,Tu RF,Gupta AK,Manandhar LK,Bao WM,Tang YM
Sat Nov 26 00:00:00 EST 2022
MiR-92a/KLF4/p110δ regulates titanium particles-induced macrophages inflammation and osteolysis.
Cell death discovery
Wen Z,Lin S,Li C,Ouyang Z,Chen Z,Li S,Huang Y,Luo W,Zheng Z,Guo P,Kuang M,Ding Y
Wed Apr 13 00:00:00 EDT 2022
Injectable hyaluronic acid hydrogel loaded with BMSC and NGF for traumatic brain injury treatment.
Materials today. Bio
Wang L,Zhang D,Ren Y,Guo S,Li J,Ma S,Yao M,Guan F
Sat Jan 01 00:00:00 EST 2022
Macrophage SCAP Contributes to Metaflammation and Lean NAFLD by Activating STING-NF-κB Signaling Pathway.
Cellular and molecular gastroenterology and hepatology
Huang X,Yao Y,Hou X,Wei L,Rao Y,Su Y,Zheng G,Ruan XZ,Li D,Chen Y
Mon Jun 20 00:00:00 EDT 2022
Apelin-13 reduces lipopolysaccharide-induced neuroinflammation and cognitive impairment via promoting glucocorticoid receptor expression and nuclear translocation.
Neuroscience letters
Hu S,Shen P,Chen B,Tian SW,You Y
Sun Sep 25 00:00:00 EDT 2022
Capsaicin ameliorates inflammation in a TRPV1-independent mechanism by inhibiting PKM2-LDHA-mediated Warburg effect in sepsis.
Cell chemical biology
Zhang Q,Luo P,Xia F,Tang H,Chen J,Zhang J,Liu D,Zhu Y,Liu Y,Gu L,Zheng L,Li Z,Yang F,Dai L,Liao F,Xu C,Wang J
Thu Aug 18 00:00:00 EDT 2022
Hypoxia-Activated Prodrugs with Dual COX-2/CA Inhibitory Effects on Attenuating Cardiac Inflammation under Hypoxia.
Journal of medicinal chemistry
Zhou W,Wang C,Liu Z,Gou S
Thu Oct 13 00:00:00 EDT 2022
Hypoxia-Activated Prodrugs with Dual COX-2/CA Inhibitory Effects on Attenuating Cardiac Inflammation under Hypoxia.
Journal of medicinal chemistry
Zhou W,Wang C,Liu Z,Gou S
Thu Oct 13 00:00:00 EDT 2022
Carbon tetrachloride exposure induces ovarian damage through oxidative stress and inflammatory mediated ovarian fibrosis.
Ecotoxicology and environmental safety
Xue L,Li X,Zhu X,Zhang J,Zhou S,Tang W,Chen D,Chen Y,Dai J,Wu M,Wu M,Wang S
Thu Sep 01 00:00:00 EDT 2022
Gastrodin ameliorates Concanavalin A-induced acute hepatitis via the IL6/JAK2/STAT3 pathway.
Immunopharmacology and immunotoxicology
Zhou Y,Chen J,Yao Z,Gu X
Thu Dec 01 00:00:00 EST 2022
Integration of clinical and transcriptomics reveals programming of the lipid metabolism in gastric cancer.
BMC cancer
Li Y,Zhao J,Chen R,Chen S,Xu Y,Cai W
Tue Sep 06 00:00:00 EDT 2022
Design, synthesis and evaluation of fused hybrids with acetylcholinesterase inhibiting and Nrf2 activating functions for Alzheimer's disease.
European journal of medicinal chemistry
Wang Y,Xiong B,Lin H,Li Q,Yang H,Qiao Y,Li Q,Xu Z,Lyu W,Qu W,Liu W,Chen Y,Feng F,Sun H
Thu Dec 15 00:00:00 EST 2022
METTL3 Attenuates Inflammation in Fusarium solani-Induced Keratitis via the PI3K/AKT Signaling Pathway.
Investigative ophthalmology & visual science
Huang L,Tang H,Hu J
Thu Sep 01 00:00:00 EDT 2022
Epithelial‑derived exosomes promote M2 macrophage polarization via Notch2/SOCS1 during mechanical ventilation.
International journal of molecular medicine
Wang Y,Xie W,Feng Y,Xu Z,He Y,Xiong Y,Chen L,Li X,Liu J,Liu G,Wu Q
Fri Jul 01 00:00:00 EDT 2022
Methionine strengthens anti-inflammation of rice protein via depressing NF-κB activation and stimulating Msr expression in rats fed cholesterol-enriched diets.
Food science and biotechnology
Wang Z,Liang M,Liu B,Yang L
Wed Jun 01 00:00:00 EDT 2022
RIPK3-Dependent Necroptosis Activates MCP-1-Mediated Inflammation in Mice after Intracerebral Hemorrhage.
Journal of stroke and cerebrovascular diseases : the official journal of National Stroke Association
Huang S,Hu W,Rao D,Wu X,Bai Q,Wang J,Chu Z,Xu Y
Sat Jan 01 00:00:00 EST 2022
Polyphenols from the Peels of Punica granatum L. and Their Bioactivity of Suppressing Lipopolysaccharide-Stimulated Inflammatory Cytokines and Mediators in RAW 264.7 Cells via Activating p38 MAPK and NF-κB Signaling Pathways.
Molecules (Basel, Switzerland)
Li HM,Kouye O,Yang DS,Zhang YQ,Ruan JY,Han LF,Zhang Y,Wang T
Wed Jul 20 00:00:00 EDT 2022
Desmethylbellidifolin Attenuates Dextran Sulfate Sodium-Induced Colitis: Impact on Intestinal Barrier, Intestinal Inflammation and Gut Microbiota.
Planta medica
Wu J,Wu Y,Chen Y,Liu M,Yu H,Zhang Y,Wang T
Wed Jun 01 00:00:00 EDT 2022
Atorvastatin Restores PPARα Inhibition of Lipid Metabolism Disorders by Downregulating miR-21 Expression to Improve Mitochondrial Function and Alleviate Diabetic Nephropathy Progression.
Frontiers in pharmacology
Xiang J,Zhang H,Zhou X,Wang D,Chen R,Tan W,Liang L,Shi M,Zhang F,Xiao Y,Zhou Y,Wang Y,Guo B
Tue Mar 01 00:00:00 EST 2022
MiR-146b-5p/TRAF6 axis is essential for Ginkgo biloba L. extract GBE to attenuate LPS-induced neuroinflammation.
Frontiers in pharmacology
Liu M,Peng Y,Che Y,Zhou M,Bai Y,Tang W,Huang S,Zhang B,Deng S,Wang C,Yu Z
Tue Sep 13 00:00:00 EDT 2022
Atractylenolide I Ameliorates Acetaminophen-Induced Acute Liver Injury via the TLR4/MAPKs/NF-κB Signaling Pathways.
Frontiers in pharmacology
Du Z,Ma Z,Lai S,Ding Q,Hu Z,Yang W,Qian Q,Zhu L,Dou X,Li S
Tue Feb 08 00:00:00 EST 2022
Sinapic Acid Alleviates Acute Pancreatitis in Association with Attenuation of Inflammation, Pyroptosis, and the AMPK/NF-[Formula: see text]B Signaling Pathway.
The American journal of Chinese medicine
Huang ZW,Tan P,Yi XK,Chen H,Sun B,Shi H,Mou ZQ,Cheng YL,Li TX,Li Q,Fu WG
Fri Nov 25 00:00:00 EST 2022
The RNF26/CBX7 axis modulates the TNF pathway to promote cell proliferation and regulate sensitivity to TKIs in ccRCC.
International journal of biological sciences
Liu W,Wang H,Jian C,Li W,Ye K,Ren J,Zhu L,Wang Y,Jin X,Yi L
Fri Apr 15 00:00:00 EDT 2022
A multifunctional black phosphorus-based adhesive patch intrinsically induces partial EMT for effective burn wound healing.
Biomaterials science
Wang X,Tang X,Li N,Sun Y,Yu R,Zhang J,Li D,Yang B,Sun H
Tue Dec 20 00:00:00 EST 2022
Inflammasome Inhibition Prevents Motor Deficit and Cerebellar Degeneration Induced by Chronic Methamphetamine Administration.
Frontiers in molecular neuroscience
Ding J,Shen L,Ye Y,Hu S,Ren Z,Liu T,Dai J,Li Z,Wang J,Luo Y,Zhang Q,Zhang X,Qi X,Huang J
Sat Jul 16 00:00:00 EDT 2022
Renoprotective activity of Ribes diacanthum pall (RDP) against inflammation in cisplatin-stimulated mice model and human renal tubular epithelial cells.
Journal of ethnopharmacology
Wu Y,Zhao L,Gu L,Tilyek A,Yu B,Chai C
Sun Jan 30 00:00:00 EST 2022
Polydatin Improves Sepsis-Associated Encephalopathy by Activating Sirt1 and Reducing p38 Phosphorylation.
The Journal of surgical research
Huang L,Chen J,Li X,Huang M,Liu J,Qin N,Zeng Z,Wang X,Li F,Yang H
Mon Aug 01 00:00:00 EDT 2022
Astaxanthin Activated the Nrf2/HO-1 Pathway to Enhance Autophagy and Inhibit Ferroptosis, Ameliorating Acetaminophen-Induced Liver Injury.
ACS applied materials & interfaces
Cai X,Hua S,Deng J,Du Z,Zhang D,Liu Z,Khan NU,Zhou M,Chen Z
Wed Sep 28 00:00:00 EDT 2022
CIDEC: A Potential Factor in Diabetic Vascular Inflammation.
Journal of vascular research
Song FQ,Zhou HM,Ma WX,Li YL,Hu BA,Shang YY,Wang ZH,Zhong M,Zhang W,Ti Y
Tue Apr 26 00:00:00 EDT 2022
Caulis Polygoni Multiflori Accelerates Megakaryopoiesis and Thrombopoiesis via Activating PI3K/Akt and MEK/ERK Signaling Pathways.
Pharmaceuticals (Basel, Switzerland)
Yang X,Wang L,Zeng J,Wu A,Qin M,Wen M,Zhang T,Chen W,Mei Q,Qin D,Yang J,Jiang Y,Wu J
Wed Sep 28 00:00:00 EDT 2022
Cathepsin C promotes breast cancer lung metastasis by modulating neutrophil infiltration and neutrophil extracellular trap formation.
Cancer cell
Xiao Y,Cong M,Li J,He D,Wu Q,Tian P,Wang Y,Yang S,Liang C,Liang Y,Wen J,Liu Y,Luo W,Lv X,He Y,Cheng DD,Zhou T,Zhao W,Zhang P,Zhang X,Xiao Y,Qian Y,Wang H,Gao Q,Yang QC,Yang Q,Hu G
Mon Mar 08 00:00:00 EST 2021
Interleukin 6 regulates the expression of programmed cell death ligand 1 in thyroid cancer.
Cancer science
Zhang GQ,Jiao Q,Shen CT,Song HJ,Zhang HZ,Qiu ZL,Luo QY
Mon Mar 01 00:00:00 EST 2021
Inflamm-Aging-Related Cytokines of IL-17 and IFN-γ Accelerate Osteoclastogenesis and Periodontal Destruction.
Journal of immunology research
Tan J,Dai A,Pan L,Zhang L,Wang Z,Ke T,Sun W,Wu Y,Ding PH,Chen L
Mon Jan 10 00:00:00 EST 2022
Renoprotective effects of levosimendan on acute kidney injury following cardiac arrest via anti-inflammation, anti-apoptosis, and ERK activation.
FEBS open bio
Tian L,Wang S,Zhao L,Lu X,Zhu C,Gong H,Yang W
Sun Aug 01 00:00:00 EDT 2021
Nr2e1 deficiency aggravates insulin resistance and chronic inflammation of visceral adipose tissues in a diet-induced obese mice model.
Life sciences
He G,Gu J,Wang H,Cheng S,Xiong Q,Ke M,Hu Y,Feng J,Song L,Liu Z,Xu Y
Sun Aug 01 00:00:00 EDT 2021
Fenofibrate Ameliorates Hepatic Ischemia/Reperfusion Injury in Mice: Involvements of Apoptosis, Autophagy, and PPAR-α Activation.
PPAR research
Zhang J,Cheng P,Dai W,Ji J,Wu L,Feng J,Wu J,Yu Q,Li J,Guo C
Sat Feb 20 00:00:00 EST 2021
Hydroxysafflor yellow A acutely attenuates blood-brain barrier permeability, oxidative stress, inflammation and apoptosis in traumatic brain injury in rats1.
Acta cirurgica brasileira
Xu J,Zhan T,Zheng W,Huang YK,Chen K,Zhang XH,Ren P,Huang X
Fri Jan 29 00:00:00 EST 2021
Vicenin-2 inhibits the Helicobacterium pylori infection associated gastric carcinogenic events through modulation of PI3K/AKT and Nrf2 signaling in GES-1 cells.
Journal of biochemical and molecular toxicology
Zhang Y,Sun J,Dong Y,Shen X,Zhang Z
Mon Mar 01 00:00:00 EST 2021
Ginsenoside Rh4 alleviates antibiotic-induced intestinal inflammation by regulating the TLR4-MyD88-MAPK pathway and gut microbiota composition.
Food & function
Bai X,Fu R,Duan Z,Liu Y,Zhu C,Fan D
Wed Apr 07 00:00:00 EDT 2021
Chronic Voluntary Alcohol Drinking Causes Anxiety-like Behavior, Thiamine Deficiency, and Brain Damage of Female Crossed High Alcohol Preferring Mice.
Frontiers in pharmacology
Xu H,Li H,Liu D,Wen W,Xu M,Frank JA,Chen J,Zhu H,Grahame NJ,Luo J
Sat Sep 25 00:00:00 EDT 2021
HIF1A-AS2 induces osimertinib resistance in lung adenocarcinoma patients by regulating the miR-146b-5p/IL-6/STAT3 axis.
Molecular therapy. Nucleic acids
Si J,Ma Y,Lv C,Hong Y,Tan H,Yang Y
Fri Dec 03 00:00:00 EST 2021
HIF1A-AS2 induces osimertinib resistance in lung adenocarcinoma patients by regulating the miR-146b-5p/IL-6/STAT3 axis.
Molecular therapy. Nucleic acids
Si J,Ma Y,Lv C,Hong Y,Tan H,Yang Y
Fri Dec 03 00:00:00 EST 2021
α-Cyperone (CYP) down-regulates NF-κB and MAPKs signaling, attenuating inflammation and extracellular matrix degradation in chondrocytes, to ameliorate osteoarthritis in mice.
Aging
Zhang H,Li S,Lu J,Jin J,Zhu G,Wang L,Yan Y,He L,Wang B,Wang X,Yu H
Thu Jul 08 00:00:00 EDT 2021
LncRNA Malat-1 From MSCs-Derived Extracellular Vesicles Suppresses Inflammation and Cartilage Degradation in Osteoarthritis.
Frontiers in bioengineering and biotechnology
Pan C,Huang W,Chen Q,Xu J,Yao G,Li B,Wu T,Yin C,Cheng X
Tue Jan 04 00:00:00 EST 2022
Catalytically active gold clusters with atomic precision for noninvasive early intervention of neurotrauma.
Journal of nanobiotechnology
Zhang Y,Sun S,Liu H,Ren Q,Hao W,Xin Q,Xu J,Wang H,Zhang XD
Wed Oct 13 00:00:00 EDT 2021
The role of TLR4/MyD88/NF-κB pathway in periodontitis-induced liver inflammation of rats.
Oral diseases
Yue Y,Liu X,Li Y,Xia B,Yu W
Sat May 01 00:00:00 EDT 2021
Si-Based Hydrogen-Producing Nanoagent Protects Fetuses From Miscarriage Caused by Mother-to-Child Transmission.
Frontiers in medical technology
Usui N,Togawa S,Sumi T,Kobayashi Y,Koyama Y,Nakamura Y,Kondo M,Shinoda K,Kobayashi H,Shimada S
Fri Jan 21 00:00:00 EST 2022
Cartilage endplate stem cells inhibit intervertebral disc degeneration by releasing exosomes to nucleus pulposus cells to activate Akt/autophagy.
Stem cells (Dayton, Ohio)
Luo L,Jian X,Sun H,Qin J,Wang Y,Zhang J,Shen Z,Yang D,Li C,Zhao P,Liu M,Tian Z,Zhou Y
Thu Apr 01 00:00:00 EDT 2021
Protective mechanisms of Agrimonia pilosa Ledeb in dextran sodium sulfate-induced colitis as determined by a network pharmacology approach.
Acta biochimica et biophysica Sinica
Li C,Wang M,Sui J,Zhou Y,Chen W
Tue Oct 12 00:00:00 EDT 2021
Cancer-associated fibroblasts induce monocytic myeloid-derived suppressor cell generation via IL-6/exosomal miR-21-activated STAT3 signaling to promote cisplatin resistance in esophageal squamous cell carcinoma.
Cancer letters
Zhao Q,Huang L,Qin G,Qiao Y,Ren F,Shen C,Wang S,Liu S,Lian J,Wang D,Yu W,Zhang Y
Sun Oct 10 00:00:00 EDT 2021
Intestinal Klebsiella pneumoniae infection enhances susceptibility to epileptic seizure which can be reduced by microglia activation.
Cell death discovery
Lin P,Lin A,Tao K,Yang M,Ye Q,Chen H,Chen Y,Ma Y,Lin Z,He M,Wang X,Tian X
Wed Jul 07 00:00:00 EDT 2021
Controlled release of KGF-2 for regulation of wound healing by KGF-2 complexed with "lotus seedpod surface-like" porous microspheres.
Journal of materials chemistry. B
Pan H,Shi C,Yang R,Xi G,Lu C,Yang X,Chen J,Wang X,Chen L,Pan J
Wed May 19 00:00:00 EDT 2021
Cordyceps acid alleviates lung cancer in nude mice.
Journal of biochemical and molecular toxicology
Wang J,Chen H,Li W,Shan L
Mon Mar 01 00:00:00 EST 2021
Regulatory T cells promote glioma cell stemness through TGF-β-NF-κB-IL6-STAT3 signaling.
Cancer immunology, immunotherapy : CII
Liu S,Zhang C,Wang B,Zhang H,Qin G,Li C,Cao L,Gao Q,Ping Y,Zhang K,Lian J,Zhao Q,Wang D,Zhang Z,Zhao X,Yang L,Huang L,Yang B,Zhang Y
Wed Sep 01 00:00:00 EDT 2021
Estrogen-Related Receptor γ Agonist DY131 Ameliorates Lipopolysaccharide-Induced Acute Liver Injury.
Frontiers in pharmacology
Ma H,Liu J,Du Y,Zhang S,Cao W,Jia Z,Gong W,Zhang A
Thu May 13 00:00:00 EDT 2021
Gancaonin N from Glycyrrhiza uralensis Attenuates the Inflammatory Response by Downregulating the NF-κB/MAPK Pathway on an Acute Pneumonia In Vitro Model.
Pharmaceutics
Ko HM,Lee SH,Jee W,Jung JH,Kim KI,Jung HJ,Jang HJ
Tue Jul 06 00:00:00 EDT 2021
Alleviation of acute pancreatitis-associated lung injury by inhibiting the p38 mitogen-activated protein kinase pathway in pulmonary microvascular endothelial cells.
World journal of gastroenterology
Zhang XX,Wang HY,Yang XF,Lin ZQ,Shi N,Chen CJ,Yao LB,Yang XM,Guo J,Xia Q,Xue P
Fri May 14 00:00:00 EDT 2021
Visfatin regulates Pg LPS-induced proinflammatory/prodegradative effects in healthy and inflammatory periodontal cells partially via NF-κB pathway.
Biochimica et biophysica acta. Molecular cell research
Yao S,Jiang C,Zhang H,Gao X,Guo Y,Cao Z
Thu Jul 01 00:00:00 EDT 2021
Suprabasin-derived bioactive peptides identified by plasma peptidomics.
Scientific reports
Taguchi T,Kodera Y,Oba K,Saito T,Nakagawa Y,Kawashima Y,Shichiri M
Wed Jan 13 00:00:00 EST 2021
Urinary exosomes-based Engineered Nanovectors for Homologously Targeted Chemo-Chemodynamic Prostate Cancer Therapy via abrogating EGFR/AKT/NF-kB/IkB signaling.
Biomaterials
Pan S,Zhang Y,Huang M,Deng Z,Zhang A,Pei L,Wang L,Zhao W,Ma L,Zhang Q,Cui D
Sun Aug 01 00:00:00 EDT 2021
Prefrontal dopamine D1 receptor manipulation influences anxiety behavior and induces neuroinflammation within the hippocampus.
International journal of bipolar disorders
Beyer DKE,Mattukat A,Freund N
Mon Mar 08 00:00:00 EST 2021
The cardioprotective effect of the sodium-glucose cotransporter 2 inhibitor dapagliflozin in rats with isoproterenol-induced cardiomyopathy.
American journal of translational research
Wang FZ,Wei WB,Li X,Huo JY,Jiang WY,Wang HY,Qian P,Li ZZ,Zhou YB
Sat Oct 16 00:00:00 EDT 2021
Maternal immune activation alters adult behavior, intestinal integrity, gut microbiota and the gut inflammation.
Brain and behavior
Li W,Chen M,Feng X,Song M,Shao M,Yang Y,Zhang L,Liu Q,Lv L,Su X
Sat May 01 00:00:00 EDT 2021
Dual-enzymatically cross-linked gelatin hydrogel promotes neural differentiation and neurotrophin secretion of bone marrow-derived mesenchymal stem cells for treatment of moderate traumatic brain injury.
International journal of biological macromolecules
Li J,Zhang D,Guo S,Zhao C,Wang L,Ma S,Guan F,Yao M
Thu Sep 30 00:00:00 EDT 2021
JQ1 as a BRD4 Inhibitor Blocks Inflammatory Pyroptosis-Related Acute Colon Injury Induced by LPS.
Frontiers in immunology
Chen L,Zhong X,Cao W,Mao M,Li W,Yang H,Li M,Shi M,Zhang Y,Deng Y,Zu X,Liu J
Fri Jul 02 00:00:00 EDT 2021
Shenyan Kangfu tablet alleviates diabetic kidney disease through attenuating inflammation and modulating the gut microbiota.
Journal of natural medicines
Chen Q,Ren D,Wu J,Yu H,Chen X,Wang J,Zhang Y,Liu M,Wang T
Fri Jan 01 00:00:00 EST 2021
Clinical significance of sarcoid-like reaction in lymph nodes of gastric cancer patients.
Journal of surgical oncology
Itai Y,Ueda Y,Etoh T,Shirasaka Y,Shiroshita H,Shiraishi N,Daa T,Inomata M
Wed Sep 01 00:00:00 EDT 2021
Melatonin alleviates LPS-induced endoplasmic reticulum stress and inflammation in spermatogonial stem cells.
Journal of cellular physiology
Yang D,Wei Y,Lu Q,Qin D,Zhang M,Du X,Xu W,Yu X,He C,Li N,Peng S,Li G,Hua J
Sat May 01 00:00:00 EDT 2021
Gasdermin D Protects Mouse Podocytes Against High-Glucose-Induced Inflammation and Apoptosis via the C-Jun N-Terminal Kinase (JNK) Pathway.
Medical science monitor : international medical journal of experimental and clinical research
Li H,Zhao K,Li Y
Tue Mar 09 00:00:00 EST 2021
Enriched environment mitigates depressive behavior by changing the inflammatory activation phenotype of microglia in the hippocampus of depression model rats.
Brain research bulletin
Gu JY,Xu YW,Feng LP,Dong J,Zhao LQ,Liu C,Wang HY,Zhang XY,Song C,Wang CH
Wed Dec 01 00:00:00 EST 2021
Endogenous hydrogen sulfide improves vascular remodeling through PPARδ/SOCS3 signaling.
Journal of advanced research
Tian D,Teng X,Jin S,Chen Y,Xue H,Xiao L,Wu Y
Fri Jan 01 00:00:00 EST 2021
Anti-NAFLD Effect of Djulis Hull and Its Major Compound, Rutin, in Mice with High-Fat Diet (HFD)-Induced Obesity.
Antioxidants (Basel, Switzerland)
Tung YT,Zeng JL,Ho ST,Xu JW,Li S,Wu JH
Wed Oct 27 00:00:00 EDT 2021
miR-128 participates in the pathogenesis of chronic constipation by regulating the p38α/M-CSF inflammatory signaling pathway.
American journal of physiology. Gastrointestinal and liver physiology
Hong Y,Ren X,Liu W,Sun K,Chen B,Liu B,Yu X,Chen Q,Qian Q,Xie X,Jiang C
Fri Oct 01 00:00:00 EDT 2021
Critical roles of sphingosine kinase 1 in the regulation of neuroinflammation and neuronal injury after spinal cord injury.
Journal of neuroinflammation
Wang C,Xu T,Lachance BB,Zhong X,Shen G,Xu T,Tang C,Jia X
Thu Feb 18 00:00:00 EST 2021
Clinical and genomic characteristics of metabolic syndrome in colorectal cancer.
Aging
Li Y,Zhao J,Wu X,Zhang Y,Jin Y,Cai W
Thu Feb 11 00:00:00 EST 2021
Clinical and genomic characteristics of metabolic syndrome in colorectal cancer.
Aging
Li Y,Zhao J,Wu X,Zhang Y,Jin Y,Cai W
Thu Feb 11 00:00:00 EST 2021
Interleukin-6 as an enhancer of anti-angiogenic therapy for ovarian clear cell carcinoma.
Scientific reports
Seki T,Yanaihara N,Shapiro JS,Saito M,Tabata J,Yokomizo R,Noguchi D,Kuroda T,Kawabata A,Suzuki J,Takahashi K,Matsuzawa H,Miyake M,Takenaka M,Iida Y,Yanagida S,Okamoto A
Thu Apr 08 00:00:00 EDT 2021
H2O2 gel bleaching induces cytotoxicity and pain conduction in dental pulp stem cells via intracellular reactive oxygen species on enamel/dentin disc.
PloS one
Chen C,Huang X,Zhu W,Ding C,Huang P,Li R
Mon Nov 22 00:00:00 EST 2021
Long-term ketamine administration induces bladder damage and upregulates autophagy-associated proteins in bladder smooth muscle tissue.
Environmental toxicology
Li Y,Dong Z,Wen G,Ren X,Ren W,Yan L,Wang X,Yu H,Wu X,Xia X,Lu Y,Wu X
Wed Dec 01 00:00:00 EST 2021
Sanweidoukou decoction, a Chinese herbal formula, ameliorates β-amyloid protein-induced neuronal insult via modulating MAPK/NF-κB signaling pathways: Studies in vivo and in vitro.
Journal of ethnopharmacology
An FM,Liu Z,Xuan XR,Liu QS,Wei CX
Sat Jun 12 00:00:00 EDT 2021
Mitochondrial ferritin attenuates cerebral ischaemia/reperfusion injury by inhibiting ferroptosis.
Cell death & disease
Wang P,Cui Y,Ren Q,Yan B,Zhao Y,Yu P,Gao G,Shi H,Chang S,Chang YZ
Wed May 05 00:00:00 EDT 2021
Study on Protection of Human Umbilical Vein Endothelial Cells from Amiodarone-Induced Damage by Intermedin through Activation of Wnt/β-Catenin Signaling Pathway.
Oxidative medicine and cellular longevity
Wang Y,Wang J,Yang J,Kang J,Xue F,Chang S,Ji H,Zang H,Zhou X,Wang G,Fan W,Yan X,Guo J,Ren X,Tian J
Wed Jan 19 00:00:00 EST 2022
Mesenchymal stem cells ameliorate renal fibrosis by galectin-3/Akt/GSK3β/Snail signaling pathway in adenine-induced nephropathy rat.
Stem cell research & therapy
Tang H,Zhang P,Zeng L,Zhao Y,Xie L,Chen B
Fri Jul 16 00:00:00 EDT 2021
WITHDRAWN: Immunohistochemical study of inflammatory responses in septa arising from type I intestinal atresia.
Histology and histopathology
Liu X,Hao P,Xie X,Li Y,Li S,Song Y,Feng C,Ye M,Wei Y,Zhao P,Li L,Jin ZW
Tue Jul 27 00:00:00 EDT 2021
Evodiamine inhibits high-fat diet-induced colitis-associated cancer in mice through regulating the gut microbiota.
Journal of integrative medicine
Zhu LQ,Zhang L,Zhang J,Chang GL,Liu G,Yu DD,Yu XM,Zhao MS,Ye B
Fri Jan 01 00:00:00 EST 2021
Maiwei Yangfei decoction prevents bleomycin-induced pulmonary fibrosis in mice.
Experimental and therapeutic medicine
Xu Y,Peng W,Han D,Feng F,Wang Z,Gu C,Zhou X,Wu Q
Mon Nov 01 00:00:00 EDT 2021
Diosmetin Ameliorates Nonalcoholic Steatohepatitis through Modulating Lipogenesis and Inflammatory Response in a STAT1/CXCL10-Dependent Manner.
Journal of agricultural and food chemistry
Luo N,Yang C,Zhu Y,Chen Q,Zhang B
Wed Jan 20 00:00:00 EST 2021
Trophoblast-derived IL-6 serves as an important factor for normal pregnancy by activating Stat3-mediated M2 macrophages polarization.
International immunopharmacology
Ding J,Yang C,Cheng Y,Wang J,Zhang S,Yan S,He F,Yin T,Yang J
Fri Jan 01 00:00:00 EST 2021
METTL3/N6-methyladenosine/ miR-21-5p promotes obstructive renal fibrosis by regulating inflammation through SPRY1/ERK/NF-κB pathway activation.
Journal of cellular and molecular medicine
Liu E,Lv L,Zhan Y,Ma Y,Feng J,He Y,Wen Y,Zhang Y,Pu Q,Ji F,Yang X,Wen JG
Sun Aug 01 00:00:00 EDT 2021
METTL3/N6-methyladenosine/ miR-21-5p promotes obstructive renal fibrosis by regulating inflammation through SPRY1/ERK/NF-κB pathway activation.
Journal of cellular and molecular medicine
Liu E,Lv L,Zhan Y,Ma Y,Feng J,He Y,Wen Y,Zhang Y,Pu Q,Ji F,Yang X,Wen JG
Sun Aug 01 00:00:00 EDT 2021
Polyphenol-Rich Extract from Litchi chinensis Seeds Alleviates Hypertension-Induced Renal Damage in Rats.
Journal of agricultural and food chemistry
Yao Y,Liu T,Yin L,Man S,Ye S,Ma L
Wed Feb 24 00:00:00 EST 2021
Mild hyperthermia promotes immune checkpoint blockade-based immunotherapy against metastatic pancreatic cancer using size-adjustable nanoparticles.
Acta biomaterialia
Yu Q,Tang X,Zhao W,Qiu Y,He J,Wan D,Li J,Wang X,He X,Liu Y,Li M,Zhang Z,He Q
Fri Oct 01 00:00:00 EDT 2021
Elevated branched-chain α-keto acids exacerbate macrophage oxidative stress and chronic inflammatory damage in type 2 diabetes mellitus.
Free radical biology & medicine
Liu S,Li L,Lou P,Zhao M,Wang Y,Tang M,Gong M,Liao G,Yuan Y,Li L,Zhang J,Chen Y,Cheng J,Lu Y,Liu J
Mon Nov 01 00:00:00 EDT 2021
Elevated branched-chain α-keto acids exacerbate macrophage oxidative stress and chronic inflammatory damage in type 2 diabetes mellitus.
Free radical biology & medicine
Liu S,Li L,Lou P,Zhao M,Wang Y,Tang M,Gong M,Liao G,Yuan Y,Li L,Zhang J,Chen Y,Cheng J,Lu Y,Liu J
Mon Nov 01 00:00:00 EDT 2021
Lack of the peroxiredoxin 6 gene causes impaired spatial memory and abnormal synaptic plasticity.
Molecular brain
Phasuk S,Jasmin S,Pairojana T,Chang HK,Liang KC,Liu IY
Mon Apr 19 00:00:00 EDT 2021
Role of Aberrantly Activated Lysophosphatidic Acid Receptor 1 Signaling Mediated Inflammation in Renal Aging.
Cells
Jin Y,Kim EN,Lim JH,Kim HD,Ban TH,Yang CW,Park CW,Choi BS
Tue Sep 28 00:00:00 EDT 2021
Periodontitis Deteriorates Cognitive Function and Impairs Neurons and Glia in a Mouse Model of Alzheimer's Disease.
Journal of Alzheimer's disease : JAD
Qian X,Zhang S,Duan L,Yang F,Zhang K,Yan F,Ge S
Wed Sep 15 00:00:00 EDT 2021
lncRNA Gm16410 Mediates PM2.5-Induced Macrophage Activation via PI3K/AKT Pathway.
Frontiers in cell and developmental biology
Xu J,Xu H,Ma K,Wang Y,Niu B,Zhang L,Li F
Sat Apr 03 00:00:00 EDT 2021
Hepatoprotective effects of ginsenoside Rk3 in acetaminophen-induced liver injury in mice by activation of autophagy.
Food & function
Qu L,Fu R,Ma X,Fan D
Mon Oct 04 00:00:00 EDT 2021
Effects of carotid baroreceptor stimulation on aortic remodeling in obese rats.
Nutrition, metabolism, and cardiovascular diseases : NMCD
Yu Q,Shu L,Wang L,Gao K,Wang J,Dai M,Cao Q,Zhang Y,Luo Q,Hu B,Dai D,Chen J,Bao M
Thu May 06 00:00:00 EDT 2021
Low-intensity pulsed ultrasound ameliorates angiotensin II-induced cardiac fibrosis by alleviating inflammation via a caveolin-1-dependent pathway.
Journal of Zhejiang University. Science. B
Zhao K,Zhang J,Xu T,Yang C,Weng L,Wu T,Wu X,Miao J,Guo X,Tu J,Zhang D,Zhou B,Sun W,Kong X
Fri Oct 15 00:00:00 EDT 2021
Cynaroside prevents macrophage polarization into pro-inflammatory phenotype and alleviates cecal ligation and puncture-induced liver injury by targeting PKM2/HIF-1α axis.
Fitoterapia
Pei L,Le Y,Chen H,Feng J,Liu Z,Zhu J,Wang C,Chen L,Dou X,Lu D
Thu Jul 01 00:00:00 EDT 2021
Cynaroside prevents macrophage polarization into pro-inflammatory phenotype and alleviates cecal ligation and puncture-induced liver injury by targeting PKM2/HIF-1α axis.
Fitoterapia
Pei L,Le Y,Chen H,Feng J,Liu Z,Zhu J,Wang C,Chen L,Dou X,Lu D
Thu Jul 01 00:00:00 EDT 2021
Shunaoxin pills improve the antihypertensive effect of nifedipine and alleviate its renal lipotoxicity in spontaneous hypertension rats.
Environmental toxicology
Liu X,Man S,Luo C,Liu Y,Liu Y,Liu C,Gao W
Mon Mar 01 00:00:00 EST 2021
Curcumin inhibits the growth of liver cancer by impairing myeloid-derived suppressor cells in murine tumor tissues.
Oncology letters
Tian S,Liao L,Zhou Q,Huang X,Zheng P,Guo Y,Deng T,Tian X
Thu Apr 01 00:00:00 EDT 2021
Dopamine receptor D2 inhibition alleviates diabetic hepatic stellate cells fibrosis by regulating the TGF-β1/Smads and NFκB pathways.
Clinical and experimental pharmacology & physiology
Zhao B,Li S,Guo Z,Chen Z,Zhang X,Xu C,Chen J,Wei C
Mon Mar 01 00:00:00 EST 2021
Application of imiquimod-induced murine psoriasis model in evaluating interleukin-17A antagonist.
BMC immunology
Li Q,Liu W,Gao S,Mao Y,Xin Y
Thu Jan 28 00:00:00 EST 2021
Identification of collagen genes related to immune infiltration and epithelial-mesenchymal transition in glioma.
Cancer cell international
Yin W,Zhu H,Tan J,Xin Z,Zhou Q,Cao Y,Wu Z,Wang L,Zhao M,Jiang X,Ren C,Tang G
Tue May 25 00:00:00 EDT 2021
PLOD2-driven IL-6/STAT3 signaling promotes the invasion and metastasis of oral squamous cell carcinoma via activation of integrin β1.
International journal of oncology
Saito K,Mitsui A,Sumardika IW,Yokoyama Y,Sakaguchi M,Kondo E
Tue Jun 01 00:00:00 EDT 2021
PLOD2-driven IL-6/STAT3 signaling promotes the invasion and metastasis of oral squamous cell carcinoma via activation of integrin β1.
International journal of oncology
Saito K,Mitsui A,Sumardika IW,Yokoyama Y,Sakaguchi M,Kondo E
Tue Jun 01 00:00:00 EDT 2021
Inhibition of sEH via stabilizing the level of EETs alleviated Alzheimer's disease through GSK3β signaling pathway.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Sun CP,Zhang XY,Zhou JJ,Huo XK,Yu ZL,Morisseau C,Hammock BD,Ma XC
Fri Oct 01 00:00:00 EDT 2021
Tear dynamics testing and quantitative proteomics analysis in patients with chronic renal failure.
Journal of proteomics
Zong RR,Zhu FF,Han W,Wang YX,Wang GL,Wang YZ,Mao YB,Guan TJ,Liu ZG,Xue YH,Li C
Thu Sep 30 00:00:00 EDT 2021
Effect of dihydromyricetin on hepatic encephalopathy associated with acute hepatic failure in mice.
Pharmaceutical biology
Cheng L,Wang X,Ma X,Xu H,Yang Y,Zhang D
Wed Dec 01 00:00:00 EST 2021
H2S improves doxorubicin-induced myocardial fibrosis by inhibiting oxidative stress and apoptosis via Keap1-Nrf2.
Technology and health care : official journal of the European Society for Engineering and Medicine
Li Y,Chandra TP,Song X,Nie L,Liu M,Yi J,Zheng X,Chu C,Yang J
Wed Aug 18 00:00:00 EDT 2021
Ameliorative effects of miR-423-5p against polarization of microglial cells of the M1 phenotype by targeting a NLRP3 inflammasome signaling pathway.
International immunopharmacology
Cheng J,Hao J,Jiang X,Ji J,Wu T,Chen X,Zhang F
Fri Oct 01 00:00:00 EDT 2021
ZBTB20-mediated titanium particle-induced peri-implant osteolysis by promoting macrophage inflammatory responses.
Biomaterials science
Qiu J,Peng P,Xin M,Wen Z,Chen Z,Lin S,Kuang M,Fu Y,Fang G,Li S,Li C,Mao J,Qin L,Ding Y
Sun Jun 07 00:00:00 EDT 2020
ZBTB20-mediated titanium particle-induced peri-implant osteolysis by promoting macrophage inflammatory responses.
Biomaterials science
Qiu J,Peng P,Xin M,Wen Z,Chen Z,Lin S,Kuang M,Fu Y,Fang G,Li S,Li C,Mao J,Qin L,Ding Y
Sun Jun 07 00:00:00 EDT 2020
Senolytic Agent Navitoclax Inhibits Angiotensin II-Induced Heart Failure in Mice.
Journal of cardiovascular pharmacology
Jia K,Dai Y,Liu A,Li X,Wu L,Lu L,Bao Y,Jin Q
Thu Oct 01 00:00:00 EDT 2020
Pulp stem cells with hepatocyte growth factor overexpression exhibit dual effects in rheumatoid arthritis.
Stem cell research & therapy
Dong X,Kong F,Liu C,Dai S,Zhang Y,Xiao F,Zhang H,Wu CT,Wang H
Wed Jun 10 00:00:00 EDT 2020
Qingda granules attenuate hypertensive cardiac remodeling and inflammation in spontaneously hypertensive rats.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Wu X,Shen A,Bao L,Wu M,Lin X,Wang H,Chen Y,Cai Q,Lin S,Zhou X,Lu Y,Peng M,Chen D,Chu J,Peng J
Tue Sep 01 00:00:00 EDT 2020
CX3CR1-microglia mediates neuroinflammation and blood pressure regulation in the nucleus tractus solitarii of fructose-induced hypertensive rats.
Journal of neuroinflammation
Ho CY,Lin YT,Chen HH,Ho WY,Sun GC,Hsiao M,Lu PJ,Cheng PW,Tseng CJ
Fri Jun 12 00:00:00 EDT 2020
Protective effects of isorhamnetin on pulmonary arterial hypertension: in vivo and in vitro studies.
Phytotherapy research : PTR
Chang Z,Wang JL,Jing ZC,Ma P,Xu QB,Na JR,Tian J,Ma X,Zhou W,Zhou R
Thu Oct 01 00:00:00 EDT 2020
The Chinese medicinal herb decoction QRZSLXF enhances anti-inflammatory effect in TNBS-induced colitis via balancing Th17/Tregs differentiation.
Journal of ethnopharmacology
Zhang M,Fan H,Tan S,Tang Q,Liu X,Zuo D,Liao Y,Nan Z,Tan C
Mon Apr 06 00:00:00 EDT 2020
Didymin ameliorates dexamethasone-induced non-alcoholic fatty liver disease by inhibiting TLR4/NF-κB and PI3K/Akt pathways in C57BL/6J mice.
International immunopharmacology
Feng Z,Pang L,Chen S,Pang X,Huang Y,Qiao Q,Wang Y,Vonglorkham S,Huang Q,Lin X,Wei J
Sun Nov 01 00:00:00 EDT 2020
WJ-39, an Aldose Reductase Inhibitor, Ameliorates Renal Lesions in Diabetic Nephropathy by Activating Nrf2 Signaling.
Oxidative medicine and cellular longevity
Zhou X,Liu Z,Ying K,Wang H,Liu P,Ji X,Chi T,Zou L,Wang S,He Z
Wed Jan 27 00:00:00 EST 2021
Dihydroartemisinin prevents dextran sodium sulphate-induced colitisthrough inhibition of the activation of NLRP3 inflammasome and p38 MAPK signaling.
International immunopharmacology
Liang R,Chen W,Fan H,Chen X,Zhang J,Zhu JS
Sun Nov 01 00:00:00 EDT 2020
LncRNA GUARDIN suppresses cellular senescence through a LRP130-PGC1α-FOXO4-p21-dependent signaling axis.
EMBO reports
Sun X,Thorne RF,Zhang XD,He M,Li J,Feng S,Liu X,Wu M
Fri Apr 03 00:00:00 EDT 2020
Sevoflurane Protects against Intestinal Ischemia-Reperfusion Injury by Activating Peroxisome Proliferator-Activated Receptor Gamma/Nuclear Factor-κB Pathway in Rats.
Pharmacology
Liu C,Ding R,Huang W,Miao L,Li J,Li Y
Tue Oct 27 00:00:00 EDT 2020
Crocin attenuates isoprenaline-induced myocardial fibrosis by targeting TLR4/NF-κB signaling: connecting oxidative stress, inflammation, and apoptosis.
Naunyn-Schmiedeberg's archives of pharmacology
Jin W,Zhang Y,Xue Y,Han X,Zhang X,Ma Z,Sun S,Chu X,Cheng J,Guan S,Li Z,Chu L
Wed Jan 01 00:00:00 EST 2020
Maresin 1 mitigates concanavalin A-induced acute liver injury in mice by inhibiting ROS-mediated activation of NF-κB signaling.
Free radical biology & medicine
Zhang P,Yin Y,Wang T,Li W,Li C,Zeng X,Yang W,Zhang R,Tang Y,Shi L,Li R,Tao K
Sat Feb 01 00:00:00 EST 2020
Echinacoside attenuates inflammatory response in a rat model of cervical spondylotic myelopathy via inhibition of excessive mitochondrial fission.
Free radical biology & medicine
Zhou L,Yao M,Tian Z,Song Y,Sun Y,Ye J,Li G,Sng KS,Xu L,Cui X,Wang Y
Wed May 20 00:00:00 EDT 2020
Preclinical Evaluation of the Novel Small-Molecule MSI-N1014 for Treating Drug-Resistant Colon Cancer via the LGR5/β-catenin/miR-142-3p Network and Reducing Cancer-Associated Fibroblast Transformation.
Cancers
Yadav VK,Huang YJ,George TA,Wei PL,Sumitra MR,Ho CL,Chang TH,Wu ATH,Huang HS
Tue Jun 16 00:00:00 EDT 2020
TLR4 knockout upregulates the expression of Mfn2 and PGC-1α in a high-fat diet and ischemia-reperfusion mice model of liver injury.
Life sciences
Zhang C,Jia Y,Liu B,Wang G,Zhang Y
Sat Aug 01 00:00:00 EDT 2020
Tannic acid ameliorates arsenic trioxide-induced nephrotoxicity, contribution of NF-κB and Nrf2 pathways.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Jin W,Xue Y,Xue Y,Han X,Song Q,Zhang J,Li Z,Cheng J,Guan S,Sun S,Chu L
Mon Jun 01 00:00:00 EDT 2020
A Novel Antidiabetic Monomers Combination Alleviates Insulin Resistance Through Bacteria-Cometabolism-Inflammation Responses.
Frontiers in microbiology
Han L,Zhao LH,Zhang ML,Li HT,Gao ZZ,Zheng XJ,Wang XM,Wu HR,Zheng YJ,Jiang XT,Ding QY,Yang HY,Jia WP,Tong XL
Mon Sep 28 00:00:00 EDT 2020
Adipose-derived exosomes protect the pulmonary endothelial barrier in ventilator-induced lung injury by inhibiting the TRPV4/Ca2+ signaling pathway.
American journal of physiology. Lung cellular and molecular physiology
Yu Q,Wang D,Wen X,Tang X,Qi D,He J,Zhao Y,Deng W,Zhu T
Wed Apr 01 00:00:00 EDT 2020
Dendrobium Alkaloids Promote Neural Function After Cerebral Ischemia-Reperfusion Injury Through Inhibiting Pyroptosis Induced Neuronal Death in both In Vivo and In Vitro Models.
Neurochemical research
Liu D,Dong Z,Xiang F,Liu H,Wang Y,Wang Q,Rao J
Sat Feb 01 00:00:00 EST 2020
KGF-2 and FGF-21 poloxamer 407 hydrogel coordinates inflammation and proliferation homeostasis to enhance wound repair of scalded skin in diabetic rats.
BMJ open diabetes research & care
Yang X,Yang R,Chen M,Zhou Q,Zheng Y,Lu C,Bi J,Sun W,Huang T,Li L,Gong J,Li X,Hui Q,Wang X
Fri May 01 00:00:00 EDT 2020
KGF-2 and FGF-21 poloxamer 407 hydrogel coordinates inflammation and proliferation homeostasis to enhance wound repair of scalded skin in diabetic rats.
BMJ open diabetes research & care
Yang X,Yang R,Chen M,Zhou Q,Zheng Y,Lu C,Bi J,Sun W,Huang T,Li L,Gong J,Li X,Hui Q,Wang X
Fri May 01 00:00:00 EDT 2020
Chronic acrylamide exposure induced glia cell activation, NLRP3 infl-ammasome upregulation and cognitive impairment.
Toxicology and applied pharmacology
Liu Y,Zhang X,Yan D,Wang Y,Wang N,Liu Y,Tan A,Chen X,Yan H
Wed Apr 15 00:00:00 EDT 2020
Critical Role of AdipoR1 in Regulating Th17 Cell Differentiation Through Modulation of HIF-1α-Dependent Glycolysis.
Frontiers in immunology
Zhang Q,Wang L,Jiang J,Lin S,Luo A,Zhao P,Tan W,Zhang M
Fri Apr 30 00:00:00 EDT 2021
MicroRNA-34a deficiency leads to impaired wound closure by augmented inflammation in mice.
Annals of translational medicine
Zhao N,Wang G,Long S,Hu M,Gao J,Ran X,Wang J,Su Y,Wang T
Wed Apr 01 00:00:00 EDT 2020
Methane-Rich Saline Alleviates CA/CPR Brain Injury by Inhibiting Oxidative Stress, Microglial Activation-Induced Inflammatory Responses, and ER Stress-Mediated Apoptosis.
Oxidative medicine and cellular longevity
Cui R,Liu S,Wang C,Liu T,Ren J,Jia Y,Tong Y,Liu C,Zhang J
Thu May 13 00:00:00 EDT 2021
C/EBPγ is a critical negative regulator of LPS-/IgG immune complex-induced acute lung injury through the downregulation of C/EBPβ-/C/EBPδ-dependent C/EBP transcription activation.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Yan C,Zhang L,Yang L,Zhang Q,Wang X
Thu Oct 01 00:00:00 EDT 2020
MicroRNA-675-3p regulates IL-1β-stimulated human chondrocyte apoptosis and cartilage degradation by targeting GNG5.
Biochemical and biophysical research communications
Shen XF,Cheng Y,Dong QR,Zheng MQ
Thu Jun 25 00:00:00 EDT 2020
ATP-Sensitive Potassium Channels Mediate the Cardioprotective Effect of Panax notoginseng Saponins against Myocardial Ischaemia-Reperfusion Injury and Inflammatory Reaction.
BioMed research international
Ning K,Jiang L,Hu T,Wang X,Liu A,Bao Y
Thu May 06 00:00:00 EDT 2021
Metamizole relieves pain by influencing cytokine levels in dorsal root ganglia in a rat model of neuropathic pain.
Pharmacological reports : PR
Zajaczkowska R,Kwiatkowski K,Pawlik K,Piotrowska A,Rojewska E,Makuch W,Wordliczek J,Mika J
Thu Oct 01 00:00:00 EDT 2020
Crocin protects against cardiotoxicity induced by doxorubicin through TLR-2/NF-κB signal pathway in vivo and vitro.
International immunopharmacology
Chu X,Zhang Y,Xue Y,Li Z,Shi J,Wang H,Chu L
Wed Jul 01 00:00:00 EDT 2020
Impact of chitosan membrane culture on the expression of pro- and anti-inflammatory cytokines in mesenchymal stem cells.
Experimental and therapeutic medicine
Yi P,Xu X,Qiu B,Li H
Thu Oct 01 00:00:00 EDT 2020
Supplementation of Inulin with Various Degree of Polymerization Ameliorates Liver Injury and Gut Microbiota Dysbiosis in High Fat-Fed Obese Mice.
Journal of agricultural and food chemistry
Du H,Zhao A,Wang Q,Yang X,Ren D
Wed Jan 22 00:00:00 EST 2020
Inhibition of interleukin-6 on matrix protein production by glomerular mesangial cells and the pathway involved.
American journal of physiology. Renal physiology
Chaudhari S,Yazdizadeh Shotorbani P,Tao Y,Davis ME,Mallet RT,Ma R
Mon Jun 01 00:00:00 EDT 2020
Mitochondrial Transplantation Attenuates Brain Dysfunction in Sepsis by Driving Microglial M2 Polarization.
Molecular neurobiology
Yan C,Ma Z,Ma H,Li Q,Zhai Q,Jiang T,Zhang Z,Wang Q
Tue Sep 01 00:00:00 EDT 2020
Pterostilbene Ameliorates Glycemic Control, Dyslipidemia and Liver Injury in Type 2 Diabetes Rats.
Biomedical and environmental sciences : BES
Zhang YJ,Sun HL,Wang T,Liu XX,Liu C,Shen F,Wang BY,Ding RR,Liu YM,Huang GY,Li WJ,Li X
Wed May 20 00:00:00 EDT 2020
The Effects of Sidt2 on the Inflammatory Pathway in Mouse Mesangial Cells.
Mediators of inflammation
Sun H,Ding JM,Zheng HH,Lv KJ,Hu YF,Luo YH,Wu X,Pei WJ,Wang LZ,Wu MC,Zhang Y,Gao JL
Fri Jul 09 00:00:00 EDT 2021
Pretreatment with the ALDH2 activator Alda‑1 protects rat livers from ischemia/reperfusion injury by inducing autophagy.
Molecular medicine reports
Liu Z,Ye S,Zhong X,Wang W,Lai CH,Yang W,Yue P,Luo J,Huang X,Zhong Z,Xiong Y,Fan X,Li L,Wang Y,Ye Q
Tue Sep 01 00:00:00 EDT 2020
HMGB1 Mediates Paraquat-Induced Neuroinflammatory Responses via Activating RAGE Signaling Pathway.
Neurotoxicity research
Huang M,Guo M,Wang K,Wu K,Li Y,Tian T,Wang Y,Yan W,Zhou Z,Yang H
Wed Apr 01 00:00:00 EDT 2020
Pachymic acid protects oocyte by improving the ovarian microenvironment in polycystic ovary syndrome mice†.
Biology of reproduction
Fu XP,Xu L,Fu BB,Wei KN,Liu Y,Liao BQ,He SW,Wang YL,Chen MH,Lin YH,Li FP,Hong ZW,Huang XH,Xu CL,Wang HL
Thu Oct 29 00:00:00 EDT 2020
Microglia polarization of hippocampus is involved in the mechanism of Apelin-13 ameliorating chronic water immersion restraint stress-induced depression-like behavior in rats.
Neuropeptides
Zhou S,Chen S,Xie W,Guo X,Zhao J
Mon Jun 01 00:00:00 EDT 2020
Sema7A, a brain immune regulator, regulates seizure activity in PTZ-kindled epileptic rats.
CNS neuroscience & therapeutics
Deng J,Xu T,Yang J,Zhang KM,Li Q,Yu XY,Li R,Fu J,Jiang Q,Ma JX,Chen YM
Wed Jan 01 00:00:00 EST 2020
Muscone suppresses inflammatory responses and neuronal damage in a rat model of cervical spondylotic myelopathy by regulating Drp1-dependent mitochondrial fission.
Journal of neurochemistry
Zhou LY,Yao M,Tian ZR,Liu SF,Song YJ,Ye J,Li G,Sun YL,Cui XJ,Wang YJ
Tue Sep 01 00:00:00 EDT 2020
IL-6 is present in beta and alpha cells in human pancreatic islets: Expression is reduced in subjects with type 1 diabetes.
Clinical immunology (Orlando, Fla.)
Rajendran S,Anquetil F,Quesada-Masachs E,Graef M,Gonzalez N,McArdle S,Chu T,Krogvold L,Dahl-Jørgensen K,von Herrath M
Sat Feb 01 00:00:00 EST 2020
IL-6 is present in beta and alpha cells in human pancreatic islets: Expression is reduced in subjects with type 1 diabetes.
Clinical immunology (Orlando, Fla.)
Rajendran S,Anquetil F,Quesada-Masachs E,Graef M,Gonzalez N,McArdle S,Chu T,Krogvold L,Dahl-Jørgensen K,von Herrath M
Sat Feb 01 00:00:00 EST 2020
Hic-5 deficiency attenuates hepatic ischemia reperfusion injury through TLR4/NF-κB signaling pathways.
Life sciences
Gao L,Qian B,Chen H,Wang A,Li Q,Li J,Tan P,Xia X,Du Y,Fu W
Fri May 15 00:00:00 EDT 2020
Maternal 25-Hydroxyvitamin D Deficiency Promoted Metabolic Syndrome and Downregulated Nrf2/CBR1 Pathway in Offspring.
Frontiers in pharmacology
Zheng J,Liu X,Zheng B,Zheng Z,Zhang H,Zheng J,Sun C,Chen H,Yang J,Wang Z,Lin M,Chen J,Zhou Q,Zheng Z,Xu X,Ying H
Mon Sep 28 00:00:00 EDT 2020
Regulation of JNK signaling pathway and RIPK3/AIF in necroptosis-mediated global cerebral ischemia/reperfusion injury in rats.
Experimental neurology
Hu W,Wu X,Yu D,Zhao L,Zhu X,Li X,Huang T,Chu Z,Xu Y
Tue Sep 01 00:00:00 EDT 2020
Androgen aggravates liver fibrosis by activation of NLRP3 inflammasome in CCl4-induced liver injury mouse model.
American journal of physiology. Endocrinology and metabolism
Ma X,Zhou Y,Qiao B,Jiang S,Shen Q,Han Y,Liu A,Chen X,Wei L,Zhou L,Zhao J
Fri May 01 00:00:00 EDT 2020
Akkermansia muciniphila promotes type H vessel formation and bone fracture healing by reducing gut permeability and inflammation.
Disease models & mechanisms
Liu JH,Yue T,Luo ZW,Cao J,Yan ZQ,Jin L,Wan TF,Shuai CJ,Wang ZG,Zhou Y,Xu R,Xie H
Tue Dec 01 00:00:00 EST 2020
Ameliorative effects and mechanism of crocetin in arsenic trioxide‑induced cardiotoxicity in rats.
Molecular medicine reports
Zhao Z,Li J,Zheng B,Liang Y,Shi J,Zhang J,Han X,Chu L,Chu X,Gao Y
Tue Dec 01 00:00:00 EST 2020
TSG-6 Inhibits Oxidative Stress and Induces M2 Polarization of Hepatic Macrophages in Mice With Alcoholic Hepatitis via Suppression of STAT3 Activation.
Frontiers in pharmacology
Wan YM,Wu HM,Li YH,Xu ZY,Yang JH,Liu C,He YF,Wang MJ,Wu XN,Zhang Y
Fri Nov 03 00:00:00 EDT 2023
A novel bile acid analog, A17, ameliorated non-alcoholic steatohepatitis in high-fat diet-fed hamsters.
Toxicology and applied pharmacology
Wang Y,Zhu Y,Niu J,Deng Q,Guo S,Jiang H,Peng Z,Xue Y,Peng H,Xuan L,Pan G
Thu Oct 01 00:00:00 EDT 2020
COMP in the Infrapatellar Fat Pad-Results of a Prospective Histological, Immunohistological, and Biochemical Case-Control Study.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society
Grevenstein D,Heilig J,Dargel J,Oppermann J,Eysel P,Brochhausen C,Niehoff A
Wed Apr 01 00:00:00 EDT 2020
RGS12 Is a Novel Critical NF-κB Activator in Inflammatory Arthritis.
iScience
Yuan G,Yang S,Ng A,Fu C,Oursler MJ,Xing L,Yang S
Fri Jun 26 00:00:00 EDT 2020
Dietary Ziziphus jujuba Fruit Attenuates Colitis-Associated Tumorigenesis: A Pivotal Role of the NF-κB/IL-6/JAK1/STAT3 Pathway.
Nutrition and cancer
Periasamy S,Wu WH,Chien SP,Liu CT,Liu MY
Thu Sep 10 00:00:00 EDT 2020
Neuroinflammation induced by secretion of acetylated HMGB1 from activated microglia in hippocampi of mice following chronic cold exposure.
Brain research
Xu B,Lang LM,Lian S,Guo JR,Wang JF,Liu J,Yang HM,Li SZ
Wed Jan 01 00:00:00 EST 2020
Dexmedetomidine ameliorates postoperative cognitive dysfunction by inhibiting Toll-like receptor 4 signaling in aged mice.
The Kaohsiung journal of medical sciences
Zhou XY,Liu J,Xu ZP,Fu Q,Wang PQ,Wang JH,Zhang H
Tue Sep 01 00:00:00 EDT 2020
Curcumin-enhanced antitumor effects of sorafenib via regulating the metabolism and tumor microenvironment.
Food & function
Man S,Yao J,Lv P,Liu Y,Yang L,Ma L
Wed Jul 22 00:00:00 EDT 2020
IL (Interleukin)-6 Contributes to Deep Vein Thrombosis and Is Negatively Regulated by miR-338-5p.
Arteriosclerosis, thrombosis, and vascular biology
Zhang Y,Zhang Z,Wei R,Miao X,Sun S,Liang G,Chu C,Zhao L,Zhu X,Guo Q,Wang B,Li X
Sat Feb 01 00:00:00 EST 2020
CircRBM33 regulates IL-6 to promote gastric cancer progression through targeting miR-149.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Wang N,Lu K,Qu H,Wang H,Chen Y,Shan T,Ge X,Wei Y,Zhou P,Xia J
Fri May 01 00:00:00 EDT 2020
TGF-β/Smad and JAK/STAT pathways are involved in the anti-fibrotic effects of propylene glycol alginate sodium sulphate on hepatic fibrosis.
Journal of cellular and molecular medicine
Xu S,Mao Y,Wu J,Feng J,Li J,Wu L,Yu Q,Zhou Y,Zhang J,Chen J,Ji J,Chen K,Wang F,Dai W,Fan X,Guo C
Fri May 01 00:00:00 EDT 2020
TGF-β/Smad and JAK/STAT pathways are involved in the anti-fibrotic effects of propylene glycol alginate sodium sulphate on hepatic fibrosis.
Journal of cellular and molecular medicine
Xu S,Mao Y,Wu J,Feng J,Li J,Wu L,Yu Q,Zhou Y,Zhang J,Chen J,Ji J,Chen K,Wang F,Dai W,Fan X,Guo C
Fri May 01 00:00:00 EDT 2020
miR-7-5p Affects Brain Edema After Intracerebral Hemorrhage and Its Possible Mechanism.
Frontiers in cell and developmental biology
Chen X,Deng S,Lei Q,He Q,Ren Y,Zhang Y,Nie J,Lu W
Tue Jan 05 00:00:00 EST 2021
Disruption of retinoid homeostasis induces RBP4 overproduction in diabetes: O-GlcNAcylation involved.
Metabolism: clinical and experimental
Shin SJ,Chen CH,Kuo WC,Chan HC,Chan HC,Lin KD,Ke LY
Tue Dec 01 00:00:00 EST 2020
Upregulated MELK Leads to Doxorubicin Chemoresistance and M2 Macrophage Polarization via the miR-34a/JAK2/STAT3 Pathway in Uterine Leiomyosarcoma.
Frontiers in oncology
Zhang Z,Sun C,Li C,Jiao X,Griffin BB,Dongol S,Wu H,Zhang C,Cao W,Dong R,Yang X,Zhang Q,Kong B
Mon Sep 28 00:00:00 EDT 2020
Thermosensitive bFGF-Modified Hydrogel with Dental Pulp Stem Cells on Neuroinflammation of Spinal Cord Injury.
ACS omega
Albashari A,He Y,Zhang Y,Ali J,Lin F,Zheng Z,Zhang K,Cao Y,Xu C,Luo L,Wang J,Ye Q
Tue Jul 07 00:00:00 EDT 2020
Thermosensitive bFGF-Modified Hydrogel with Dental Pulp Stem Cells on Neuroinflammation of Spinal Cord Injury.
ACS omega
Albashari A,He Y,Zhang Y,Ali J,Lin F,Zheng Z,Zhang K,Cao Y,Xu C,Luo L,Wang J,Ye Q
Tue Jul 07 00:00:00 EDT 2020
CP-25 improves nephropathy in collagen-induced arthritis rats by inhibiting the renal inflammatory response.
International immunopharmacology
Wang C,Wei X,Wu Y,Tang H,Wang B,Wang Y,Sun W,Asenso J,Xiao F,Wei W
Sun Nov 01 00:00:00 EDT 2020
Peptidome analysis of umbilical cord mesenchymal stem cell (hUC-MSC) conditioned medium from preterm and term infants.
Stem cell research & therapy
Wang Y,Zhang L,Wu Y,Zhu R,Wang Y,Cao Y,Long W,Ji C,Wang H,You L
Wed Sep 23 00:00:00 EDT 2020
Hirudin Ameliorates Renal Interstitial Fibrosis via Regulating TGF-β1/Smad and NF-κB Signaling in UUO Rat Model.
Evidence-based complementary and alternative medicine : eCAM
Yang K,Fan B,Zhao Q,Ji Y,Liu P,Gao S,Ren T,Dou Y,Pei M,Yang H
Fri Apr 15 00:00:00 EDT 2022
Chitosan-calcium alginate dressing promotes wound healing: A preliminary study.
Wound repair and regeneration : official publication of the Wound Healing Society [and] the European Tissue Repair Society
Zhao WY,Fang QQ,Wang XF,Wang XW,Zhang T,Shi BH,Zheng B,Zhang DD,Hu YY,Ma L,Tan WQ
Fri May 01 00:00:00 EDT 2020
Oxymatrine Liposomes for Intervertebral Disc Treatment: Formulation, in vitro and vivo Assessments.
Drug design, development and therapy
Wang H,Ding Y,Zhang W,Wei K,Pei Y,Zou C,Zhang C,Ding J,Fang H,Tan S
Thu Feb 04 00:00:00 EST 2021
Integrin-α9 and Its Corresponding Ligands Play Regulatory Roles in Chronic Periodontitis.
Inflammation
Xu S,Jiang C,Liu H,Zhang H,Liao H,Wang X,Yao S,Ma L,Guo Y,Cao Z
Sat Aug 01 00:00:00 EDT 2020
Polysaccharides from the Fleshy Fruits of Camellia oleifera Attenuate the Kidney Injury in High-Fat Diet/Streptozotocin-Induced Diabetic Mice.
Journal of medicinal food
Li J,Xie F,Zhu X,Hu H,Fang X,Huang Z,Liu Z,Wu S
Tue Dec 01 00:00:00 EST 2020
Polysaccharides from the Fleshy Fruits of Camellia oleifera Attenuate the Kidney Injury in High-Fat Diet/Streptozotocin-Induced Diabetic Mice.
Journal of medicinal food
Li J,Xie F,Zhu X,Hu H,Fang X,Huang Z,Liu Z,Wu S
Tue Dec 01 00:00:00 EST 2020
PTP1B promotes macrophage activation by regulating the NF-κB pathway in alcoholic liver injury.
Toxicology letters
Yang L,Sun YY,Liu YR,Yin NN,Bu FT,Yu HX,Du XS,Li J,Huang C
Sat Feb 01 00:00:00 EST 2020
Proinflammatory and regulatory mechanisms in allergic contact dermatitis caused by methylchloroisothiazolinone and methylisothiazolinone.
Experimental dermatology
Goes HFO,Virgens AR,de Carvalho GC,Pietrobon AJ,Branco ACCC,Oliveira LMDS,Fernandes IG,Pereira NV,Sotto MN,Dos Reis VMS,Sato MN
Fri May 01 00:00:00 EDT 2020
Ginsenoside Rb1 Ameliorates Age-Related Myocardial Dysfunction by Regulating the NF-[Formula: see text]B Signaling Pathway.
The American journal of Chinese medicine
Ke SY,Liu DH,Wu L,Yu XG,Wang M,Shi GY,Wen RH,Zhou B,Hao BS,Liu Y,Zhu JM,Qian XX
Wed Oct 21 00:00:00 EDT 2020
miR-124-3p increases in high glucose induced osteocyte-derived exosomes and regulates galectin-3 expression: A possible mechanism in bone remodeling alteration in diabetic periodontitis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Li J,Guo Y,Chen YY,Liu Q,Chen Y,Tan L,Zhang SH,Gao ZR,Zhou YH,Zhang GY,Feng YZ
Sun Nov 01 00:00:00 EDT 2020
Intravaginal Administration of Human Type III Collagen-Derived Biomaterial with High Cell-Adhesion Activity to Treat Vaginal Atrophy in Rats.
ACS biomaterials science & engineering
You S,Liu S,Dong X,Li H,Zhu Y,Hu L
Mon Apr 13 00:00:00 EDT 2020
Trehalose attenuates renal ischemia-reperfusion injury by enhancing autophagy and inhibiting oxidative stress and inflammation.
American journal of physiology. Renal physiology
Liu S,Yang Y,Gao H,Zhou N,Wang P,Zhang Y,Zhang A,Jia Z,Huang S
Wed Apr 01 00:00:00 EDT 2020
Trehalose attenuates renal ischemia-reperfusion injury by enhancing autophagy and inhibiting oxidative stress and inflammation.
American journal of physiology. Renal physiology
Liu S,Yang Y,Gao H,Zhou N,Wang P,Zhang Y,Zhang A,Jia Z,Huang S
Wed Apr 01 00:00:00 EDT 2020
Huoxin Pill Attenuates Cardiac Inflammation by Suppression of TLR4/NF-κB in Acute Myocardial Ischemia Injury Rats.
Evidence-based complementary and alternative medicine : eCAM
Chu J,Zhou X,Peng M,Lu Y,Farman A,Peng L,Gao H,Li Q,Chen X,Xie L,Chen Y,Shen A,Peng J
Mon Sep 28 00:00:00 EDT 2020
Chronic ethanol exposure induces neuroinflammation in H4 cells through TLR3 / NF-κB pathway and anxiety-like behavior in male C57BL/6 mice.
Toxicology
Wang X,Yu H,Wang C,Liu Y,You J,Wang P,Xu G,Shen H,Yao H,Lan X,Zhao R,Wu X,Zhang G
Tue Dec 15 00:00:00 EST 2020
GABA-mediated activated microglia induce neuroinflammation in the hippocampus of mice following cold exposure through the NLRP3 inflammasome and NF-κB signaling pathways.
International immunopharmacology
Lang L,Xu B,Yuan J,Li S,Lian S,Chen Y,Guo J,Yang H
Tue Dec 01 00:00:00 EST 2020
EZH2-Inhibited MicroRNA-454-3p Promotes M2 Macrophage Polarization in Glioma.
Frontiers in cell and developmental biology
Qi B,Yang C,Zhu Z,Chen H
Tue Dec 29 00:00:00 EST 2020
Protective Effects of Cinnamaldehyde against Mesenteric Ischemia-Reperfusion-Induced Lung and Liver Injuries in Rats.
Oxidative medicine and cellular longevity
Almoiliqy M,Wen J,Qaed E,Sun Y,Lian M,Mousa H,Al-Azab M,Zaky MY,Chen D,Wang L,Al-Sharabi A,Liu Z,Sun P,Lin Y
Thu Sep 02 00:00:00 EDT 2021
Visfatin Exerts Immunotherapeutic Effects in Lipopolysaccharide-Induced Acute Lung Injury in Murine Model.
Inflammation
Luo Y,Pang XX,Ansari AR,Wu XT,Li HZ,Zhang ZW,Song H
Sat Feb 01 00:00:00 EST 2020
Chronotherapy targeting cytokine secretion attenuates collagen-induced arthritis in mice.
International immunopharmacology
Yaekura A,Yoshida K,Morii K,Oketani Y,Okumura I,Kaneshiro K,Shibanuma N,Sakai Y,Hashiramoto A
Wed Jul 01 00:00:00 EDT 2020
HMGB1-mediated differential response on hippocampal neurotransmitter disorder and neuroinflammation in adolescent male and female mice following cold exposure.
Brain, behavior, and immunity
Xu B,Zang SC,Li SZ,Guo JR,Wang JF,Wang D,Zhang LP,Yang HM,Lian S
Fri Feb 01 00:00:00 EST 2019
HMGB1-mediated differential response on hippocampal neurotransmitter disorder and neuroinflammation in adolescent male and female mice following cold exposure.
Brain, behavior, and immunity
Xu B,Zang SC,Li SZ,Guo JR,Wang JF,Wang D,Zhang LP,Yang HM,Lian S
Fri Feb 01 00:00:00 EST 2019
OAB-14, a bexarotene derivative, improves Alzheimer's disease-related pathologies and cognitive impairments by increasing β-amyloid clearance in APP/PS1 mice.
Biochimica et biophysica acta. Molecular basis of disease
Yuan C,Guo X,Zhou Q,Du F,Jiang W,Zhou X,Liu P,Chi T,Ji X,Gao J,Chen C,Lang H,Xu J,Liu D,Yang Y,Qiu S,Tang X,Chen G,Zou L
Tue Jan 01 00:00:00 EST 2019
OAB-14, a bexarotene derivative, improves Alzheimer's disease-related pathologies and cognitive impairments by increasing β-amyloid clearance in APP/PS1 mice.
Biochimica et biophysica acta. Molecular basis of disease
Yuan C,Guo X,Zhou Q,Du F,Jiang W,Zhou X,Liu P,Chi T,Ji X,Gao J,Chen C,Lang H,Xu J,Liu D,Yang Y,Qiu S,Tang X,Chen G,Zou L
Tue Jan 01 00:00:00 EST 2019
Hepatoprotective effects and structure-activity relationship of five flavonoids against lipopolysaccharide/d-galactosamine induced acute liver failure in mice.
International immunopharmacology
He Y,Xia Z,Yu D,Wang J,Jin L,Huang D,Ye X,Li X,Zhang B
Fri Mar 01 00:00:00 EST 2019
Hepatoprotective effects and structure-activity relationship of five flavonoids against lipopolysaccharide/d-galactosamine induced acute liver failure in mice.
International immunopharmacology
He Y,Xia Z,Yu D,Wang J,Jin L,Huang D,Ye X,Li X,Zhang B
Fri Mar 01 00:00:00 EST 2019
Therapeutic Effects of Human Urine-Derived Stem Cells in a Rat Model of Cisplatin-Induced Acute Kidney Injury In Vivo and In Vitro.
Stem cells international
Sun B,Luo X,Yang C,Liu P,Yang Y,Dong X,Yang Z,Xu J,Zhang Y,Li L
Thu May 13 00:00:00 EDT 2021
Selective Serotonin Reuptake Inhibitors Aggravate Depression-Associated Dry Eye Via Activating the NF-κB Pathway.
Investigative ophthalmology & visual science
Zhang X,Yin Y,Yue L,Gong L
Wed Jan 02 00:00:00 EST 2019
Metformin suppresses the esophageal carcinogenesis in rats treated with NMBzA through inhibiting AMPK/mTOR signaling pathway.
Carcinogenesis
Fan H,Yu X,Zou Z,Zheng W,Deng X,Guo L,Jiang W,Zhan Q,Lu SH
Thu Jul 04 00:00:00 EDT 2019
Metformin suppresses the esophageal carcinogenesis in rats treated with NMBzA through inhibiting AMPK/mTOR signaling pathway.
Carcinogenesis
Fan H,Yu X,Zou Z,Zheng W,Deng X,Guo L,Jiang W,Zhan Q,Lu SH
Thu Jul 04 00:00:00 EDT 2019
Macrophage polarization in human gingival tissue in response to periodontal disease.
Oral diseases
Zhou LN,Bi CS,Gao LN,An Y,Chen F,Chen FM
Tue Jan 01 00:00:00 EST 2019
Macrophage polarization in human gingival tissue in response to periodontal disease.
Oral diseases
Zhou LN,Bi CS,Gao LN,An Y,Chen F,Chen FM
Tue Jan 01 00:00:00 EST 2019
Roseburia intestinalis supernatant ameliorates colitis induced in mice by regulating the immune response.
Molecular medicine reports
Luo W,Shen Z,Deng M,Li X,Tan B,Xiao M,Wu S,Yang Z,Zhu C,Tian L,Wu X,Meng X,Quan Y,Wang X
Thu Aug 01 00:00:00 EDT 2019
FOXA1 is Prognostic of Triple Negative Breast Cancers by Transcriptionally Suppressing SOD2 and IL6.
International journal of biological sciences
Dai X,Cheng H,Chen X,Li T,Zhang J,Jin G,Cai D,Huang Z
Mon Mar 16 00:00:00 EDT 2020
Survival-time dependent increase in neuronal IL-6 and astroglial GFAP expression in fatally injured human brain tissue.
Scientific reports
Trautz F,Franke H,Bohnert S,Hammer N,Müller W,Stassart R,Tse R,Zwirner J,Dreßler J,Ondruschka B
Thu Aug 15 00:00:00 EDT 2019
Sweroside Alleviated LPS-Induced Inflammation via SIRT1 Mediating NF-κB and FOXO1 Signaling Pathways in RAW264.7 Cells.
Molecules (Basel, Switzerland)
Wang R,Dong Z,Lan X,Liao Z,Chen M
Fri Mar 01 00:00:00 EST 2019
Sedum sarmentosum Bunge extract alleviates inflammation and kidney injury via inhibition of M1-macrophage polarization.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Lu H,Cheng S,Wu C,Zheng S,Hong W,Liu L,Bai Y
Sun Sep 01 00:00:00 EDT 2019
LncRNA HIF1A-AS1 contributes to ventricular remodeling after myocardial ischemia/reperfusion injury by adsorption of microRNA-204 to regulating SOCS2 expression.
Cell cycle (Georgetown, Tex.)
Xue X,Luo L
Tue Oct 01 00:00:00 EDT 2019
Exploration of the mechanisms of Ge Gen Decoction against influenza A virus infection.
Chinese journal of natural medicines
Geng ZK,Li YQ,Cui QH,DU RK,Tian JZ
Sun Sep 01 00:00:00 EDT 2019
Juxtaposition of IL-1β and IFN-γ expression and apoptosis of keratinocytes in adult-onset Still's disease.
Expert review of clinical immunology
Rao S,Li Q,Wu H,Zhao M,Wang A,Zhang G,Li J,Lu L,Shi W,Lu Q
Sun Dec 01 00:00:00 EST 2019
Anti-interleukin-6 receptor antibody treatment ameliorates postoperative adhesion formation.
Scientific reports
Uyama N,Tsutsui H,Wu S,Yasuda K,Hatano E,Qin XY,Kojima S,Fujimoto J
Tue Nov 26 00:00:00 EST 2019
TLR4 gene in the regulation of periodontitis and its molecular mechanism.
Experimental and therapeutic medicine
Qi W,Yang X,Ye N,Li S,Han Q,Huang J,Wu B
Sun Sep 01 00:00:00 EDT 2019
Differential immune responses of C57BL/6 mice to infection by Salmonella enterica serovar Typhimurium strain SL1344, CVCC541 and CMCC50115.
Virulence
Wei S,Huang J,Liu Z,Wang M,Zhang B,Lian Z,Guo Y,Han H
Sun Dec 01 00:00:00 EST 2019
Curcumin inhibits cigarette smoke-induced inflammation via modulating the PPARγ-NF-κB signaling pathway.
Food & function
Li Q,Sun J,Mohammadtursun N,Wu J,Dong J,Li L
Wed Dec 11 00:00:00 EST 2019
Nrf2 activation in osteoblasts suppresses osteoclastogenesis via inhibiting IL-6 expression.
Bone reports
Narimiya T,Kanzaki H,Yamaguchi Y,Wada S,Katsumata Y,Tanaka K,Tomonari H
Sun Dec 01 00:00:00 EST 2019
Nrf2 activation in osteoblasts suppresses osteoclastogenesis via inhibiting IL-6 expression.
Bone reports
Narimiya T,Kanzaki H,Yamaguchi Y,Wada S,Katsumata Y,Tanaka K,Tomonari H
Sun Dec 01 00:00:00 EST 2019
IMM-H007, a novel small molecule inhibitor for atherosclerosis, represses endothelium inflammation by regulating the activity of NF-κB and JNK/AP1 signaling.
Toxicology and applied pharmacology
Yu J,Ming H,Li HY,Yu B,Chu M,Zhu H,Zhu X
Tue Oct 15 00:00:00 EDT 2019
Olanzapine-induced endoplasmic reticulum stress and inflammation in the hypothalamus were inhibited by an ER stress inhibitor 4-phenylbutyrate.
Psychoneuroendocrinology
He M,Huang XF,Gao G,Zhou T,Li W,Hu J,Chen J,Li J,Sun T
Sat Jun 01 00:00:00 EDT 2019
Nr2e1 ablation impairs liver glucolipid metabolism and induces inflammation, high-fat diets amplify the damage.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Xiong Q,Wu Y,Yang M,Wu G,Wang Y,Wang H,Feng J,Song L,Tong B,He G,Xu Y
Sun Dec 01 00:00:00 EST 2019
TAK242 suppresses the TLR4 signaling pathway and ameliorates DCD liver IRI in rats.
Molecular medicine reports
Zhong X,Xiao Q,Liu Z,Wang W,Lai CH,Yang W,Yue P,Ye Q,Xiao J
Sun Sep 01 00:00:00 EDT 2019
TAK242 suppresses the TLR4 signaling pathway and ameliorates DCD liver IRI in rats.
Molecular medicine reports
Zhong X,Xiao Q,Liu Z,Wang W,Lai CH,Yang W,Yue P,Ye Q,Xiao J
Sun Sep 01 00:00:00 EDT 2019
Inhibition of Fibroblast Growth Factor Receptor by AZD4547 Protects Against Inflammation in Septic Mice.
Inflammation
Huang Y,Wang F,Li H,Xu S,Xu W,Pan X,Hu Y,Mao L,Qian S,Pan J
Sun Dec 01 00:00:00 EST 2019
Clock mutant promotes osteoarthritis by inhibiting the acetylation of NFκB.
Osteoarthritis and cartilage
Yuan G,Xu L,Cai T,Hua B,Sun N,Yan Z,Lu C,Qian R
Sat Jun 01 00:00:00 EDT 2019
Liver-target nanotechnology facilitates berberine to ameliorate cardio-metabolic diseases.
Nature communications
Guo HH,Feng CL,Zhang WX,Luo ZG,Zhang HJ,Zhang TT,Ma C,Zhan Y,Li R,Wu S,Abliz Z,Li C,Li XL,Ma XL,Wang LL,Zheng WS,Han YX,Jiang JD
Tue Apr 30 00:00:00 EDT 2019
Suppression of miR-143 contributes to overexpression of IL-6, HIF-1α and NF-κB p65 in Cr(VI)-induced human exposure and tumor growth.
Toxicology and applied pharmacology
Wang L,Qiu JG,He J,Liu WJ,Ge X,Zhou FM,Huang YX,Jiang BH,Liu LZ
Sun Sep 01 00:00:00 EDT 2019
Comparison of the Hepatoprotective Effects of the Three Main Stilbenes from Mulberry Twigs.
Journal of agricultural and food chemistry
Jia YN,Peng YL,Zhao YP,Cheng XF,Zhou Y,Chai CL,Zeng LS,Pan MH,Xu L
Wed May 15 00:00:00 EDT 2019
Gut microbe-derived metabolite trimethylamine N-oxide activates the cardiac autonomic nervous system and facilitates ischemia-induced ventricular arrhythmia via two different pathways.
EBioMedicine
Meng G,Zhou X,Wang M,Zhou L,Wang Z,Wang M,Deng J,Wang Y,Zhou Z,Zhang Y,Lai Y,Zhang Q,Yang X,Yu L,Jiang H
Sat Jun 01 00:00:00 EDT 2019
Upregulation of IL-6 in CUL4B-deficient myeloid-derived suppressive cells increases the aggressiveness of cancer cells.
Oncogene
Xu Z,Li L,Qian Y,Song Y,Qin L,Duan Y,Wang M,Li P,Jiang B,Ma C,Shao C,Gong Y
Mon Jul 01 00:00:00 EDT 2019
Knockdown of periostin attenuates 5/6 nephrectomy-induced intrarenal renin-angiotensin system activation, fibrosis, and inflammation in rats.
Journal of cellular physiology
Bian X,Bai Y,Su X,Zhao G,Sun G,Li D
Sun Dec 01 00:00:00 EST 2019
Sulforaphane Inhibits the Acquisition of Tobacco Smoke-Induced Lung Cancer Stem Cell-Like Properties via the IL-6/ΔNp63α/Notch Axis.
Theranostics
Xie C,Zhu J,Jiang Y,Chen J,Wang X,Geng S,Wu J,Zhong C,Li X,Meng Z
Tue Aug 25 00:00:00 EDT 2020
Toxicological exploration of peptide-based cationic liposomes in siRNA delivery.
Colloids and surfaces. B, Biointerfaces
Zhu Y,Meng Y,Zhao Y,Zhu J,Xu H,Zhang E,Shi L,Du L,Liu G,Zhang C,Xu X,Kang X,Zhen Y,Zhang S
Mon Jul 01 00:00:00 EDT 2019
Rice Protein Exerts Anti-Inflammatory Effect in Growing and Adult Rats via Suppressing NF-κB Pathway.
International journal of molecular sciences
Wang Z,Liang M,Li H,Cai L,Yang L
Fri Dec 06 00:00:00 EST 2019
Sinomenine contributes to the inhibition of the inflammatory response and the improvement of osteoarthritis in mouse-cartilage cells by acting on the Nrf2/HO-1 and NF-κB signaling pathways.
International immunopharmacology
Wu Y,Lin Z,Yan Z,Wang Z,Fu X,Yu K
Tue Oct 01 00:00:00 EDT 2019
Interaction with tumor‑associated macrophages promotes PRL‑3‑induced invasion of colorectal cancer cells via MAPK pathway‑induced EMT and NF‑κB signaling‑induced angiogenesis.
Oncology reports
Zhang T,Liu L,Lai W,Zeng Y,Xu H,Lan Q,Su P,Chu Z
Wed May 01 00:00:00 EDT 2019
Protective effects of levo-tetrahydropalmatine on hepatic ischemia/reperfusion injury are mediated by inhibition of the ERK/NF-κB pathway.
International immunopharmacology
Yu Q,Wu L,Liu T,Li S,Feng J,Mao Y,Fan X,Guo C,Wu J
Wed May 01 00:00:00 EDT 2019
Three kinds of corneal host cells contribute differently to corneal neovascularization.
EBioMedicine
Yu H,Sun L,Cui J,Li Y,Yan Y,Wei X,Wang C,Song F,Jiang W,Liu Y,Ge H,Qian H,Li X,Tang X,Liu P
Sat Jun 01 00:00:00 EDT 2019
Inhibition of myeloid differentiation 1 specifically in colon with antisense oligonucleotide exacerbates dextran sodium sulfate-induced colitis.
Journal of cellular biochemistry
Chen X,Pan H,Li J,Zhang G,Cheng S,Zuo N,Zhao Q,Peng Z
Tue Oct 01 00:00:00 EDT 2019
Protective effects of ginsenoside Rk3 against chronic alcohol-induced liver injury in mice through inhibition of inflammation, oxidative stress, and apoptosis.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Qu L,Zhu Y,Liu Y,Yang H,Zhu C,Ma P,Deng J,Fan D
Mon Apr 01 00:00:00 EDT 2019
Aspirin inhibits inflammation and scar formation in the injury tendon healing through regulating JNK/STAT-3 signalling pathway.
Cell proliferation
Wang Y,He G,Tang H,Shi Y,Kang X,Lyu J,Zhu M,Zhou M,Yang M,Mu M,Chen W,Zhou B,Zhang J,Tang K
Mon Jul 01 00:00:00 EDT 2019
Bioactive Constituents from the Roots of Eurycoma longifolia.
Molecules (Basel, Switzerland)
Ruan J,Li Z,Zhang Y,Chen Y,Liu M,Han L,Zhang Y,Wang T
Fri Aug 30 00:00:00 EDT 2019
Thyrotropin aggravates atherosclerosis by promoting macrophage inflammation in plaques.
The Journal of experimental medicine
Yang C,Lu M,Chen W,He Z,Hou X,Feng M,Zhang H,Bo T,Zhou X,Yu Y,Zhang H,Zhao M,Wang L,Yu C,Gao L,Jiang W,Zhang Q,Zhao J
Mon May 06 00:00:00 EDT 2019
Vaccaria n-Butanol Extract Lower the Production of Proinflammatory Cytokines and the Infection Risk of T. spiralis In Vivo.
Acta parasitologica
Xu F,Hou B,Zhu X,Liu Y,Shi X,Li S,Li Z,Cai W,Zhou Y,Qiu L
Sun Sep 01 00:00:00 EDT 2019
Bax inhibitor-1 overexpression in prelimbic cortex protects rats against depression-like behavior induced by olfactory bulbectomy and reduces apoptotic and inflammatory signals.
Neurological research
Li D,Cai Z,Wu J,Zhang Y
Mon Apr 01 00:00:00 EDT 2019
Pigment epithelium-derived factor inhibits adipogenesis in 3T3-L1 adipocytes and protects against high-fat diet-induced obesity and metabolic disorders in mice.
Translational research : the journal of laboratory and clinical medicine
Chen CC,Lee TY,Leu YL,Wang SH
Thu Aug 01 00:00:00 EDT 2019
Anti-tumour effects of red blood cell membrane-camouflaged black phosphorous quantum dots combined with chemotherapy and anti-inflammatory therapy.
Artificial cells, nanomedicine, and biotechnology
Huang X,Wu B,Li J,Shang Y,Chen W,Nie X,Gui R
Sun Dec 01 00:00:00 EST 2019
Anti-tumour effects of red blood cell membrane-camouflaged black phosphorous quantum dots combined with chemotherapy and anti-inflammatory therapy.
Artificial cells, nanomedicine, and biotechnology
Huang X,Wu B,Li J,Shang Y,Chen W,Nie X,Gui R
Sun Dec 01 00:00:00 EST 2019
MicroRNA-34a/IL-6R pathway as a potential therapeutic target for ovarian high-grade serous carcinoma.
Oncotarget
Yokomizo R,Yanaihara N,Yamaguchi N,Saito M,Kawabata A,Takahashi K,Takenaka M,Yamada K,Shapiro JS,Okamoto A
Tue Aug 06 00:00:00 EDT 2019
Taraxacum officinale extract ameliorates dextran sodium sulphate-induced colitis by regulating fatty acid degradation and microbial dysbiosis.
Journal of cellular and molecular medicine
Chen W,Fan H,Liang R,Zhang R,Zhang J,Zhu J
Sun Dec 01 00:00:00 EST 2019
FA2H Exhibits Tumor Suppressive Roles on Breast Cancers via Cancer Stemness Control.
Frontiers in oncology
Dai X,Zhang S,Cheng H,Cai D,Chen X,Huang Z
Sat Oct 14 00:00:00 EDT 2023
M1 macrophages regulate TLR4/AP1 via paracrine to promote alveolar bone destruction in periodontitis.
Oral diseases
Zhu LF,Li L,Wang XQ,Pan L,Mei YM,Fu YW,Xu Y
Fri Nov 01 00:00:00 EDT 2019
Omentin-1 prevents inflammation-induced osteoporosis by downregulating the pro-inflammatory cytokines.
Bone research
Rao SS,Hu Y,Xie PL,Cao J,Wang ZX,Liu JH,Yin H,Huang J,Tan YJ,Luo J,Luo MJ,Tang SY,Chen TH,Yuan LQ,Liao EY,Xu R,Liu ZZ,Chen CY,Xie H
Sun Nov 12 00:00:00 EST 2023
Omentin-1 prevents inflammation-induced osteoporosis by downregulating the pro-inflammatory cytokines.
Bone research
Rao SS,Hu Y,Xie PL,Cao J,Wang ZX,Liu JH,Yin H,Huang J,Tan YJ,Luo J,Luo MJ,Tang SY,Chen TH,Yuan LQ,Liao EY,Xu R,Liu ZZ,Chen CY,Xie H
Sun Nov 12 00:00:00 EST 2023
Omentin-1 prevents inflammation-induced osteoporosis by downregulating the pro-inflammatory cytokines.
Bone research
Rao SS,Hu Y,Xie PL,Cao J,Wang ZX,Liu JH,Yin H,Huang J,Tan YJ,Luo J,Luo MJ,Tang SY,Chen TH,Yuan LQ,Liao EY,Xu R,Liu ZZ,Chen CY,Xie H
Sun Nov 12 00:00:00 EST 2023
Omentin-1 prevents inflammation-induced osteoporosis by downregulating the pro-inflammatory cytokines.
Bone research
Rao SS,Hu Y,Xie PL,Cao J,Wang ZX,Liu JH,Yin H,Huang J,Tan YJ,Luo J,Luo MJ,Tang SY,Chen TH,Yuan LQ,Liao EY,Xu R,Liu ZZ,Chen CY,Xie H
Sun Nov 12 00:00:00 EST 2023
CAPE-pNO2 attenuates diabetic cardiomyopathy through the NOX4/NF-κB pathway in STZ-induced diabetic mice.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Fan L,Xiao Q,Zhang L,Wang X,Huang Q,Li S,Zhao X,Li Z
Sat Dec 01 00:00:00 EST 2018
CAPE-pNO2 attenuates diabetic cardiomyopathy through the NOX4/NF-κB pathway in STZ-induced diabetic mice.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Fan L,Xiao Q,Zhang L,Wang X,Huang Q,Li S,Zhao X,Li Z
Sat Dec 01 00:00:00 EST 2018
Total glucosides of paeony improves the immunomodulatory capacity of MSCs partially via the miR-124/STAT3 pathway in oral lichen planus.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhao Z,Han Y,Zhang Z,Li W,Ji X,Liu X,Jin J,Xu S,Cui H,Cheng Z,Wang Q,Wang X,Guo X,Wang Y,Liu H
Sat Sep 01 00:00:00 EDT 2018
Total glucosides of paeony improves the immunomodulatory capacity of MSCs partially via the miR-124/STAT3 pathway in oral lichen planus.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhao Z,Han Y,Zhang Z,Li W,Ji X,Liu X,Jin J,Xu S,Cui H,Cheng Z,Wang Q,Wang X,Guo X,Wang Y,Liu H
Sat Sep 01 00:00:00 EDT 2018
Isorhamnetin: A hepatoprotective flavonoid inhibits apoptosis and autophagy via P38/PPAR-α pathway in mice.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Lu X,Liu T,Chen K,Xia Y,Dai W,Xu S,Xu L,Wang F,Wu L,Li J,Li S,Wang W,Yu Q,Feng J,Fan X,Zhou Y,Niu P,Guo C
Sun Jul 01 00:00:00 EDT 2018
Isorhamnetin: A hepatoprotective flavonoid inhibits apoptosis and autophagy via P38/PPAR-α pathway in mice.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Lu X,Liu T,Chen K,Xia Y,Dai W,Xu S,Xu L,Wang F,Wu L,Li J,Li S,Wang W,Yu Q,Feng J,Fan X,Zhou Y,Niu P,Guo C
Sun Jul 01 00:00:00 EDT 2018
Isorhamnetin: A hepatoprotective flavonoid inhibits apoptosis and autophagy via P38/PPAR-α pathway in mice.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Lu X,Liu T,Chen K,Xia Y,Dai W,Xu S,Xu L,Wang F,Wu L,Li J,Li S,Wang W,Yu Q,Feng J,Fan X,Zhou Y,Niu P,Guo C
Sun Jul 01 00:00:00 EDT 2018
Isorhamnetin: A hepatoprotective flavonoid inhibits apoptosis and autophagy via P38/PPAR-α pathway in mice.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Lu X,Liu T,Chen K,Xia Y,Dai W,Xu S,Xu L,Wang F,Wu L,Li J,Li S,Wang W,Yu Q,Feng J,Fan X,Zhou Y,Niu P,Guo C
Sun Jul 01 00:00:00 EDT 2018
miR-19b-3p induces cell proliferation and reduces heterochromatin-mediated senescence through PLZF in goat male germline stem cells.
Journal of cellular physiology
Daguia Zambe JC,Zhai Y,Zhou Z,Du X,Wei Y,Ma F,Hua J
Fri Jun 01 00:00:00 EDT 2018
miR-19b-3p induces cell proliferation and reduces heterochromatin-mediated senescence through PLZF in goat male germline stem cells.
Journal of cellular physiology
Daguia Zambe JC,Zhai Y,Zhou Z,Du X,Wei Y,Ma F,Hua J
Fri Jun 01 00:00:00 EDT 2018
Abberent expression of NOR1 protein in tumor associated macrophages contributes to the development of DEN-induced hepatocellular carcinoma.
Journal of cellular physiology
Chen S,Zheng P,Wang W,Yi M,Chen P,Cai J,Li J,Peng Q,Ban Y,Zhou Y,Zeng Z,Li X,Xiong W,Li G,Xiang B
Fri Jun 01 00:00:00 EDT 2018
Abberent expression of NOR1 protein in tumor associated macrophages contributes to the development of DEN-induced hepatocellular carcinoma.
Journal of cellular physiology
Chen S,Zheng P,Wang W,Yi M,Chen P,Cai J,Li J,Peng Q,Ban Y,Zhou Y,Zeng Z,Li X,Xiong W,Li G,Xiang B
Fri Jun 01 00:00:00 EDT 2018
Quercetin ameliorates kidney injury and fibrosis by modulating M1/M2 macrophage polarization.
Biochemical pharmacology
Lu H,Wu L,Liu L,Ruan Q,Zhang X,Hong W,Wu S,Jin G,Bai Y
Wed Aug 01 00:00:00 EDT 2018
Quercetin ameliorates kidney injury and fibrosis by modulating M1/M2 macrophage polarization.
Biochemical pharmacology
Lu H,Wu L,Liu L,Ruan Q,Zhang X,Hong W,Wu S,Jin G,Bai Y
Wed Aug 01 00:00:00 EDT 2018
Hydrogen sulfide protects neonatal rat medulla oblongata against prenatal cigarette smoke exposure via anti-oxidative and anti-inflammatory effects.
Environmental toxicology and pharmacology
Yan X,Lei F,Hu Y,Nie L,Jia Q,Zhou H,Zhao F,Zheng Y
Mon Jan 01 00:00:00 EST 2018
Hydrogen sulfide protects neonatal rat medulla oblongata against prenatal cigarette smoke exposure via anti-oxidative and anti-inflammatory effects.
Environmental toxicology and pharmacology
Yan X,Lei F,Hu Y,Nie L,Jia Q,Zhou H,Zhao F,Zheng Y
Mon Jan 01 00:00:00 EST 2018
Asiatic acid from Potentilla chinensis alleviates non-alcoholic fatty liver by regulating endoplasmic reticulum stress and lipid metabolism.
International immunopharmacology
Wang D,Lao L,Pang X,Qiao Q,Pang L,Feng Z,Bai F,Sun X,Lin X,Wei J
Sat Dec 01 00:00:00 EST 2018
Asiatic acid from Potentilla chinensis alleviates non-alcoholic fatty liver by regulating endoplasmic reticulum stress and lipid metabolism.
International immunopharmacology
Wang D,Lao L,Pang X,Qiao Q,Pang L,Feng Z,Bai F,Sun X,Lin X,Wei J
Sat Dec 01 00:00:00 EST 2018
Oxyresveratrol prevents lipopolysaccharide/d-galactosamine-induced acute liver injury in mice.
International immunopharmacology
Jia YN,Lu HP,Peng YL,Zhang BS,Gong XB,Su J,Zhou Y,Pan MH,Xu L
Thu Mar 01 00:00:00 EST 2018
Oxyresveratrol prevents lipopolysaccharide/d-galactosamine-induced acute liver injury in mice.
International immunopharmacology
Jia YN,Lu HP,Peng YL,Zhang BS,Gong XB,Su J,Zhou Y,Pan MH,Xu L
Thu Mar 01 00:00:00 EST 2018
Photobiomodulation therapy promotes neurogenesis by improving post-stroke local microenvironment and stimulating neuroprogenitor cells.
Experimental neurology
Yang L,Tucker D,Dong Y,Wu C,Lu Y,Li Y,Zhang J,Liu TC,Zhang Q
Mon Jan 01 00:00:00 EST 2018
Photobiomodulation therapy promotes neurogenesis by improving post-stroke local microenvironment and stimulating neuroprogenitor cells.
Experimental neurology
Yang L,Tucker D,Dong Y,Wu C,Lu Y,Li Y,Zhang J,Liu TC,Zhang Q
Mon Jan 01 00:00:00 EST 2018
Relevance of interferon-gamma in pathogenesis of life-threatening rapidly progressive interstitial lung disease in patients with dermatomyositis.
Arthritis research & therapy
Ishikawa Y,Iwata S,Hanami K,Nawata A,Zhang M,Yamagata K,Hirata S,Sakata K,Todoroki Y,Nakano K,Nakayamada S,Satoh M,Tanaka Y
Fri Oct 26 00:00:00 EDT 2018
Relevance of interferon-gamma in pathogenesis of life-threatening rapidly progressive interstitial lung disease in patients with dermatomyositis.
Arthritis research & therapy
Ishikawa Y,Iwata S,Hanami K,Nawata A,Zhang M,Yamagata K,Hirata S,Sakata K,Todoroki Y,Nakano K,Nakayamada S,Satoh M,Tanaka Y
Fri Oct 26 00:00:00 EDT 2018
The Protective Effects of Levo-Tetrahydropalmatine on ConA-Induced Liver Injury Are via TRAF6/JNK Signaling.
Mediators of inflammation
Yu Q,Liu T,Li S,Feng J,Wu L,Wang W,Chen K,Xia Y,Niu P,Xu L,Wang F,Dai W,Zhou Y,Guo C
Fri Apr 19 00:00:00 EDT 2019
The Protective Effects of Levo-Tetrahydropalmatine on ConA-Induced Liver Injury Are via TRAF6/JNK Signaling.
Mediators of inflammation
Yu Q,Liu T,Li S,Feng J,Wu L,Wang W,Chen K,Xia Y,Niu P,Xu L,Wang F,Dai W,Zhou Y,Guo C
Fri Apr 19 00:00:00 EDT 2019
The Protective Effects of Levo-Tetrahydropalmatine on ConA-Induced Liver Injury Are via TRAF6/JNK Signaling.
Mediators of inflammation
Yu Q,Liu T,Li S,Feng J,Wu L,Wang W,Chen K,Xia Y,Niu P,Xu L,Wang F,Dai W,Zhou Y,Guo C
Fri Apr 19 00:00:00 EDT 2019
The Protective Effects of Levo-Tetrahydropalmatine on ConA-Induced Liver Injury Are via TRAF6/JNK Signaling.
Mediators of inflammation
Yu Q,Liu T,Li S,Feng J,Wu L,Wang W,Chen K,Xia Y,Niu P,Xu L,Wang F,Dai W,Zhou Y,Guo C
Fri Apr 19 00:00:00 EDT 2019
Neuroprotective Effect of Anethole Against Neuropathic Pain Induced by Chronic Constriction Injury of the Sciatic Nerve in Mice.
Neurochemical research
Wang B,Zhang G,Yang M,Liu N,Li YX,Ma H,Ma L,Sun T,Tan H,Yu J
Sat Dec 01 00:00:00 EST 2018
Neuroprotective Effect of Anethole Against Neuropathic Pain Induced by Chronic Constriction Injury of the Sciatic Nerve in Mice.
Neurochemical research
Wang B,Zhang G,Yang M,Liu N,Li YX,Ma H,Ma L,Sun T,Tan H,Yu J
Sat Dec 01 00:00:00 EST 2018
Tert-butylhydroquinone post-treatment attenuates neonatal hypoxic-ischemic brain damage in rats.
Neurochemistry international
Zhang J,Tucker LD,Lu Y,Yang L,Wu C,Li Y,Zhang Q
Fri Jun 01 00:00:00 EDT 2018
Mini-peptide RPL41 attenuated retinal neovascularization by inducing degradation of ATF4 in oxygen-induced retinopathy mice.
Experimental cell research
Geng W,Qin F,Ren J,Xiao S,Wang A
Wed Aug 15 00:00:00 EDT 2018
Mini-peptide RPL41 attenuated retinal neovascularization by inducing degradation of ATF4 in oxygen-induced retinopathy mice.
Experimental cell research
Geng W,Qin F,Ren J,Xiao S,Wang A
Wed Aug 15 00:00:00 EDT 2018
GABAB receptor mediate hippocampal neuroinflammation in adolescent male and female mice after cold expose.
Brain research bulletin
Xu B,Lian S,Li SZ,Guo JR,Wang JF,Wang D,Zhang LP,Yang HM
Sat Sep 01 00:00:00 EDT 2018
GABAB receptor mediate hippocampal neuroinflammation in adolescent male and female mice after cold expose.
Brain research bulletin
Xu B,Lian S,Li SZ,Guo JR,Wang JF,Wang D,Zhang LP,Yang HM
Sat Sep 01 00:00:00 EDT 2018
Mechanism of salvianolic acid B neuroprotection against ischemia/reperfusion induced cerebral injury.
Brain research
Fan Y,Luo Q,Wei J,Lin R,Lin L,Li Y,Chen Z,Lin W,Chen Q
Mon Jan 15 00:00:00 EST 2018
Mechanism of salvianolic acid B neuroprotection against ischemia/reperfusion induced cerebral injury.
Brain research
Fan Y,Luo Q,Wei J,Lin R,Lin L,Li Y,Chen Z,Lin W,Chen Q
Mon Jan 15 00:00:00 EST 2018
Skeletal muscle secretion of IL-6 is muscle type specific: Ex vivo evidence.
Biochemical and biophysical research communications
Liang AP,Drazick AT,Gao H,Li Y
Sat Oct 20 00:00:00 EDT 2018
Skeletal muscle secretion of IL-6 is muscle type specific: Ex vivo evidence.
Biochemical and biophysical research communications
Liang AP,Drazick AT,Gao H,Li Y
Sat Oct 20 00:00:00 EDT 2018
Rapamycin and FTY720 Alleviate Atherosclerosis by Cross Talk of Macrophage Polarization and Autophagy.
BioMed research international
Sun RZ,Fan Y,Liang X,Gong TT,Wang Q,Liu H,Shan ZY,Lei L
Wed Apr 24 00:00:00 EDT 2019
Rapamycin and FTY720 Alleviate Atherosclerosis by Cross Talk of Macrophage Polarization and Autophagy.
BioMed research international
Sun RZ,Fan Y,Liang X,Gong TT,Wang Q,Liu H,Shan ZY,Lei L
Wed Apr 24 00:00:00 EDT 2019
TAM Infiltration Differences in "Tumor-First" and " ZHENG-First" Models and the Underlying Inflammatory Molecular Mechanism in Pancreatic Cancer.
Integrative cancer therapies
Wang FJ,Wang P,Chen LY,Geng YW,Chen H,Meng ZQ,Liu LM,Chen Z
Sat Sep 01 00:00:00 EDT 2018
TAM Infiltration Differences in "Tumor-First" and " ZHENG-First" Models and the Underlying Inflammatory Molecular Mechanism in Pancreatic Cancer.
Integrative cancer therapies
Wang FJ,Wang P,Chen LY,Geng YW,Chen H,Meng ZQ,Liu LM,Chen Z
Sat Sep 01 00:00:00 EDT 2018
Influence of vitamin D deficiency on T cell subsets and related indices during spinal tuberculosis.
Experimental and therapeutic medicine
Zhou SH,Wang X,Fan MY,Li HL,Bian F,Huang T,Fang HY
Wed Aug 01 00:00:00 EDT 2018
Influence of vitamin D deficiency on T cell subsets and related indices during spinal tuberculosis.
Experimental and therapeutic medicine
Zhou SH,Wang X,Fan MY,Li HL,Bian F,Huang T,Fang HY
Wed Aug 01 00:00:00 EDT 2018
Effects of SB202190 on expression levels of IL-6 and NF-κB in flap ischemia-reperfusion injury.
Experimental and therapeutic medicine
Guo K,Ma J,Liang W
Sat Sep 01 00:00:00 EDT 2018
Effects of SB202190 on expression levels of IL-6 and NF-κB in flap ischemia-reperfusion injury.
Experimental and therapeutic medicine
Guo K,Ma J,Liang W
Sat Sep 01 00:00:00 EDT 2018
Underexpression of Receptor for Activated C Kinase 1 (RACK1) in Leukocytes from Patients with Severe Acute Pancreatitis.
The Tohoku journal of experimental medicine
Zhang X,Guo J,Wang H,Zhang C,Shi N,Cai W,Jin T,Lin Z,Ma Y,Yang X,Xia Q,Xue P
Sun Jul 01 00:00:00 EDT 2018
Underexpression of Receptor for Activated C Kinase 1 (RACK1) in Leukocytes from Patients with Severe Acute Pancreatitis.
The Tohoku journal of experimental medicine
Zhang X,Guo J,Wang H,Zhang C,Shi N,Cai W,Jin T,Lin Z,Ma Y,Yang X,Xia Q,Xue P
Sun Jul 01 00:00:00 EDT 2018
Upregulation of phosphorylated sphingosine kinase 1 expression in colitis-associated cancer.
The Journal of surgical research
Yuza K,Nagahashi M,Shimada Y,Nakano M,Tajima Y,Kameyama H,Nakajima M,Takabe K,Wakai T
Thu Nov 01 00:00:00 EDT 2018
Upregulation of phosphorylated sphingosine kinase 1 expression in colitis-associated cancer.
The Journal of surgical research
Yuza K,Nagahashi M,Shimada Y,Nakano M,Tajima Y,Kameyama H,Nakajima M,Takabe K,Wakai T
Thu Nov 01 00:00:00 EDT 2018
Vaccarin alleviates hypertension and nephropathy in renovascular hypertensive rats.
Experimental and therapeutic medicine
Cai W,Zhang Z,Huang Y,Sun H,Qiu L
Mon Jan 01 00:00:00 EST 2018
Butaselen prevents hepatocarcinogenesis and progression through inhibiting thioredoxin reductase activity.
Redox biology
Zheng X,Ma W,Sun R,Yin H,Lin F,Liu Y,Xu W,Zeng H
Sun Apr 01 00:00:00 EDT 2018
Butaselen prevents hepatocarcinogenesis and progression through inhibiting thioredoxin reductase activity.
Redox biology
Zheng X,Ma W,Sun R,Yin H,Lin F,Liu Y,Xu W,Zeng H
Sun Apr 01 00:00:00 EDT 2018
Effects of platelet-rich plasma on the activity of human menstrual blood-derived stromal cells in vitro.
Stem cell research & therapy
Zhang S,Li P,Yuan Z,Tan J
Mon Feb 26 00:00:00 EST 2018
Effects of platelet-rich plasma on the activity of human menstrual blood-derived stromal cells in vitro.
Stem cell research & therapy
Zhang S,Li P,Yuan Z,Tan J
Mon Feb 26 00:00:00 EST 2018
Lipocalin 2 Suppresses Ocular Inflammation by Inhibiting the Activation of NF-κβ Pathway in Endotoxin-Induced Uveitis.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Tang W,Ma J,Gu R,Ding X,Lei B,Wang X,Zhuang H,Xu G
Mon Jun 25 00:00:00 EDT 2018
Lipocalin 2 Suppresses Ocular Inflammation by Inhibiting the Activation of NF-κβ Pathway in Endotoxin-Induced Uveitis.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Tang W,Ma J,Gu R,Ding X,Lei B,Wang X,Zhuang H,Xu G
Mon Jun 25 00:00:00 EDT 2018
Protective Effects of Costunolide Against D-Galactosamine and Lipopolysaccharide-Induced Acute Liver Injury in Mice.
Frontiers in pharmacology
Mao J,Yi M,Wang R,Huang Y,Chen M
Sat Apr 09 00:00:00 EDT 2022
Protective Effects of Costunolide Against D-Galactosamine and Lipopolysaccharide-Induced Acute Liver Injury in Mice.
Frontiers in pharmacology
Mao J,Yi M,Wang R,Huang Y,Chen M
Sat Apr 09 00:00:00 EDT 2022
The development of salt-sensitive hypertension regulated by PSGL-1 gene in mice.
Cell & bioscience
Yang Y,Liu X,Liu Y,Fu H,Gao Y,Liu X,Jiang X
Wed Mar 16 00:00:00 EDT 2022
Cardioprotection of CAPE-oNO2 against myocardial ischemia/reperfusion induced ROS generation via regulating the SIRT1/eNOS/NF-κB pathway in vivo and in vitro.
Redox biology
Li D,Wang X,Huang Q,Li S,Zhou Y,Li Z
Tue May 01 00:00:00 EDT 2018
Cardioprotection of CAPE-oNO2 against myocardial ischemia/reperfusion induced ROS generation via regulating the SIRT1/eNOS/NF-κB pathway in vivo and in vitro.
Redox biology
Li D,Wang X,Huang Q,Li S,Zhou Y,Li Z
Tue May 01 00:00:00 EDT 2018
Surgical incision induces learning impairment in mice partially through inhibition of the brain-derived neurotrophic factor signaling pathway in the hippocampus and amygdala.
Molecular pain
Liu Z,Liu F,Liu X,Ma C,Zhao J
Mon Jan 14 00:00:00 EST 2019
Surgical incision induces learning impairment in mice partially through inhibition of the brain-derived neurotrophic factor signaling pathway in the hippocampus and amygdala.
Molecular pain
Liu Z,Liu F,Liu X,Ma C,Zhao J
Mon Jan 14 00:00:00 EST 2019
Ultraviolet B Inhibits IL-17A/TNF-α-Stimulated Activation of Human Dermal Fibroblasts by Decreasing the Expression of IL-17RA and IL-17RC on Fibroblasts.
Frontiers in immunology
Yin L,Hu Y,Xu J,Guo J,Tu J,Yin Z
Thu Mar 31 00:00:00 EDT 2022
Ultraviolet B Inhibits IL-17A/TNF-α-Stimulated Activation of Human Dermal Fibroblasts by Decreasing the Expression of IL-17RA and IL-17RC on Fibroblasts.
Frontiers in immunology
Yin L,Hu Y,Xu J,Guo J,Tu J,Yin Z
Thu Mar 31 00:00:00 EDT 2022
Ultraviolet B Inhibits IL-17A/TNF-α-Stimulated Activation of Human Dermal Fibroblasts by Decreasing the Expression of IL-17RA and IL-17RC on Fibroblasts.
Frontiers in immunology
Yin L,Hu Y,Xu J,Guo J,Tu J,Yin Z
Thu Mar 31 00:00:00 EDT 2022
Ultraviolet B Inhibits IL-17A/TNF-α-Stimulated Activation of Human Dermal Fibroblasts by Decreasing the Expression of IL-17RA and IL-17RC on Fibroblasts.
Frontiers in immunology
Yin L,Hu Y,Xu J,Guo J,Tu J,Yin Z
Thu Mar 31 00:00:00 EDT 2022
Diffuse large B cell lymphoma associated with chronic inflammation arising within atrial myxoma: aggressive histological features but indolent clinical behaviour.
Histopathology
Yan J,Luo D,Zhang F,He J,Yao S,Luo X,Xu F,Chen Y,Fu L,Xu J,Liu Y
Fri Dec 01 00:00:00 EST 2017
Diffuse large B cell lymphoma associated with chronic inflammation arising within atrial myxoma: aggressive histological features but indolent clinical behaviour.
Histopathology
Yan J,Luo D,Zhang F,He J,Yao S,Luo X,Xu F,Chen Y,Fu L,Xu J,Liu Y
Fri Dec 01 00:00:00 EST 2017
Antioxidation, anti-inflammation and anti-apoptosis by paeonol in LPS/d-GalN-induced acute liver failure in mice.
International immunopharmacology
Gong X,Yang Y,Huang L,Zhang Q,Wan RZ,Zhang P,Zhang B
Mon May 01 00:00:00 EDT 2017
Antioxidation, anti-inflammation and anti-apoptosis by paeonol in LPS/d-GalN-induced acute liver failure in mice.
International immunopharmacology
Gong X,Yang Y,Huang L,Zhang Q,Wan RZ,Zhang P,Zhang B
Mon May 01 00:00:00 EDT 2017
Prognostic impact of interleukin-6 expression in stage I ovarian clear cell carcinoma.
Gynecologic oncology
Kawabata A,Yanaihara N,Nagata C,Saito M,Noguchi D,Takenaka M,Iida Y,Takano H,Yamada K,Iwamoto M,Kiyokawa T,Okamoto A
Fri Sep 01 00:00:00 EDT 2017
Prognostic impact of interleukin-6 expression in stage I ovarian clear cell carcinoma.
Gynecologic oncology
Kawabata A,Yanaihara N,Nagata C,Saito M,Noguchi D,Takenaka M,Iida Y,Takano H,Yamada K,Iwamoto M,Kiyokawa T,Okamoto A
Fri Sep 01 00:00:00 EDT 2017
The acute exposure of tetrachloro-p-benzoquinone (a.k.a. chloranil) triggers inflammation and neurological dysfunction via Toll-like receptor 4 signaling: The protective role of melatonin preconditioning.
Toxicology
Fu J,Xia X,Liu Z,Wang Y,Wang Y,Shi Q,Song X,Song E,Song Y
Sat Apr 15 00:00:00 EDT 2017
The acute exposure of tetrachloro-p-benzoquinone (a.k.a. chloranil) triggers inflammation and neurological dysfunction via Toll-like receptor 4 signaling: The protective role of melatonin preconditioning.
Toxicology
Fu J,Xia X,Liu Z,Wang Y,Wang Y,Shi Q,Song X,Song E,Song Y
Sat Apr 15 00:00:00 EDT 2017
Neuroprotective Effects of Lycium barbarum Polysaccharide on Focal Cerebral Ischemic Injury in Mice.
Neurochemical research
Zhao P,Zhou R,Zhu XY,Liu G,Zhao YP,Ma PS,Wu W,Niu Y,Sun T,Li YX,Yu JQ,Qian ZM
Sun Oct 01 00:00:00 EDT 2017
Neuroprotective Effects of Lycium barbarum Polysaccharide on Focal Cerebral Ischemic Injury in Mice.
Neurochemical research
Zhao P,Zhou R,Zhu XY,Liu G,Zhao YP,Ma PS,Wu W,Niu Y,Sun T,Li YX,Yu JQ,Qian ZM
Sun Oct 01 00:00:00 EDT 2017
Inflammatory and oxidative stress‑associated factors in chronic intermittent hypoxia in Chinese patients, rats, lymphocytes and endotheliocytes.
Molecular medicine reports
Ren H,Hu K
Fri Dec 01 00:00:00 EST 2017
The novel preventive effect of Daikenchuto (TJ-100), a Japanese herbal drug, against neonatal necrotizing enterocolitis in rats.
Pediatric surgery international
Shinyama S,Kaji T,Mukai M,Nakame K,Matsufuji H,Takamatsu H,Ieiri S
Sun Oct 01 00:00:00 EDT 2017
The novel preventive effect of Daikenchuto (TJ-100), a Japanese herbal drug, against neonatal necrotizing enterocolitis in rats.
Pediatric surgery international
Shinyama S,Kaji T,Mukai M,Nakame K,Matsufuji H,Takamatsu H,Ieiri S
Sun Oct 01 00:00:00 EDT 2017
The liver protection of propylene glycol alginate sodium sulfate preconditioning against ischemia reperfusion injury: focusing MAPK pathway activity.
Scientific reports
Xu S,Niu P,Chen K,Xia Y,Yu Q,Liu N,Li J,Li S,Wu L,Feng J,Wang W,Lu X,Liu T,Wang F,Dai W,Fan X,Mo W,Xu L,Guo C
Thu Nov 09 00:00:00 EST 2017
The liver protection of propylene glycol alginate sodium sulfate preconditioning against ischemia reperfusion injury: focusing MAPK pathway activity.
Scientific reports
Xu S,Niu P,Chen K,Xia Y,Yu Q,Liu N,Li J,Li S,Wu L,Feng J,Wang W,Lu X,Liu T,Wang F,Dai W,Fan X,Mo W,Xu L,Guo C
Thu Nov 09 00:00:00 EST 2017
The liver protection of propylene glycol alginate sodium sulfate preconditioning against ischemia reperfusion injury: focusing MAPK pathway activity.
Scientific reports
Xu S,Niu P,Chen K,Xia Y,Yu Q,Liu N,Li J,Li S,Wu L,Feng J,Wang W,Lu X,Liu T,Wang F,Dai W,Fan X,Mo W,Xu L,Guo C
Thu Nov 09 00:00:00 EST 2017
The liver protection of propylene glycol alginate sodium sulfate preconditioning against ischemia reperfusion injury: focusing MAPK pathway activity.
Scientific reports
Xu S,Niu P,Chen K,Xia Y,Yu Q,Liu N,Li J,Li S,Wu L,Feng J,Wang W,Lu X,Liu T,Wang F,Dai W,Fan X,Mo W,Xu L,Guo C
Thu Nov 09 00:00:00 EST 2017
Mechanism of Lycium barbarum polysaccharides on primary cultured rat hippocampal neurons.
Cell and tissue research
Zhao P,Ma NT,Chang RY,Li YX,Hao YJ,Yang WL,Zheng J,Niu Y,Sun T,Yu JQ
Fri Sep 01 00:00:00 EDT 2017
hCLOCK induction by hypoxia promotes inflammatory responses by activating the NF‑κB pathway.
Molecular medicine reports
Tang X,Guo D,Lin C,Shi Z,Qian R,Fu W,Liu J,Li X,Fan L
Wed Mar 01 00:00:00 EST 2017
Effects of Ketamine on Levels of Inflammatory Cytokines IL-6, IL-1β, and TNF-α in the Hippocampus of Mice Following Acute or Chronic Administration.
Frontiers in pharmacology
Li Y,Shen R,Wen G,Ding R,Du A,Zhou J,Dong Z,Ren X,Yao H,Zhao R,Zhang G,Lu Y,Wu X
Fri Mar 11 00:00:00 EST 2022
Effects of Ketamine on Levels of Inflammatory Cytokines IL-6, IL-1β, and TNF-α in the Hippocampus of Mice Following Acute or Chronic Administration.
Frontiers in pharmacology
Li Y,Shen R,Wen G,Ding R,Du A,Zhou J,Dong Z,Ren X,Yao H,Zhao R,Zhang G,Lu Y,Wu X
Fri Mar 11 00:00:00 EST 2022
BRD7 plays an anti-inflammatory role during early acute inflammation by inhibiting activation of the NF-кB signaling pathway.
Cellular & molecular immunology
Zhao R,Liu Y,Wang H,Yang J,Niu W,Fan S,Xiong W,Ma J,Li X,Phillips JB,Tan M,Qiu Y,Li G,Zhou M
Sun Oct 01 00:00:00 EDT 2017
Ovatodiolide suppresses colon tumorigenesis and prevents polarization of M2 tumor-associated macrophages through YAP oncogenic pathways.
Journal of hematology & oncology
Huang YJ,Yang CK,Wei PL,Huynh TT,Whang-Peng J,Meng TC,Hsiao M,Tzeng YM,Wu AT,Yen Y
Tue Feb 28 00:00:00 EST 2017
Ovatodiolide suppresses colon tumorigenesis and prevents polarization of M2 tumor-associated macrophages through YAP oncogenic pathways.
Journal of hematology & oncology
Huang YJ,Yang CK,Wei PL,Huynh TT,Whang-Peng J,Meng TC,Hsiao M,Tzeng YM,Wu AT,Yen Y
Tue Feb 28 00:00:00 EST 2017
CAPE-pNO2 ameliorated diabetic nephropathy through regulating the Akt/NF-κB/ iNOS pathway in STZ-induced diabetic mice.
Oncotarget
Wang X,Li D,Fan L,Xiao Q,Zuo H,Li Z
Fri Dec 29 00:00:00 EST 2017
CAPE-pNO2 ameliorated diabetic nephropathy through regulating the Akt/NF-κB/ iNOS pathway in STZ-induced diabetic mice.
Oncotarget
Wang X,Li D,Fan L,Xiao Q,Zuo H,Li Z
Fri Dec 29 00:00:00 EST 2017
Synergistic IL-6 and IL-8 paracrine signalling pathway infers a strategy to inhibit tumour cell migration.
Nature communications
Jayatilaka H,Tyle P,Chen JJ,Kwak M,Ju J,Kim HJ,Lee JSH,Wu PH,Gilkes DM,Fan R,Wirtz D
Fri May 26 00:00:00 EDT 2017
Synergistic IL-6 and IL-8 paracrine signalling pathway infers a strategy to inhibit tumour cell migration.
Nature communications
Jayatilaka H,Tyle P,Chen JJ,Kwak M,Ju J,Kim HJ,Lee JSH,Wu PH,Gilkes DM,Fan R,Wirtz D
Fri May 26 00:00:00 EDT 2017
Naoling decoction restores cognitive function by inhibiting the neuroinflammatory network in a rat model of Alzheimer's disease.
Oncotarget
Xia Z,Peng W,Cheng S,Zhong B,Sheng C,Zhang C,Gong W,Cheng S,Li J,Wang Z
Tue Jun 27 00:00:00 EDT 2017
Naoling decoction restores cognitive function by inhibiting the neuroinflammatory network in a rat model of Alzheimer's disease.
Oncotarget
Xia Z,Peng W,Cheng S,Zhong B,Sheng C,Zhang C,Gong W,Cheng S,Li J,Wang Z
Tue Jun 27 00:00:00 EDT 2017
Diosmetin exerts anti-oxidative, anti-inflammatory and anti-apoptotic effects to protect against endotoxin-induced acute hepatic failure in mice.
Oncotarget
Yang Y,Gong XB,Huang LG,Wang ZX,Wan RZ,Zhang P,Zhang QY,Chen Z,Zhang BS
Tue May 09 00:00:00 EDT 2017
Diosmetin exerts anti-oxidative, anti-inflammatory and anti-apoptotic effects to protect against endotoxin-induced acute hepatic failure in mice.
Oncotarget
Yang Y,Gong XB,Huang LG,Wang ZX,Wan RZ,Zhang P,Zhang QY,Chen Z,Zhang BS
Tue May 09 00:00:00 EDT 2017
Neuroprotective effect of liquiritin against neuropathic pain induced by chronic constriction injury of the sciatic nerve in mice.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhang MT,Wang B,Jia YN,Liu N,Ma PS,Gong SS,Niu Y,Sun T,Li YX,Yu JQ
Wed Nov 01 00:00:00 EDT 2017
Elevated Level of Circulating Platelet-derived Microparticles in Oral Cancer.
Journal of dental research
Ren JG,Man QW,Zhang W,Li C,Xiong XP,Zhu JY,Wang WM,Sun ZJ,Jia J,Zhang WF,Zhao YF,Chen G,Liu B
Fri Jan 01 00:00:00 EST 2016
Elevated Level of Circulating Platelet-derived Microparticles in Oral Cancer.
Journal of dental research
Ren JG,Man QW,Zhang W,Li C,Xiong XP,Zhu JY,Wang WM,Sun ZJ,Jia J,Zhang WF,Zhao YF,Chen G,Liu B
Fri Jan 01 00:00:00 EST 2016
Low molecular weight fucoidan modulates P-selectin and alleviates diabetic nephropathy.
International journal of biological macromolecules
Xu Y,Zhang Q,Luo D,Wang J,Duan D
Sat Oct 01 00:00:00 EDT 2016
Low molecular weight fucoidan modulates P-selectin and alleviates diabetic nephropathy.
International journal of biological macromolecules
Xu Y,Zhang Q,Luo D,Wang J,Duan D
Sat Oct 01 00:00:00 EDT 2016
Low molecular weight fucoidan modulates P-selectin and alleviates diabetic nephropathy.
International journal of biological macromolecules
Xu Y,Zhang Q,Luo D,Wang J,Duan D
Sat Oct 01 00:00:00 EDT 2016
Low molecular weight fucoidan modulates P-selectin and alleviates diabetic nephropathy.
International journal of biological macromolecules
Xu Y,Zhang Q,Luo D,Wang J,Duan D
Sat Oct 01 00:00:00 EDT 2016
Interleukin-6 enhances cancer stemness and promotes metastasis of hepatocellular carcinoma via up-regulating osteopontin expression.
American journal of cancer research
Wang CQ,Sun HT,Gao XM,Ren N,Sheng YY,Wang Z,Zheng Y,Wei JW,Zhang KL,Yu XX,Zhu Y,Luo Q,Yang LY,Dong QZ,Qin LX
Fri Apr 08 00:00:00 EDT 2022
Interleukin-6 enhances cancer stemness and promotes metastasis of hepatocellular carcinoma via up-regulating osteopontin expression.
American journal of cancer research
Wang CQ,Sun HT,Gao XM,Ren N,Sheng YY,Wang Z,Zheng Y,Wei JW,Zhang KL,Yu XX,Zhu Y,Luo Q,Yang LY,Dong QZ,Qin LX
Fri Apr 08 00:00:00 EDT 2022
Tetrachlorobenzoquinone exhibits neurotoxicity by inducing inflammatory responses through ROS-mediated IKK/IκB/NF-κB signaling.
Environmental toxicology and pharmacology
Fu J,Shi Q,Song X,Xia X,Su C,Liu Z,Song E,Song Y
Fri Jan 01 00:00:00 EST 2016
Tetrachlorobenzoquinone exhibits neurotoxicity by inducing inflammatory responses through ROS-mediated IKK/IκB/NF-κB signaling.
Environmental toxicology and pharmacology
Fu J,Shi Q,Song X,Xia X,Su C,Liu Z,Song E,Song Y
Fri Jan 01 00:00:00 EST 2016
Human β-defensin-3 alleviates the progression of atherosclerosis accelerated by Porphyromonas gingivalis lipopolysaccharide.
International immunopharmacology
Li L,Bian T,Lyu J,Cui D,Lei L,Yan F
Thu Sep 01 00:00:00 EDT 2016
Human β-defensin-3 alleviates the progression of atherosclerosis accelerated by Porphyromonas gingivalis lipopolysaccharide.
International immunopharmacology
Li L,Bian T,Lyu J,Cui D,Lei L,Yan F
Thu Sep 01 00:00:00 EDT 2016
Palmitic acid aggravates inflammation of pancreatic acinar cells by enhancing unfolded protein response induced CCAAT-enhancer-binding protein β-CCAAT-enhancer-binding protein α activation.
The international journal of biochemistry & cell biology
Wu J,Hu G,Lu Y,Zheng J,Chen J,Wang X,Zeng Y
Sat Oct 01 00:00:00 EDT 2016
Palmitic acid aggravates inflammation of pancreatic acinar cells by enhancing unfolded protein response induced CCAAT-enhancer-binding protein β-CCAAT-enhancer-binding protein α activation.
The international journal of biochemistry & cell biology
Wu J,Hu G,Lu Y,Zheng J,Chen J,Wang X,Zeng Y
Sat Oct 01 00:00:00 EDT 2016
MicroRNA-499 Rs3746444 polymorphism and biliary atresia.
Digestive and liver disease : official journal of the Italian Society of Gastroenterology and the Italian Association for the Study of the Liver
Shan Y,Shen N,Han L,Chen Q,Zhang J,Long X,Xia Q
Fri Apr 01 00:00:00 EDT 2016
MicroRNA-499 Rs3746444 polymorphism and biliary atresia.
Digestive and liver disease : official journal of the Italian Society of Gastroenterology and the Italian Association for the Study of the Liver
Shan Y,Shen N,Han L,Chen Q,Zhang J,Long X,Xia Q
Fri Apr 01 00:00:00 EDT 2016
A fundamental study on the dynamics of multiple biomarkers in mouse excisional wounds for wound age estimation.
Journal of forensic and legal medicine
Wang LL,Zhao R,Liu CS,Liu M,Li SS,Li JY,Jiang SK,Zhang M,Tian ZL,Wang M,Zhang MZ,Guan DW
Fri Apr 01 00:00:00 EDT 2016
A fundamental study on the dynamics of multiple biomarkers in mouse excisional wounds for wound age estimation.
Journal of forensic and legal medicine
Wang LL,Zhao R,Liu CS,Liu M,Li SS,Li JY,Jiang SK,Zhang M,Tian ZL,Wang M,Zhang MZ,Guan DW
Fri Apr 01 00:00:00 EDT 2016
Non-toxic retention of nanoceria in murine eyes.
Molecular vision
Cai X,Seal S,McGinnis JF
Mon Jan 15 00:00:00 EST 2018
Non-toxic retention of nanoceria in murine eyes.
Molecular vision
Cai X,Seal S,McGinnis JF
Mon Jan 15 00:00:00 EST 2018
From the Cover: Tetrachlorobenzoquinone Exerts Neurological Proinflammatory Activity by Promoting HMGB1 Release, Which Induces TLR4 Clustering within the Lipid Raft.
Toxicological sciences : an official journal of the Society of Toxicology
Fu J,Shi Q,Song X,Liu Z,Wang Y,Wang Y,Song E,Song Y
Sat Oct 01 00:00:00 EDT 2016
Senescent Vascular Smooth Muscle Cells Drive Inflammation Through an Interleukin-1α-Dependent Senescence-Associated Secretory Phenotype.
Arteriosclerosis, thrombosis, and vascular biology
Gardner SE,Humphry M,Bennett MR,Clarke MC
Tue Sep 01 00:00:00 EDT 2015
Senescent Vascular Smooth Muscle Cells Drive Inflammation Through an Interleukin-1α-Dependent Senescence-Associated Secretory Phenotype.
Arteriosclerosis, thrombosis, and vascular biology
Gardner SE,Humphry M,Bennett MR,Clarke MC
Tue Sep 01 00:00:00 EDT 2015
hCLOCK Causes Rho-Kinase-Mediated Endothelial Dysfunction and NF-κB-Mediated Inflammatory Responses.
Oxidative medicine and cellular longevity
Tang X,Guo D,Lin C,Shi Z,Qian R,Fu W,Liu J,Li X,Fan L
Mon Jul 25 00:00:00 EDT 2016
hCLOCK Causes Rho-Kinase-Mediated Endothelial Dysfunction and NF-κB-Mediated Inflammatory Responses.
Oxidative medicine and cellular longevity
Tang X,Guo D,Lin C,Shi Z,Qian R,Fu W,Liu J,Li X,Fan L
Mon Jul 25 00:00:00 EDT 2016
Artificial sweetener neohesperidin dihydrochalcone showed antioxidative, anti-inflammatory and anti-apoptosis effects against paraquat-induced liver injury in mice.
International immunopharmacology
Shi Q,Song X,Fu J,Su C,Xia X,Song E,Song Y
Tue Dec 01 00:00:00 EST 2015
Artificial sweetener neohesperidin dihydrochalcone showed antioxidative, anti-inflammatory and anti-apoptosis effects against paraquat-induced liver injury in mice.
International immunopharmacology
Shi Q,Song X,Fu J,Su C,Xia X,Song E,Song Y
Tue Dec 01 00:00:00 EST 2015
The quantum of initial transformed cells potentially modulates the type of local inflammation mechanism elicited by surrounding normal epithelial tissues and systemic immune pattern for tumor arrest or progression.
Journal of Cancer
Owusu L,Wang B,Du Y,Li W,Xin Y
Tue Jan 06 00:00:00 EST 2015
The quantum of initial transformed cells potentially modulates the type of local inflammation mechanism elicited by surrounding normal epithelial tissues and systemic immune pattern for tumor arrest or progression.
Journal of Cancer
Owusu L,Wang B,Du Y,Li W,Xin Y
Tue Jan 06 00:00:00 EST 2015
The effect of maternal obesity on the expression and functionality of placental P-glycoprotein: Implications in the individualized transplacental digoxin treatment for fetal heart failure.
Placenta
Wang C,Li H,Luo C,Li Y,Zhang Y,Yun D,Mu D,Zhou K,Hua Y
Thu Oct 01 00:00:00 EDT 2015
The effect of maternal obesity on the expression and functionality of placental P-glycoprotein: Implications in the individualized transplacental digoxin treatment for fetal heart failure.
Placenta
Wang C,Li H,Luo C,Li Y,Zhang Y,Yun D,Mu D,Zhou K,Hua Y
Thu Oct 01 00:00:00 EDT 2015
Mitochondrion-Targeted Peptide SS-31 Inhibited Oxidized Low-Density Lipoproteins-Induced Foam Cell Formation through both ROS Scavenging and Inhibition of Cholesterol Influx in RAW264.7 Cells.
Molecules (Basel, Switzerland)
Hao S,Ji J,Zhao H,Shang L,Wu J,Li H,Qiao T,Li K
Tue Dec 01 00:00:00 EST 2015
Mitochondrion-Targeted Peptide SS-31 Inhibited Oxidized Low-Density Lipoproteins-Induced Foam Cell Formation through both ROS Scavenging and Inhibition of Cholesterol Influx in RAW264.7 Cells.
Molecules (Basel, Switzerland)
Hao S,Ji J,Zhao H,Shang L,Wu J,Li H,Qiao T,Li K
Tue Dec 01 00:00:00 EST 2015
MFHAS1 Is Associated with Sepsis and Stimulates TLR2/NF-κB Signaling Pathway Following Negative Regulation.
PloS one
Zhong J,Shi QQ,Zhu MM,Shen J,Wang HH,Ma D,Miao CH
Mon Jun 20 00:00:00 EDT 2016
MFHAS1 Is Associated with Sepsis and Stimulates TLR2/NF-κB Signaling Pathway Following Negative Regulation.
PloS one
Zhong J,Shi QQ,Zhu MM,Shen J,Wang HH,Ma D,Miao CH
Mon Jun 20 00:00:00 EDT 2016
Role of α-lipoic acid in LPS/d-GalN induced fulminant hepatic failure in mice: studies on oxidative stress, inflammation and apoptosis.
International immunopharmacology
Xia X,Su C,Fu J,Zhang P,Jiang X,Xu D,Hu L,Song E,Song Y
Wed Oct 01 00:00:00 EDT 2014
Role of α-lipoic acid in LPS/d-GalN induced fulminant hepatic failure in mice: studies on oxidative stress, inflammation and apoptosis.
International immunopharmacology
Xia X,Su C,Fu J,Zhang P,Jiang X,Xu D,Hu L,Song E,Song Y
Wed Oct 01 00:00:00 EDT 2014
Protective effects of neohesperidin dihydrochalcone against carbon tetrachloride-induced oxidative damage in vivo and in vitro.
Chemico-biological interactions
Hu L,Li L,Xu D,Xia X,Pi R,Xu D,Wang W,Du H,Song E,Song Y
Fri Apr 25 00:00:00 EDT 2014
Protective effects of neohesperidin dihydrochalcone against carbon tetrachloride-induced oxidative damage in vivo and in vitro.
Chemico-biological interactions
Hu L,Li L,Xu D,Xia X,Pi R,Xu D,Wang W,Du H,Song E,Song Y
Fri Apr 25 00:00:00 EDT 2014
Tetrachloro-p-benzoquinone induces hepatic oxidative damage and inflammatory response, but not apoptosis in mouse: the prevention of curcumin.
Toxicology and applied pharmacology
Xu D,Hu L,Su C,Xia X,Zhang P,Fu J,Wang W,Xu D,Du H,Hu Q,Song E,Song Y
Wed Oct 15 00:00:00 EDT 2014
Tetrachloro-p-benzoquinone induces hepatic oxidative damage and inflammatory response, but not apoptosis in mouse: the prevention of curcumin.
Toxicology and applied pharmacology
Xu D,Hu L,Su C,Xia X,Zhang P,Fu J,Wang W,Xu D,Du H,Hu Q,Song E,Song Y
Wed Oct 15 00:00:00 EDT 2014
Ethyl pyruvate pretreatment attenuates concanavalin a-induced autoimmune hepatitis in mice.
PloS one
Shen M,Lu J,Cheng P,Lin C,Dai W,Wang F,Wang C,Zhang Y,Chen K,Xu L,Zhou Y,Guo C
Mon Oct 20 00:00:00 EDT 2014
Ethyl pyruvate pretreatment attenuates concanavalin a-induced autoimmune hepatitis in mice.
PloS one
Shen M,Lu J,Cheng P,Lin C,Dai W,Wang F,Wang C,Zhang Y,Chen K,Xu L,Zhou Y,Guo C
Mon Oct 20 00:00:00 EDT 2014
N-acetylcysteine attenuates ischemia-reperfusion-induced apoptosis and autophagy in mouse liver via regulation of the ROS/JNK/Bcl-2 pathway.
PloS one
Wang C,Chen K,Xia Y,Dai W,Wang F,Shen M,Cheng P,Wang J,Lu J,Zhang Y,Yang J,Zhu R,Zhang H,Li J,Zheng Y,Zhou Y,Guo C
Mon Sep 28 00:00:00 EDT 2015
N-acetylcysteine attenuates ischemia-reperfusion-induced apoptosis and autophagy in mouse liver via regulation of the ROS/JNK/Bcl-2 pathway.
PloS one
Wang C,Chen K,Xia Y,Dai W,Wang F,Shen M,Cheng P,Wang J,Lu J,Zhang Y,Yang J,Zhu R,Zhang H,Li J,Zheng Y,Zhou Y,Guo C
Mon Sep 28 00:00:00 EDT 2015
Impaired autophagy contributes to adverse cardiac remodeling in acute myocardial infarction.
PloS one
Wu X,He L,Chen F,He X,Cai Y,Zhang G,Yi Q,He M,Luo J
Tue Jul 14 00:00:00 EDT 2015
Impaired autophagy contributes to adverse cardiac remodeling in acute myocardial infarction.
PloS one
Wu X,He L,Chen F,He X,Cai Y,Zhang G,Yi Q,He M,Luo J
Tue Jul 14 00:00:00 EDT 2015
Lactoferrin deficiency promotes colitis-associated colorectal dysplasia in mice.
PloS one
Ye Q,Zheng Y,Fan S,Qin Z,Li N,Tang A,Ai F,Zhang X,Bian Y,Dang W,Huang J,Zhou M,Zhou Y,Xiong W,Yan Q,Ma J,Li G
Fri Nov 13 00:00:00 EST 2015
Lactoferrin deficiency promotes colitis-associated colorectal dysplasia in mice.
PloS one
Ye Q,Zheng Y,Fan S,Qin Z,Li N,Tang A,Ai F,Zhang X,Bian Y,Dang W,Huang J,Zhou M,Zhou Y,Xiong W,Yan Q,Ma J,Li G
Fri Nov 13 00:00:00 EST 2015
Ethyl pyruvate ameliorates hepatic ischemia-reperfusion injury by inhibiting intrinsic pathway of apoptosis and autophagy.
Mediators of inflammation
Shen M,Lu J,Dai W,Wang F,Xu L,Chen K,He L,Cheng P,Zhang Y,Wang C,Wu D,Yang J,Zhu R,Zhang H,Zhou Y,Guo C
Tue Oct 14 00:00:00 EDT 2014
Ethyl pyruvate ameliorates hepatic ischemia-reperfusion injury by inhibiting intrinsic pathway of apoptosis and autophagy.
Mediators of inflammation
Shen M,Lu J,Dai W,Wang F,Xu L,Chen K,He L,Cheng P,Zhang Y,Wang C,Wu D,Yang J,Zhu R,Zhang H,Zhou Y,Guo C
Tue Oct 14 00:00:00 EDT 2014