Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).
This antibody has specificity for Human IL10RB/CRFB4/CDw210b.
Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDw210b; CRF2-4; cytokine receptor class-II CRF2-4; Cytokine receptor class-II member 4; Cytokine receptor family 2 member 4; cytokine receptor family II, member 4; IL-10 receptor subunit beta; IL-10R subunit 2; IL-10R subunit beta; IL-10RB; inte; interleukin 10 receptor, beta; Interleukin-10 receptor subunit 2; Interleukin-10 receptor subunit beta
Gene Aliases: CDW210B; CRF2-4; CRFB4; D21S58; D21S66; IL-10R2; IL10RB
UniProt ID: (Human) Q08334
Entrez Gene ID: (Human) 3588
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support