Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GPPPGPPRVS PDPRAELDST VLLTRSLLAD TRQLAAQLRD KFPADGDHNL DSLPTLAMSA GALGALQLPG VLTRLRADLL SYLRHVQWLR RAGGSSLKTL EPELGTLQAR LDRLLRRLQL LMSRLALPQP PPDPPAPPLA PPSSAWGGIR AAHAILGGLH LTLDWAVRGL LLLKTRL
Interleukins (ILs) are a large group of cytokines that are produced mainly by leukocytes, although some are produced by certain phagocytes and auxiliary cells. ILs have a variety of functions, but most function to direct other immune cells to divide and differentiate. Each IL acts on a specific, limited group of cells through a receptor specific for that IL.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adipogenesis inhibitory factor; AGIF; il 11; IL-11; IL-11ra; IL10RA; IL11 inhibitor; ILN; Interleukin; Interleukin-11; Interleukin11; MGC2146; oprelvekin
Gene Aliases: AGIF; IL-11; IL11
UniProt ID: (Human) P20809
Entrez Gene ID: (Human) 3589
Molecular Function:
interleukin superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support