Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MAHTFRGCSL AFMFIITWLL IKAKIDACKR GDVTVKPSHV ILLGSTVNIT CSLKPRQGCF HYSRRNKLIL YKFDRRINFH HGHSLNSQVT GLPLGTTLFV CKLACINSDE IQICGAEIFV GVAPEQPQNL SCIQKGEQGT VACTWERGRD THLYTEYTLQ LSGPKNLTWQ KQCKDIYCDY LDFGINLTPE SPESNFTAKV TAVNSLGSSS SLPSTFTFLD IVRPLPPWDI RIKFQKASVS RCTLYWRDEG LVLLNRLRYR PSNSRLWNMV NVTKAKGRHD LLDLKPFTEY EFQISSKLHL YKGSWSDWSE SLRAQTPEEE PTGMLDVWYM KRHIDYSRQQ ISLFWKNLSV SEARGKILHY QVTLQELTGG KAMTQNITGH TSWTTVIPRT GNWAVAVSAA NSKGSSLPTR INIMNLCEAG LLAPRQVSAN SEGMDNILVT WQPPRKDPSA VQEYVVEWRE LHPGGDTQVP LNWLRSRPYN VSALISENIK SYICYEIRVY ALSGDQGGCS SILGNSKHKA PLSGPHINAI TEEKGSILIS WNSIPVQEQM GCLLHYRIYW KERDSNSQPQ LCEIPYRVSQ NSHPINSLQP RVTYVLWMTA LTAAGESSHG NEREFCLQGK ANWMAFVAPS ICIAIIMVGI FSTHYFQQKV FVLLAALRPQ WCSREIPDPA NSTCAKKYPI AEEKTQLPLD RLLIDWPTPE DPEPLVISEV LHQVTPVFRH PPCSNWPQRE KGIQGHQASE KDMMHSASSP PPPRALQAES RQLVDLYKVL ESRGSDPKPE NPACPWTVLP AGDLPTHDGY LPSNIDDLPS HEAPLADSLE ELEPQHISLS VFPSSSLHPL TFSCGDKLTL DQLKMRCDSL ML
The protein encoded by this gene is a type I transmembrane protein identified as a subunit of the interleukin 12 receptor complex. The coexpression of this and IL12RB1 proteins was shown to lead to the formation of high-affinity IL12 binding sites and reconstitution of IL12 dependent signaling. The expression of this gene is up-regulated by interferon gamma in Th1 cells, and plays a role in Th1 cell differentiation. The up-regulation of this gene is found to be associated with a number of infectious diseases, such as Crohn's disease and leprosy, which is thought to contribute to the inflammatory response and host defense.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IL-12 receptor subunit beta-2; IL-12R subunit beta-2; interleukin 12 receptor, beta 2; interleukin-12 receptor beta-2 chain; Interleukin-12 receptor subunit beta-2; RP11-102M16.1
Gene Aliases: IL12RB2
UniProt ID: (Human) Q99665
Entrez Gene ID: (Human) 3595
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support