Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MECPARLCGL WALLLCAGGG GGSGGAAPTE TQPPVTNLSV SVENLCTVIW TWNPPEGASS NCSLWYFSHF GDKQDKKIAP ETRRSIEVPL NERICLQVGS QCSTNESEKP SILVEKCISP PEGDPESAVT ELQCIWHNLS YMKCSWLPGR NTSPDTNYTL YYWHRSLEKI HQCENIFREG QYFGCSFDLT KVKDSSFEQH SVQIMVKDNA GKIKPSFNIV PLTSRVKPDP PHIKNLSFHN DDLYVQWENP QNFISRCLFY EVEVNNSQTE THNVFYVQEA KCENPEFERN VENTSCFMVP GVLPDTLNTV RIRVKTNKLC YEDDKLWSNW SQEMSIGKKR NSTLYITMLL IVPVIVAGAI IVLLLYLKRL KIIIFPPIPD PGKIFKEMFG DQNDDTLHWK KYDIYEKQTK EETDSVVLIE NLKKASQ
The protein encoded by this gene is a subunit of the interleukin 13 receptor. This subunit forms a receptor complex with IL4 receptor alpha, a subunit shared by IL13 and IL4 receptors. This subunit serves as a primary IL13-binding subunit of the IL13 receptor, and may also be a component of IL4 receptors. This protein has been shown to bind tyrosine kinase TYK2, and thus may mediate the signaling processes that lead to the activation of JAK1, STAT3 and STAT6 induced by IL13 and IL4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 19; CD antigen CD213a1; CD213a1; CD213a1 antigen; CT19; IL-13 receptor subunit alpha-1; IL-13R subunit alpha-1; IL-13RA1; IL13 receptor alpha-1 chain; interleukin 13 receptor, alpha 1; Interleukin-13 receptor subunit alpha-1
Gene Aliases: CD213A1; CT19; IL-13Ra; IL13R; IL13RA; IL13RA1; NR4
UniProt ID: (Human) P78552
Entrez Gene ID: (Human) 3597
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support