Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGAARSPPSA VPGPLLGLLL LLLGVLAPGG ASLRLLDHRA LVCSQPGLNC TVKNSTCLDD SWIHPRNLTP SSPKDLQIQL HFAHTQQGDL FPVAHIEWTL QTDASILYLE GAELSVLQLN TNERLCVRFE FLSKLRHHHR RWRFTFSHFV VDPDQEYEVT VHHLPKPIPD GDPNHQSKNF LVPDCEHARM KVTTPCMSSG SLWDPNITVE TLEAHQLRVS FTLWNESTHY QILLTSFPHM ENHSCFEHMH HIPAPRPEEF HQRSNVTLTL RNLKGCCRHQ VQIQPFFSSC LNDCLRHSAT VSCPEMPDTP EPIPDYMPLW VYWFITGISI LLVGSVILLI VCMTWRLAGP GSEKYSDDTK YTDGLPVADL IPPPLKPRKV WIIYSADHPL YVDVVLKFAQ FLLTACGTEV ALDLLEEQAI SEAGVMTWVG RQKQEMVESN SKIIVLCSRG TRAKWQALLG RGAPVRLRCD HGKPVGDLFT AAMNMILPDF KRPACFGTYV VCYFSEVSCD GDVPDLFGAA PRYPLMDRFE EVYFRIQDLE MFQPGRMHRV GELSGDNYLR SPGGRQLRAA LDRFRDWQVH CPDWFECENL YSADDQDAPS LDEEVFEEPL LPPGTGIVKR APLVREPGSQ ACLAIDPLVG EEGGAAVAKL EPHLQPRGQP APQPLHTLVL AAEEGALVAA VEPGPLADGA AVRLALAGEG EACPLLGSPG AGRNSVLFLP VDPEDSPLGS STPMASPDLL PEDVREHLEG LMLSLFEQSL SCQAQGGCSR PAMVLTDPHT PYEEEQRQSV QSDQGYISRS SPQPPEGLTE MEEEEEEEQD PGKPALPLSP EDLESLRSLQ RQLLFRQLQK NSGWDTMGSE SEGPSA
Interleukin 17A (IL-17A) is a proinflammatory cytokine secreted by activated T-lymphocytes. It is a potent inducer of the maturation of CD34-positive hematopoietic precursors into neutrophils. IL-17RA is a ubiquitous type I membrane glycoprotein that binds with low affinity to IL-17A and IL-17F. IL-17A and its receptor play a pathogenic role in many inflammatory and autoimmune diseases such as rheumatoid arthritis. Defects in IL-17RA are the cause of familial candidiasis type 5 (CANDF5) which is a rare disorder with altered immune responses and impaired clearance of fungal infections.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD217; CDw217; IL-17 receptor A; Interleukin-17 receptor A; MGC10262
Gene Aliases: CANDF5; CD217; CDw217; hIL-17R; IL-17RA; IL17R; IL17RA
UniProt ID: (Human) Q96F46
Entrez Gene ID: (Human) 23765
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support