Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: AGIGFRHLPH WNTRCPLASH TDDSFTGSSA YIPCRTWWAL FSTKPWCVRV WHCSRCLCQH LLSGGSGLQR GLFHLLVQKS KKSSTFKFYR RHKMPAPAQR KLLPRRHLSE KSHHISIPSP DISHKGLRSK RTQPSDPETW ESLPRLDSQR HGGPEFSFDL LPEARAIRVT ISSGPEVSVR LCHQWALECE ELSSPYDVQK IVSGGHTVEL PYEFLLPCLC IEASYLQEDT VRRKKCP; Positive Samples: HT-29, A-549, A-431, Mouse kidney, Mouse thymus; Cellular Location: Cell membrane, Cytoplasm, Single-pass type I membrane protein
The interleukin 17 receptor E (IL-17RE) shares limited similarity to the receptor of IL-17A, a ubiquitous type I membrane glycoprotein that with IL-17A plays a pathogenic role in many inflammatory and autoimmune diseases such as rheumatoid arthritis. IL-17RE is the functional receptor of IL-17C and has been suggested to mediate mucosal immunity to infection with intestinal pathogens. IL-17C is produced by epithelia in response to bacterial challenge and binds to a receptor complex formed by IL-17RE and IL-17RA on the epithelial cell surface, thereby regulating the immune function of epithelial cells in an autocrine manner.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IL-17 receptor E; Interleukin-17 receptor E
Gene Aliases: AA589509; IL-17RE; IL17RE; Il25r; UNQ3056/PRO9877
UniProt ID: (Human) Q8NFR9, (Mouse) Q8BH06
Entrez Gene ID: (Human) 132014, (Mouse) 57890
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support